Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75234547-75235037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGA3
Entrez Gene ID
23163
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 3
Gene Aliases
-
UniGene
Hs.87726
Gene Chromosome Location
Chr.17: 75225464-75262393 on Build GRCh38
Transcripts
XM_011524563.2 NM_001303276.1 NM_024844.4 XM_005257692.3 XM_006722094.1 XM_011525267.1 XM_017025108.1 XM_017025109.1 XM_017025110.1

Gene Symbol
NUP85
Entrez Gene ID
79902
Gene Name
nucleoporin 85
Gene Aliases
FROUNT, Nup75
UniGene
Hs.362817
Gene Chromosome Location
Chr.17: 75205450-75235759 on Build GRCh38
Transcripts
XM_011524563.2 NM_001303276.1 NM_024844.4 XM_005257692.3 XM_006722094.1 XM_011525267.1 XM_017025108.1 XM_017025109.1 XM_017025110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6042077 c.1317_1318delTA p.S440fs*>23 Chr.17: 75234842-75234843 on Build GRCh38
6016819 c.1353C>T p.S451S Chr.17: 75234878-75234878 on Build GRCh38
4069515 c.1134T>C p.R378R Chr.17: 75234659-75234659 on Build GRCh38
6731694 c.1666G>A p.A556T Chr.17: 75234687-75234687 on Build GRCh38
6731695 c.1162G>A p.A388T Chr.17: 75234687-75234687 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767265411 Chr.17: 75234893-75234893 on Build GRCh38
rs760516189 Chr.17: 75234898-75234898 on Build GRCh38
rs573048590 Chr.17: 75234911-75234911 on Build GRCh38
rs570687318 Chr.17: 75234620-75234620 on Build GRCh38
rs930366904 Chr.17: 75234894-75234894 on Build GRCh38
rs750715149 Chr.17: 75234861-75234861 on Build GRCh38
rs1023302753 Chr.17: 75234574-75234574 on Build GRCh38
rs762504317 Chr.17: 75234827-75234827 on Build GRCh38
rs749309995 Chr.17: 75234648-75234648 on Build GRCh38
rs771144534 Chr.17: 75234657-75234657 on Build GRCh38
rs772983865 Chr.17: 75234822-75234822 on Build GRCh38
rs530510838 Chr.17: 75234583-75234583 on Build GRCh38
rs750471677 Chr.17: 75234900-75234900 on Build GRCh38
rs530782747 Chr.17: 75234865-75234865 on Build GRCh38
rs62086215 Chr.17: 75234630-75234630 on Build GRCh38
rs145404739 Chr.17: 75234721-75234721 on Build GRCh38
rs767461817 Chr.17: 75234840-75234840 on Build GRCh38
rs758185970 Chr.17: 75234957-75234958 on Build GRCh38
rs770210906 Chr.17: 75234915-75234915 on Build GRCh38
rs775161317 Chr.17: 75234941-75234941 on Build GRCh38
rs199756304 Chr.17: 75234684-75234684 on Build GRCh38
rs759180068 Chr.17: 75234934-75234934 on Build GRCh38
rs148227295 Chr.17: 75234783-75234783 on Build GRCh38
rs375906535 Chr.17: 75234818-75234818 on Build GRCh38
rs778369261 Chr.17: 75234611-75234611 on Build GRCh38
rs755007791 Chr.17: 75234979-75234979 on Build GRCh38
rs754925338 Chr.17: 75234722-75234722 on Build GRCh38
rs759765425 Chr.17: 75234699-75234701 on Build GRCh38
rs371329944 Chr.17: 75234809-75234809 on Build GRCh38
rs774028711 Chr.17: 75234932-75234932 on Build GRCh38
rs994606920 Chr.17: 75234746-75234746 on Build GRCh38
rs147921665 Chr.17: 75234646-75234646 on Build GRCh38
rs767861181 Chr.17: 75234778-75234783 on Build GRCh38
rs773941386 Chr.17: 75234890-75234890 on Build GRCh38
rs541797333 Chr.17: 75234942-75234942 on Build GRCh38
rs747318919 Chr.17: 75234779-75234779 on Build GRCh38
rs771912889 Chr.17: 75234937-75234937 on Build GRCh38
rs774871258 Chr.17: 75234629-75234629 on Build GRCh38
rs3214915 Chr.17: 75234842-75234843 on Build GRCh38
rs1021182261 Chr.17: 75234990-75234990 on Build GRCh38
rs752693082 Chr.17: 75234897-75234897 on Build GRCh38
rs753039026 Chr.17: 75234809-75234809 on Build GRCh38
rs2291032 Chr.17: 75234868-75234868 on Build GRCh38
rs766002365 Chr.17: 75234829-75234829 on Build GRCh38
rs772555152 Chr.17: 75234771-75234771 on Build GRCh38
rs200639832 Chr.17: 75234664-75234664 on Build GRCh38
rs771304601 Chr.17: 75234595-75234597 on Build GRCh38
rs931954785 Chr.17: 75234867-75234867 on Build GRCh38
rs760953241 Chr.17: 75234812-75234815 on Build GRCh38
rs757394827 Chr.17: 75234845-75234845 on Build GRCh38
rs9988 Chr.17: 75234761-75234761 on Build GRCh38
rs750025024 Chr.17: 75234702-75234702 on Build GRCh38
rs533381930 Chr.17: 75234681-75234681 on Build GRCh38
rs758258054 Chr.17: 75234716-75234716 on Build GRCh38
rs778306228 Chr.17: 75234917-75234917 on Build GRCh38
rs1034887197 Chr.17: 75234803-75234803 on Build GRCh38
rs533156111 Chr.17: 75234889-75234889 on Build GRCh38
rs550305037 Chr.17: 75234597-75234597 on Build GRCh38
rs112194253 Chr.17: 75234686-75234686 on Build GRCh38
rs751363224 Chr.17: 75234833-75234833 on Build GRCh38
rs1004758928 Chr.17: 75234725-75234725 on Build GRCh38
rs754730231 Chr.17: 75234836-75234836 on Build GRCh38
rs779241237 Chr.17: 75234740-75234740 on Build GRCh38
rs748599990 Chr.17: 75234869-75234869 on Build GRCh38
rs376654764 Chr.17: 75234815-75234815 on Build GRCh38
rs899038224 Chr.17: 75234758-75234758 on Build GRCh38
rs759372199 Chr.17: 75234891-75234891 on Build GRCh38
rs1045011615 Chr.17: 75234921-75234921 on Build GRCh38
rs778853695 Chr.17: 75234860-75234860 on Build GRCh38
rs750432533 Chr.17: 75234910-75234910 on Build GRCh38
rs755461892 Chr.17: 75234909-75234909 on Build GRCh38
rs752605043 Chr.17: 75234841-75234841 on Build GRCh38
rs780475181 Chr.17: 75234904-75234904 on Build GRCh38
rs112734534 Chr.17: 75235001-75235001 on Build GRCh38
rs539120556 Chr.17: 75234623-75234624 on Build GRCh38
rs763662878 Chr.17: 75234591-75234591 on Build GRCh38
rs369490158 Chr.17: 75234729-75234729 on Build GRCh38
rs761256079 Chr.17: 75234675-75234675 on Build GRCh38
rs746265050 Chr.17: 75234658-75234658 on Build GRCh38
rs770402782 Chr.17: 75234926-75234926 on Build GRCh38
rs545989200 Chr.17: 75234885-75234885 on Build GRCh38
rs367921124 Chr.17: 75234647-75234647 on Build GRCh38
rs750246813 Chr.17: 75234919-75234920 on Build GRCh38
rs776291455 Chr.17: 75234683-75234683 on Build GRCh38
rs769602381 Chr.17: 75234616-75234616 on Build GRCh38
rs1034768890 Chr.17: 75234844-75234844 on Build GRCh38
rs762460843 Chr.17: 75234884-75234884 on Build GRCh38
rs775703648 Chr.17: 75234774-75234774 on Build GRCh38
rs772615510 Chr.17: 75234881-75234881 on Build GRCh38
rs751802495 Chr.17: 75234908-75234908 on Build GRCh38
rs758560807 Chr.17: 75234863-75234863 on Build GRCh38
rs746923382 Chr.17: 75234645-75234645 on Build GRCh38
rs563354357 Chr.17: 75234786-75234786 on Build GRCh38
rs757232047 Chr.17: 75234733-75234733 on Build GRCh38
rs750266918 Chr.17: 75234618-75234618 on Build GRCh38
rs561720137 Chr.17: 75234980-75234980 on Build GRCh38
rs370744021 Chr.17: 75234598-75234598 on Build GRCh38
rs543886237 Chr.17: 75235006-75235006 on Build GRCh38
rs35857535 Chr.17: 75234701-75234701 on Build GRCh38
rs776099938 Chr.17: 75234665-75234665 on Build GRCh38
rs753793247 Chr.17: 75234959-75234959 on Build GRCh38
rs577099697 Chr.17: 75234871-75234871 on Build GRCh38
rs763968320 Chr.17: 75234943-75234943 on Build GRCh38
rs202239179 Chr.17: 75234720-75234720 on Build GRCh38
rs930542145 Chr.17: 75234916-75234916 on Build GRCh38
rs552915306 Chr.17: 75234899-75234899 on Build GRCh38
rs141865454 Chr.17: 75234688-75234688 on Build GRCh38
rs757239505 Chr.17: 75234868-75234869 on Build GRCh38
rs749399274 Chr.17: 75234732-75234732 on Build GRCh38
rs892178412 Chr.17: 75235000-75235000 on Build GRCh38
rs753608510 Chr.17: 75234593-75234593 on Build GRCh38

View Full Product Details