Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75913835-75914342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBF1
Entrez Gene ID
85302
Gene Name
Fas binding factor 1
Gene Aliases
Alb, FBF-1
UniGene
Hs.720068
Gene Chromosome Location
Chr.17: 75909574-75938232 on Build GRCh38
Transcripts
NM_001319193.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1206685 c.3032C>T p.A1011V Chr.17: 75913965-75913965 on Build GRCh38
3388146 c.3050A>T p.E1017V Chr.17: 75913947-75913947 on Build GRCh38
4069721 c.2898C>T p.T966T Chr.17: 75914170-75914170 on Build GRCh38
1206686 c.2986C>T p.R996C Chr.17: 75914011-75914011 on Build GRCh38
3388147 c.3049G>T p.E1017* Chr.17: 75913948-75913948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745639394 Chr.17: 75914132-75914132 on Build GRCh38
rs768113815 Chr.17: 75913938-75913938 on Build GRCh38
rs759793256 Chr.17: 75913866-75913866 on Build GRCh38
rs903948904 Chr.17: 75914203-75914203 on Build GRCh38
rs766378424 Chr.17: 75913876-75913876 on Build GRCh38
rs180748834 Chr.17: 75913961-75913961 on Build GRCh38
rs779470655 Chr.17: 75913897-75913897 on Build GRCh38
rs1021012703 Chr.17: 75914199-75914199 on Build GRCh38
rs758160836 Chr.17: 75913906-75913906 on Build GRCh38
rs888808677 Chr.17: 75914179-75914179 on Build GRCh38
rs905068295 Chr.17: 75914139-75914139 on Build GRCh38
rs918584693 Chr.17: 75913878-75913878 on Build GRCh38
rs758149843 Chr.17: 75914236-75914236 on Build GRCh38
rs773199429 Chr.17: 75914255-75914255 on Build GRCh38
rs369895761 Chr.17: 75914011-75914011 on Build GRCh38
rs757218381 Chr.17: 75914207-75914207 on Build GRCh38
rs752412883 Chr.17: 75914192-75914192 on Build GRCh38
rs370541289 Chr.17: 75914113-75914113 on Build GRCh38
rs373679795 Chr.17: 75914148-75914148 on Build GRCh38
rs779806594 Chr.17: 75913916-75913916 on Build GRCh38
rs377502076 Chr.17: 75914170-75914170 on Build GRCh38
rs374332434 Chr.17: 75914284-75914284 on Build GRCh38
rs55793071 Chr.17: 75913867-75913867 on Build GRCh38
rs770835758 Chr.17: 75914117-75914117 on Build GRCh38
rs778614457 Chr.17: 75914120-75914120 on Build GRCh38
rs3826274 Chr.17: 75914314-75914314 on Build GRCh38
rs770054332 Chr.17: 75914149-75914149 on Build GRCh38
rs147541272 Chr.17: 75914293-75914293 on Build GRCh38
rs553617516 Chr.17: 75914198-75914198 on Build GRCh38
rs746530564 Chr.17: 75913932-75913932 on Build GRCh38
rs944498850 Chr.17: 75913889-75913889 on Build GRCh38
rs765802801 Chr.17: 75913992-75913992 on Build GRCh38
rs909809159 Chr.17: 75913901-75913901 on Build GRCh38
rs199551553 Chr.17: 75914265-75914265 on Build GRCh38
rs573489562 Chr.17: 75914237-75914237 on Build GRCh38
rs777262131 Chr.17: 75914114-75914114 on Build GRCh38
rs750444987 Chr.17: 75914303-75914303 on Build GRCh38
rs776608208 Chr.17: 75914037-75914037 on Build GRCh38
rs768786129 Chr.17: 75914134-75914134 on Build GRCh38
rs759955938 Chr.17: 75914262-75914262 on Build GRCh38
rs759526959 Chr.17: 75914024-75914033 on Build GRCh38
rs767534166 Chr.17: 75914041-75914042 on Build GRCh38
rs369024350 Chr.17: 75913959-75913959 on Build GRCh38
rs761938623 Chr.17: 75914038-75914038 on Build GRCh38
rs754618020 Chr.17: 75914102-75914102 on Build GRCh38
rs774182959 Chr.17: 75914029-75914029 on Build GRCh38
rs780846474 Chr.17: 75914107-75914107 on Build GRCh38
rs776859185 Chr.17: 75914145-75914145 on Build GRCh38
rs568266767 Chr.17: 75914024-75914024 on Build GRCh38
rs777629604 Chr.17: 75914026-75914026 on Build GRCh38
rs542535100 Chr.17: 75914257-75914257 on Build GRCh38
rs998766057 Chr.17: 75914278-75914278 on Build GRCh38
rs765070344 Chr.17: 75913860-75913860 on Build GRCh38
rs144888266 Chr.17: 75914084-75914084 on Build GRCh38
rs61736662 Chr.17: 75913946-75913946 on Build GRCh38
rs754372856 Chr.17: 75913883-75913883 on Build GRCh38
rs766866106 Chr.17: 75914165-75914165 on Build GRCh38
rs937947618 Chr.17: 75914142-75914142 on Build GRCh38
rs747652849 Chr.17: 75914020-75914020 on Build GRCh38
rs781412045 Chr.17: 75914238-75914238 on Build GRCh38
rs1048950284 Chr.17: 75913907-75913907 on Build GRCh38
rs897689011 Chr.17: 75913892-75913892 on Build GRCh38
rs751223422 Chr.17: 75914016-75914016 on Build GRCh38
rs760947548 Chr.17: 75913975-75913975 on Build GRCh38
rs753933930 Chr.17: 75913988-75913988 on Build GRCh38
rs114455179 Chr.17: 75913890-75913890 on Build GRCh38
rs376833713 Chr.17: 75914010-75914010 on Build GRCh38
rs753316838 Chr.17: 75914204-75914204 on Build GRCh38
rs773400065 Chr.17: 75914150-75914150 on Build GRCh38
rs971864944 Chr.17: 75914079-75914079 on Build GRCh38
rs752794969 Chr.17: 75913868-75913868 on Build GRCh38
rs773453385 Chr.17: 75914250-75914250 on Build GRCh38
rs748082925 Chr.17: 75913944-75913944 on Build GRCh38
rs139333367 Chr.17: 75914310-75914310 on Build GRCh38
rs552443727 Chr.17: 75913870-75913870 on Build GRCh38
rs140747613 Chr.17: 75914060-75914060 on Build GRCh38
rs764140634 Chr.17: 75913979-75913979 on Build GRCh38
rs751033795 Chr.17: 75914056-75914056 on Build GRCh38
rs917060926 Chr.17: 75914112-75914112 on Build GRCh38
rs765856394 Chr.17: 75914048-75914048 on Build GRCh38
rs762269675 Chr.17: 75913989-75913989 on Build GRCh38
rs570414210 Chr.17: 75914109-75914109 on Build GRCh38
rs749281513 Chr.17: 75914115-75914115 on Build GRCh38
rs1026169521 Chr.17: 75914021-75914021 on Build GRCh38
rs368267518 Chr.17: 75914286-75914286 on Build GRCh38
rs775509470 Chr.17: 75913974-75913974 on Build GRCh38
rs776189544 Chr.17: 75913942-75913942 on Build GRCh38
rs1018549088 Chr.17: 75914272-75914272 on Build GRCh38
rs1021527020 Chr.17: 75913964-75913964 on Build GRCh38
rs745665757 Chr.17: 75914210-75914210 on Build GRCh38
rs1049378270 Chr.17: 75914129-75914129 on Build GRCh38
rs765095267 Chr.17: 75914302-75914302 on Build GRCh38
rs749085238 Chr.17: 75914027-75914027 on Build GRCh38
rs775810816 Chr.17: 75914268-75914268 on Build GRCh38
rs1040374974 Chr.17: 75914263-75914263 on Build GRCh38
rs774854401 Chr.17: 75914260-75914260 on Build GRCh38
rs771752676 Chr.17: 75914241-75914241 on Build GRCh38
rs772965713 Chr.17: 75913865-75913865 on Build GRCh38
rs759885597 Chr.17: 75914176-75914176 on Build GRCh38
rs377618256 Chr.17: 75914032-75914032 on Build GRCh38
rs773020817 Chr.17: 75913949-75913949 on Build GRCh38
rs763536761 Chr.17: 75914058-75914058 on Build GRCh38
rs991392931 Chr.17: 75914090-75914090 on Build GRCh38
rs780604600 Chr.17: 75914019-75914019 on Build GRCh38
rs369262043 Chr.17: 75914033-75914033 on Build GRCh38
rs754633811 Chr.17: 75914018-75914018 on Build GRCh38
rs764669454 Chr.17: 75913872-75913872 on Build GRCh38
rs935770930 Chr.17: 75914146-75914146 on Build GRCh38
rs749728329 Chr.17: 75913939-75913939 on Build GRCh38
rs778864232 Chr.17: 75914208-75914208 on Build GRCh38
rs113839911 Chr.17: 75913928-75913928 on Build GRCh38
rs751715906 Chr.17: 75913892-75913895 on Build GRCh38
rs746207344 Chr.17: 75913900-75913900 on Build GRCh38
rs373656567 Chr.17: 75914014-75914014 on Build GRCh38
rs752662481 Chr.17: 75914140-75914144 on Build GRCh38
rs985772589 Chr.17: 75913859-75913859 on Build GRCh38
rs758243151 Chr.17: 75914309-75914309 on Build GRCh38
rs200283832 Chr.17: 75914163-75914163 on Build GRCh38
rs767081032 Chr.17: 75914182-75914182 on Build GRCh38
rs544253818 Chr.17: 75914299-75914299 on Build GRCh38

View Full Product Details