Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 47155270-47155803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IER3IP1
Entrez Gene ID
51124
Gene Name
immediate early response 3 interacting protein 1
Gene Aliases
HSPC039, MEDS, PRO2309
UniGene
Hs.130917
Gene Chromosome Location
Chr.18: 47155019-47176374 on Build GRCh38
Transcripts
NM_016097.4

SNP Detail

SNP ID Chromosome Location
rs191595406 Chr.18: 47155683-47155683 on Build GRCh38
rs551870994 Chr.18: 47155744-47155744 on Build GRCh38
rs769608786 Chr.18: 47155461-47155461 on Build GRCh38
rs915151422 Chr.18: 47155471-47155471 on Build GRCh38
rs528591728 Chr.18: 47155674-47155677 on Build GRCh38
rs898736483 Chr.18: 47155627-47155627 on Build GRCh38
rs187175176 Chr.18: 47155372-47155372 on Build GRCh38
rs774159168 Chr.18: 47155738-47155738 on Build GRCh38
rs1013609058 Chr.18: 47155630-47155630 on Build GRCh38
rs887232992 Chr.18: 47155547-47155547 on Build GRCh38
rs764415950 Chr.18: 47155581-47155581 on Build GRCh38
rs35726685 Chr.18: 47155594-47155595 on Build GRCh38
rs750179892 Chr.18: 47155583-47155583 on Build GRCh38
rs542820094 Chr.18: 47155540-47155542 on Build GRCh38
rs529253691 Chr.18: 47155426-47155426 on Build GRCh38
rs929345347 Chr.18: 47155538-47155538 on Build GRCh38
rs956158599 Chr.18: 47155714-47155714 on Build GRCh38
rs1055062860 Chr.18: 47155368-47155368 on Build GRCh38
rs540392061 Chr.18: 47155354-47155354 on Build GRCh38
rs368065227 Chr.18: 47155479-47155480 on Build GRCh38
rs1050838268 Chr.18: 47155545-47155545 on Build GRCh38
rs141766622 Chr.18: 47155377-47155377 on Build GRCh38
rs1046615648 Chr.18: 47155745-47155745 on Build GRCh38
rs902225627 Chr.18: 47155422-47155422 on Build GRCh38
rs745352280 Chr.18: 47155451-47155451 on Build GRCh38
rs1031944995 Chr.18: 47155429-47155431 on Build GRCh38
rs902362830 Chr.18: 47155755-47155755 on Build GRCh38
rs1038579934 Chr.18: 47155611-47155611 on Build GRCh38
rs1055043415 Chr.18: 47155667-47155667 on Build GRCh38
rs946656404 Chr.18: 47155506-47155506 on Build GRCh38
rs976691111 Chr.18: 47155488-47155488 on Build GRCh38
rs933727953 Chr.18: 47155531-47155531 on Build GRCh38
rs941904223 Chr.18: 47155356-47155356 on Build GRCh38
rs201115046 Chr.18: 47155433-47155434 on Build GRCh38
rs548996218 Chr.18: 47155514-47155514 on Build GRCh38
rs1002921901 Chr.18: 47155664-47155664 on Build GRCh38
rs1013874799 Chr.18: 47155374-47155374 on Build GRCh38
rs16949034 Chr.18: 47155652-47155652 on Build GRCh38
rs907206677 Chr.18: 47155663-47155663 on Build GRCh38
rs940166722 Chr.18: 47155591-47155591 on Build GRCh38
rs200246920 Chr.18: 47155468-47155469 on Build GRCh38
rs887782500 Chr.18: 47155320-47155320 on Build GRCh38
rs573836137 Chr.18: 47155519-47155519 on Build GRCh38
rs775239916 Chr.18: 47155516-47155516 on Build GRCh38
rs200570860 Chr.18: 47155450-47155452 on Build GRCh38
rs148825959 Chr.18: 47155444-47155446 on Build GRCh38

View Full Product Details