Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 49483364-49483866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C18orf32
Entrez Gene ID
497661
Gene Name
chromosome 18 open reading frame 32
Gene Aliases
-
UniGene
Hs.603040
Gene Chromosome Location
Chr.18: 49481178-49487274 on Build GRCh38
Transcripts
NM_001035005.3 NM_001199346.1 NM_001199355.1 NM_001199356.1

Gene Symbol
RPL17-C18orf32
Entrez Gene ID
100526842
Gene Name
RPL17-C18orf32 readthrough
Gene Aliases
-
UniGene
Hs.374588
Gene Chromosome Location
Chr.18: 49481178-49492565 on Build GRCh38
Transcripts
NM_001035005.3 NM_001199346.1 NM_001199355.1 NM_001199356.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6769178 c.93C>T p.F31F Chr.18: 49483656-49483656 on Build GRCh38
709430 c.66A>G p.P22P Chr.18: 49483683-49483683 on Build GRCh38
6148545 c.89C>T p.P30L Chr.18: 49483660-49483660 on Build GRCh38
6614806 c.22G>A p.V8I Chr.18: 49483727-49483727 on Build GRCh38
1198287 c.127G>T p.E43* Chr.18: 49483622-49483622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777139159 Chr.18: 49483739-49483739 on Build GRCh38
rs768753352 Chr.18: 49483642-49483642 on Build GRCh38
rs140347847 Chr.18: 49483593-49483593 on Build GRCh38
rs191869034 Chr.18: 49483616-49483616 on Build GRCh38
rs1021124345 Chr.18: 49483502-49483502 on Build GRCh38
rs760295150 Chr.18: 49483575-49483575 on Build GRCh38
rs751544895 Chr.18: 49483680-49483680 on Build GRCh38
rs780243305 Chr.18: 49483699-49483699 on Build GRCh38
rs1008043226 Chr.18: 49483425-49483425 on Build GRCh38
rs766028505 Chr.18: 49483655-49483655 on Build GRCh38
rs769307126 Chr.18: 49483757-49483757 on Build GRCh38
rs1011122324 Chr.18: 49483469-49483469 on Build GRCh38
rs747588792 Chr.18: 49483727-49483727 on Build GRCh38
rs576383326 Chr.18: 49483625-49483625 on Build GRCh38
rs758131734 Chr.18: 49483606-49483606 on Build GRCh38
rs775042329 Chr.18: 49483679-49483679 on Build GRCh38
rs762567484 Chr.18: 49483768-49483768 on Build GRCh38
rs375259579 Chr.18: 49483825-49483825 on Build GRCh38
rs761794204 Chr.18: 49483791-49483791 on Build GRCh38
rs751348939 Chr.18: 49483779-49483779 on Build GRCh38
rs376746396 Chr.18: 49483604-49483604 on Build GRCh38
rs748469635 Chr.18: 49483646-49483646 on Build GRCh38
rs748046543 Chr.18: 49483790-49483790 on Build GRCh38
rs764810508 Chr.18: 49483661-49483661 on Build GRCh38
rs373778829 Chr.18: 49483724-49483724 on Build GRCh38
rs369428991 Chr.18: 49483764-49483764 on Build GRCh38
rs972646924 Chr.18: 49483667-49483667 on Build GRCh38
rs768155639 Chr.18: 49483549-49483549 on Build GRCh38
rs769110907 Chr.18: 49483541-49483541 on Build GRCh38
rs80201970 Chr.18: 49483728-49483728 on Build GRCh38
rs11537626 Chr.18: 49483639-49483639 on Build GRCh38
rs372463077 Chr.18: 49483557-49483557 on Build GRCh38
rs918546080 Chr.18: 49483682-49483682 on Build GRCh38
rs768637811 Chr.18: 49483715-49483718 on Build GRCh38
rs779031886 Chr.18: 49483807-49483807 on Build GRCh38
rs138012802 Chr.18: 49483647-49483647 on Build GRCh38
rs757152863 Chr.18: 49483685-49483685 on Build GRCh38
rs1033430767 Chr.18: 49483530-49483530 on Build GRCh38
rs572227147 Chr.18: 49483656-49483656 on Build GRCh38
rs762446233 Chr.18: 49483544-49483544 on Build GRCh38
rs746907370 Chr.18: 49483617-49483617 on Build GRCh38
rs372217659 Chr.18: 49483648-49483648 on Build GRCh38
rs750338395 Chr.18: 49483798-49483798 on Build GRCh38
rs749561223 Chr.18: 49483640-49483640 on Build GRCh38
rs749060876 Chr.18: 49483674-49483675 on Build GRCh38
rs202115703 Chr.18: 49483710-49483710 on Build GRCh38
rs959774832 Chr.18: 49483820-49483820 on Build GRCh38
rs1041613633 Chr.18: 49483435-49483435 on Build GRCh38
rs769797410 Chr.18: 49483533-49483533 on Build GRCh38
rs555008646 Chr.18: 49483404-49483404 on Build GRCh38
rs1041952354 Chr.18: 49483788-49483788 on Build GRCh38
rs377011626 Chr.18: 49483752-49483752 on Build GRCh38
rs774331787 Chr.18: 49483748-49483748 on Build GRCh38
rs753578142 Chr.18: 49483588-49483588 on Build GRCh38
rs143746104 Chr.18: 49483722-49483722 on Build GRCh38
rs765146925 Chr.18: 49483602-49483602 on Build GRCh38
rs767363331 Chr.18: 49483796-49483796 on Build GRCh38
rs895640224 Chr.18: 49483393-49483393 on Build GRCh38
rs201224790 Chr.18: 49483649-49483649 on Build GRCh38
rs748334926 Chr.18: 49483715-49483715 on Build GRCh38
rs574841089 Chr.18: 49483459-49483459 on Build GRCh38
rs867268383 Chr.18: 49483693-49483694 on Build GRCh38
rs749418926 Chr.18: 49483539-49483539 on Build GRCh38
rs752328241 Chr.18: 49483665-49483665 on Build GRCh38
rs758219583 Chr.18: 49483672-49483672 on Build GRCh38
rs759291520 Chr.18: 49483659-49483659 on Build GRCh38
rs561008485 Chr.18: 49483756-49483756 on Build GRCh38
rs1000167975 Chr.18: 49483504-49483504 on Build GRCh38
rs774544285 Chr.18: 49483542-49483542 on Build GRCh38
rs375834505 Chr.18: 49483704-49483704 on Build GRCh38
rs1035210902 Chr.18: 49483794-49483794 on Build GRCh38
rs1007634820 Chr.18: 49483600-49483600 on Build GRCh38
rs775719203 Chr.18: 49483535-49483535 on Build GRCh38
rs369128238 Chr.18: 49483626-49483626 on Build GRCh38
rs777690741 Chr.18: 49483615-49483615 on Build GRCh38
rs755040185 Chr.18: 49483799-49483799 on Build GRCh38
rs1014066507 Chr.18: 49483426-49483426 on Build GRCh38
rs369030829 Chr.18: 49483706-49483706 on Build GRCh38
rs991181511 Chr.18: 49483758-49483758 on Build GRCh38
rs201004082 Chr.18: 49483644-49483644 on Build GRCh38
rs200854465 Chr.18: 49483774-49483774 on Build GRCh38
rs765941547 Chr.18: 49483587-49483587 on Build GRCh38

View Full Product Details