Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 54365529-54365949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C18orf54
Entrez Gene ID
162681
Gene Name
chromosome 18 open reading frame 54
Gene Aliases
LAS2
UniGene
Hs.208701
Gene Chromosome Location
Chr.18: 54357914-54382035 on Build GRCh38
Transcripts
NM_001288980.1 NM_001288981.1 NM_001288982.1 NM_173529.5 NR_110234.1 XM_005258201.1 XM_005258203.1 XM_017025567.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
357270 c.843+1G>A p.? Chr.18: 54365822-54365822 on Build GRCh38
5790621 c.800G>A p.S267N Chr.18: 54365778-54365778 on Build GRCh38
6614811 c.757G>A p.V253I Chr.18: 54365735-54365735 on Build GRCh38
563722 c.756C>A p.P252P Chr.18: 54365734-54365734 on Build GRCh38
386893 c.757G>T p.V253F Chr.18: 54365735-54365735 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746558887 Chr.18: 54365832-54365832 on Build GRCh38
rs771678339 Chr.18: 54365813-54365813 on Build GRCh38
rs959087273 Chr.18: 54365596-54365596 on Build GRCh38
rs779148516 Chr.18: 54365800-54365800 on Build GRCh38
rs564715637 Chr.18: 54365788-54365788 on Build GRCh38
rs748500557 Chr.18: 54365621-54365621 on Build GRCh38
rs576628455 Chr.18: 54365563-54365563 on Build GRCh38
rs150002374 Chr.18: 54365733-54365733 on Build GRCh38
rs761384515 Chr.18: 54365698-54365698 on Build GRCh38
rs987593657 Chr.18: 54365699-54365699 on Build GRCh38
rs768295349 Chr.18: 54365834-54365835 on Build GRCh38
rs370519789 Chr.18: 54365729-54365729 on Build GRCh38
rs962644425 Chr.18: 54365625-54365625 on Build GRCh38
rs765147351 Chr.18: 54365735-54365735 on Build GRCh38
rs773878976 Chr.18: 54365865-54365867 on Build GRCh38
rs1029978744 Chr.18: 54365609-54365609 on Build GRCh38
rs867197636 Chr.18: 54365874-54365874 on Build GRCh38
rs775108995 Chr.18: 54365724-54365726 on Build GRCh38
rs116395327 Chr.18: 54365889-54365889 on Build GRCh38
rs748874671 Chr.18: 54365827-54365828 on Build GRCh38
rs749689818 Chr.18: 54365669-54365669 on Build GRCh38
rs781330301 Chr.18: 54365745-54365745 on Build GRCh38
rs756579110 Chr.18: 54365749-54365749 on Build GRCh38
rs760646263 Chr.18: 54365727-54365727 on Build GRCh38
rs751750581 Chr.18: 54365737-54365737 on Build GRCh38
rs1042566467 Chr.18: 54365785-54365785 on Build GRCh38
rs994277270 Chr.18: 54365559-54365559 on Build GRCh38
rs376096474 Chr.18: 54365692-54365692 on Build GRCh38
rs13380956 Chr.18: 54365587-54365587 on Build GRCh38
rs946972654 Chr.18: 54365763-54365763 on Build GRCh38
rs747832140 Chr.18: 54365807-54365807 on Build GRCh38
rs113166902 Chr.18: 54365858-54365858 on Build GRCh38
rs772755551 Chr.18: 54365826-54365826 on Build GRCh38
rs542230935 Chr.18: 54365635-54365635 on Build GRCh38
rs929355351 Chr.18: 54365696-54365696 on Build GRCh38
rs772662781 Chr.18: 54365721-54365721 on Build GRCh38
rs374556847 Chr.18: 54365734-54365734 on Build GRCh38
rs9946193 Chr.18: 54365594-54365594 on Build GRCh38
rs749814296 Chr.18: 54365787-54365787 on Build GRCh38
rs531931422 Chr.18: 54365861-54365861 on Build GRCh38
rs370707130 Chr.18: 54365677-54365677 on Build GRCh38
rs757429277 Chr.18: 54365742-54365742 on Build GRCh38
rs771544033 Chr.18: 54365718-54365718 on Build GRCh38
rs894058111 Chr.18: 54365758-54365758 on Build GRCh38
rs1034777426 Chr.18: 54365910-54365910 on Build GRCh38
rs2339994 Chr.18: 54365645-54365645 on Build GRCh38
rs948322608 Chr.18: 54365731-54365731 on Build GRCh38
rs377493692 Chr.18: 54365668-54365668 on Build GRCh38
rs780495544 Chr.18: 54365778-54365778 on Build GRCh38
rs770598273 Chr.18: 54365847-54365847 on Build GRCh38
rs3760543 Chr.18: 54365859-54365859 on Build GRCh38
rs1047165262 Chr.18: 54365566-54365566 on Build GRCh38
rs73958766 Chr.18: 54365746-54365746 on Build GRCh38
rs187867814 Chr.18: 54365607-54365607 on Build GRCh38
rs769036574 Chr.18: 54365682-54365682 on Build GRCh38

View Full Product Details