Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7440396-7440906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF18
Entrez Gene ID
23370
Gene Name
Rho/Rac guanine nucleotide exchange factor 18
Gene Aliases
P114-RhoGEF
UniGene
Hs.465761
Gene Chromosome Location
Chr.19: 7348901-7476990 on Build GRCh38
Transcripts
NM_001130955.1 NM_015318.3 XM_005272464.4 XM_006722705.3 XM_006722706.3 XM_006722708.2 XM_011527835.2 XM_011527836.2 XM_011527837.2 XM_011527838.2 XM_011527839.2 XM_011527840.1 XM_011527841.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7177654 c.487C>G p.Q163E Chr.19: 7440427-7440427 on Build GRCh38
3541228 c.31C>T p.P11S Chr.19: 7440445-7440445 on Build GRCh38
6604102 c.19delG p.G8fs*30 Chr.19: 7440433-7440433 on Build GRCh38
4082722 c.497G>T p.G166V Chr.19: 7440437-7440437 on Build GRCh38
6225725 c.44C>T p.P15L Chr.19: 7440458-7440458 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs948191707 Chr.19: 7440672-7440672 on Build GRCh38
rs942156905 Chr.19: 7440699-7440699 on Build GRCh38
rs950927609 Chr.19: 7440670-7440670 on Build GRCh38
rs908143746 Chr.19: 7440510-7440510 on Build GRCh38
rs952433671 Chr.19: 7440870-7440870 on Build GRCh38
rs547057092 Chr.19: 7440509-7440509 on Build GRCh38
rs940940232 Chr.19: 7440564-7440564 on Build GRCh38
rs369247436 Chr.19: 7440550-7440550 on Build GRCh38
rs921965335 Chr.19: 7440772-7440772 on Build GRCh38
rs984959658 Chr.19: 7440880-7440880 on Build GRCh38
rs768732615 Chr.19: 7440511-7440511 on Build GRCh38
rs76993643 Chr.19: 7440659-7440659 on Build GRCh38
rs933298763 Chr.19: 7440775-7440775 on Build GRCh38
rs200633564 Chr.19: 7440459-7440459 on Build GRCh38
rs552616985 Chr.19: 7440803-7440803 on Build GRCh38
rs903899458 Chr.19: 7440685-7440685 on Build GRCh38
rs1021383766 Chr.19: 7440554-7440554 on Build GRCh38
rs879519515 Chr.19: 7440835-7440835 on Build GRCh38
rs992448778 Chr.19: 7440647-7440647 on Build GRCh38
rs781292482 Chr.19: 7440498-7440498 on Build GRCh38
rs959684093 Chr.19: 7440591-7440591 on Build GRCh38
rs892426211 Chr.19: 7440723-7440723 on Build GRCh38
rs115555106 Chr.19: 7440481-7440481 on Build GRCh38
rs1034050270 Chr.19: 7440588-7440588 on Build GRCh38
rs757729503 Chr.19: 7440497-7440497 on Build GRCh38
rs35114657 Chr.19: 7440432-7440432 on Build GRCh38
rs772177440 Chr.19: 7440506-7440506 on Build GRCh38
rs780255449 Chr.19: 7440507-7440507 on Build GRCh38
rs188358798 Chr.19: 7440805-7440805 on Build GRCh38
rs371300946 Chr.19: 7440515-7440515 on Build GRCh38
rs1051703348 Chr.19: 7440791-7440791 on Build GRCh38
rs956575348 Chr.19: 7440423-7440423 on Build GRCh38
rs762758574 Chr.19: 7440422-7440422 on Build GRCh38
rs1038970699 Chr.19: 7440461-7440461 on Build GRCh38
rs34009822 Chr.19: 7440601-7440602 on Build GRCh38
rs774116610 Chr.19: 7440534-7440534 on Build GRCh38
rs983989577 Chr.19: 7440677-7440677 on Build GRCh38
rs945904909 Chr.19: 7440794-7440794 on Build GRCh38
rs570414418 Chr.19: 7440508-7440508 on Build GRCh38
rs11878820 Chr.19: 7440573-7440573 on Build GRCh38
rs913108381 Chr.19: 7440792-7440792 on Build GRCh38
rs930829545 Chr.19: 7440493-7440493 on Build GRCh38
rs1030829934 Chr.19: 7440712-7440712 on Build GRCh38
rs773143261 Chr.19: 7440419-7440419 on Build GRCh38
rs1043321471 Chr.19: 7440802-7440802 on Build GRCh38
rs762842388 Chr.19: 7440530-7440530 on Build GRCh38
rs751248837 Chr.19: 7440452-7440452 on Build GRCh38
rs1055536574 Chr.19: 7440819-7440819 on Build GRCh38
rs754601209 Chr.19: 7440453-7440453 on Build GRCh38
rs754348870 Chr.19: 7440482-7440482 on Build GRCh38
rs1019303850 Chr.19: 7440763-7440763 on Build GRCh38
rs28489511 Chr.19: 7440446-7440446 on Build GRCh38
rs904234515 Chr.19: 7440561-7440561 on Build GRCh38
rs895503608 Chr.19: 7440881-7440881 on Build GRCh38
rs538977209 Chr.19: 7440751-7440751 on Build GRCh38
rs891648796 Chr.19: 7440513-7440513 on Build GRCh38
rs770758435 Chr.19: 7440531-7440531 on Build GRCh38
rs779391912 Chr.19: 7440500-7440500 on Build GRCh38
rs1019695738 Chr.19: 7440451-7440451 on Build GRCh38
rs752147981 Chr.19: 7440478-7440478 on Build GRCh38
rs376793767 Chr.19: 7440502-7440502 on Build GRCh38
rs1030926113 Chr.19: 7440422-7440423 on Build GRCh38
rs1025626122 Chr.19: 7440665-7440665 on Build GRCh38
rs1001282524 Chr.19: 7440567-7440567 on Build GRCh38
rs2287907 Chr.19: 7440680-7440680 on Build GRCh38

View Full Product Details