Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7698650-7699149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCER2
Entrez Gene ID
2208
Gene Name
Fc fragment of IgE receptor II
Gene Aliases
BLAST-2, CD23, CD23A, CLEC4J, FCE2, IGEBF
UniGene
Hs.465778
Gene Chromosome Location
Chr.19: 7688757-7702755 on Build GRCh38
Transcripts
NM_001207019.2 NM_001220500.1 NM_002002.4 XM_005272462.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1003350 c.24G>T p.E8D Chr.19: 7698853-7698853 on Build GRCh38
4774494 c.72C>T p.I24I Chr.19: 7698805-7698805 on Build GRCh38
3721804 c.60T>C p.R20R Chr.19: 7698817-7698817 on Build GRCh38
3541422 c.105G>A p.W35* Chr.19: 7698772-7698772 on Build GRCh38
5391146 c.27C>T p.I9I Chr.19: 7698850-7698850 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs988959509 Chr.19: 7698687-7698687 on Build GRCh38
rs933406092 Chr.19: 7698710-7698710 on Build GRCh38
rs537287800 Chr.19: 7698774-7698774 on Build GRCh38
rs62111765 Chr.19: 7698985-7698985 on Build GRCh38
rs762556859 Chr.19: 7698773-7698773 on Build GRCh38
rs150243563 Chr.19: 7698776-7698776 on Build GRCh38
rs2287866 Chr.19: 7698817-7698817 on Build GRCh38
rs754772969 Chr.19: 7698720-7698720 on Build GRCh38
rs755966040 Chr.19: 7698854-7698854 on Build GRCh38
rs369367063 Chr.19: 7698758-7698758 on Build GRCh38
rs145650191 Chr.19: 7698740-7698740 on Build GRCh38
rs866807750 Chr.19: 7699034-7699034 on Build GRCh38
rs185214885 Chr.19: 7698781-7698781 on Build GRCh38
rs897989889 Chr.19: 7698912-7698912 on Build GRCh38
rs143388680 Chr.19: 7698850-7698850 on Build GRCh38
rs12981234 Chr.19: 7698992-7698992 on Build GRCh38
rs1037838433 Chr.19: 7698900-7698900 on Build GRCh38
rs369730396 Chr.19: 7698719-7698719 on Build GRCh38
rs138454013 Chr.19: 7698747-7698747 on Build GRCh38
rs138026836 Chr.19: 7698849-7698849 on Build GRCh38
rs769978800 Chr.19: 7698884-7698884 on Build GRCh38
rs765935864 Chr.19: 7698830-7698830 on Build GRCh38
rs71179126 Chr.19: 7698812-7698813 on Build GRCh38
rs8110107 Chr.19: 7699044-7699044 on Build GRCh38
rs138917660 Chr.19: 7698783-7698783 on Build GRCh38
rs539091249 Chr.19: 7698913-7698913 on Build GRCh38
rs376326260 Chr.19: 7698784-7698784 on Build GRCh38
rs546158506 Chr.19: 7698670-7698670 on Build GRCh38
rs149266114 Chr.19: 7698804-7698804 on Build GRCh38
rs748353352 Chr.19: 7698986-7698986 on Build GRCh38
rs75301084 Chr.19: 7698840-7698840 on Build GRCh38
rs745892615 Chr.19: 7698785-7698785 on Build GRCh38
rs746078341 Chr.19: 7698741-7698741 on Build GRCh38
rs8110128 Chr.19: 7699091-7699091 on Build GRCh38
rs758552077 Chr.19: 7698794-7698794 on Build GRCh38
rs562872186 Chr.19: 7698964-7698964 on Build GRCh38
rs375723520 Chr.19: 7698860-7698860 on Build GRCh38
rs72558006 Chr.19: 7698831-7698831 on Build GRCh38
rs757356321 Chr.19: 7698731-7698731 on Build GRCh38
rs193197320 Chr.19: 7698767-7698767 on Build GRCh38
rs903569009 Chr.19: 7698991-7698992 on Build GRCh38
rs923214934 Chr.19: 7699106-7699106 on Build GRCh38
rs532036972 Chr.19: 7698735-7698735 on Build GRCh38
rs751798756 Chr.19: 7698870-7698870 on Build GRCh38
rs141436817 Chr.19: 7698757-7698757 on Build GRCh38
rs986066765 Chr.19: 7699111-7699111 on Build GRCh38
rs989957518 Chr.19: 7698799-7698799 on Build GRCh38
rs764771051 Chr.19: 7698759-7698759 on Build GRCh38
rs764710531 Chr.19: 7698700-7698700 on Build GRCh38
rs748145712 Chr.19: 7698728-7698728 on Build GRCh38
rs933258688 Chr.19: 7699109-7699109 on Build GRCh38
rs763868178 Chr.19: 7698857-7698857 on Build GRCh38
rs936395833 Chr.19: 7698991-7698991 on Build GRCh38
rs775882255 Chr.19: 7698745-7698745 on Build GRCh38
rs780346824 Chr.19: 7698861-7698861 on Build GRCh38
rs752893524 Chr.19: 7698801-7698801 on Build GRCh38
rs376204616 Chr.19: 7698896-7698896 on Build GRCh38
rs752383168 Chr.19: 7698852-7698852 on Build GRCh38
rs551858136 Chr.19: 7698744-7698744 on Build GRCh38
rs10415518 Chr.19: 7699031-7699031 on Build GRCh38
rs369921807 Chr.19: 7698999-7698999 on Build GRCh38
rs139634066 Chr.19: 7698929-7698929 on Build GRCh38
rs770161837 Chr.19: 7698818-7698818 on Build GRCh38
rs781053482 Chr.19: 7698725-7698725 on Build GRCh38
rs993761913 Chr.19: 7698914-7698914 on Build GRCh38
rs748395997 Chr.19: 7698903-7698903 on Build GRCh38
rs906852375 Chr.19: 7698948-7698948 on Build GRCh38
rs376301496 Chr.19: 7698706-7698706 on Build GRCh38
rs992469435 Chr.19: 7698941-7698941 on Build GRCh38
rs754758835 Chr.19: 7698780-7698780 on Build GRCh38
rs773775018 Chr.19: 7698819-7698819 on Build GRCh38
rs755298422 Chr.19: 7698873-7698873 on Build GRCh38
rs769136391 Chr.19: 7698805-7698805 on Build GRCh38
rs971090665 Chr.19: 7698775-7698775 on Build GRCh38
rs559731890 Chr.19: 7698713-7698713 on Build GRCh38
rs72558013 Chr.19: 7699097-7699097 on Build GRCh38
rs544103337 Chr.19: 7699014-7699014 on Build GRCh38
rs777173432 Chr.19: 7698812-7698812 on Build GRCh38

View Full Product Details