Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7699589-7700080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCER2
Entrez Gene ID
2208
Gene Name
Fc fragment of IgE receptor II
Gene Aliases
BLAST-2, CD23, CD23A, CLEC4J, FCE2, IGEBF
UniGene
Hs.465778
Gene Chromosome Location
Chr.19: 7688757-7702755 on Build GRCh38
Transcripts
NM_001220500.1 NM_002002.4 XM_005272462.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6153396 c.21A>C p.S7S Chr.19: 7699740-7699740 on Build GRCh38
3423357 c.13C>A p.Q5K Chr.19: 7699748-7699748 on Build GRCh38
6477299 c.22+1G>T p.? Chr.19: 7699738-7699738 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs990958360 Chr.19: 7699936-7699936 on Build GRCh38
rs765852287 Chr.19: 7699942-7699942 on Build GRCh38
rs149217740 Chr.19: 7699812-7699812 on Build GRCh38
rs879708108 Chr.19: 7700027-7700027 on Build GRCh38
rs11672997 Chr.19: 7699979-7699979 on Build GRCh38
rs934356682 Chr.19: 7699981-7699981 on Build GRCh38
rs376251026 Chr.19: 7699811-7699811 on Build GRCh38
rs201405965 Chr.19: 7699991-7699992 on Build GRCh38
rs764006533 Chr.19: 7699690-7699690 on Build GRCh38
rs758395297 Chr.19: 7699743-7699744 on Build GRCh38
rs201171143 Chr.19: 7699703-7699703 on Build GRCh38
rs531275645 Chr.19: 7699677-7699677 on Build GRCh38
rs972337627 Chr.19: 7699978-7699978 on Build GRCh38
rs778064818 Chr.19: 7700040-7700040 on Build GRCh38
rs768304041 Chr.19: 7699761-7699761 on Build GRCh38
rs1030080591 Chr.19: 7699676-7699676 on Build GRCh38
rs928111720 Chr.19: 7699756-7699756 on Build GRCh38
rs562527082 Chr.19: 7699659-7699659 on Build GRCh38
rs184734088 Chr.19: 7700019-7700019 on Build GRCh38
rs759675233 Chr.19: 7699783-7699783 on Build GRCh38
rs894136483 Chr.19: 7699624-7699624 on Build GRCh38
rs3214456 Chr.19: 7699659-7699660 on Build GRCh38
rs11672993 Chr.19: 7699924-7699924 on Build GRCh38
rs142479170 Chr.19: 7699941-7699941 on Build GRCh38
rs112871344 Chr.19: 7699991-7699991 on Build GRCh38
rs397698363 Chr.19: 7699662-7699663 on Build GRCh38
rs200865107 Chr.19: 7699793-7699793 on Build GRCh38
rs569462141 Chr.19: 7699610-7699610 on Build GRCh38
rs764247173 Chr.19: 7699788-7699788 on Build GRCh38
rs766543517 Chr.19: 7699704-7699704 on Build GRCh38
rs771381661 Chr.19: 7699739-7699739 on Build GRCh38
rs2287868 Chr.19: 7699886-7699886 on Build GRCh38
rs959462132 Chr.19: 7699935-7699935 on Build GRCh38
rs368902365 Chr.19: 7699764-7699764 on Build GRCh38
rs1011682356 Chr.19: 7699628-7699628 on Build GRCh38
rs749260823 Chr.19: 7699749-7699749 on Build GRCh38
rs2287867 Chr.19: 7699855-7699855 on Build GRCh38
rs112897528 Chr.19: 7699878-7699878 on Build GRCh38
rs865915044 Chr.19: 7700029-7700029 on Build GRCh38
rs113802533 Chr.19: 7699988-7699988 on Build GRCh38
rs781214296 Chr.19: 7699713-7699713 on Build GRCh38
rs867120374 Chr.19: 7699911-7699911 on Build GRCh38
rs753715211 Chr.19: 7699696-7699696 on Build GRCh38
rs561853433 Chr.19: 7699955-7699955 on Build GRCh38
rs765200662 Chr.19: 7699770-7699770 on Build GRCh38
rs754165362 Chr.19: 7699790-7699790 on Build GRCh38
rs533773452 Chr.19: 7699899-7699899 on Build GRCh38
rs72558012 Chr.19: 7699843-7699843 on Build GRCh38
rs909535984 Chr.19: 7699961-7699961 on Build GRCh38
rs113327021 Chr.19: 7699988-7699988 on Build GRCh38
rs995156350 Chr.19: 7700032-7700032 on Build GRCh38
rs371068137 Chr.19: 7699784-7699784 on Build GRCh38
rs72558008 Chr.19: 7699905-7699905 on Build GRCh38
rs538486964 Chr.19: 7699632-7699632 on Build GRCh38
rs757838614 Chr.19: 7699805-7699805 on Build GRCh38
rs949542078 Chr.19: 7699985-7699985 on Build GRCh38
rs749775571 Chr.19: 7699738-7699738 on Build GRCh38
rs752360163 Chr.19: 7699664-7699664 on Build GRCh38
rs748244100 Chr.19: 7699826-7699826 on Build GRCh38
rs72558011 Chr.19: 7699900-7699900 on Build GRCh38
rs938063796 Chr.19: 7700014-7700014 on Build GRCh38
rs545424080 Chr.19: 7699652-7699652 on Build GRCh38
rs890274247 Chr.19: 7699839-7699839 on Build GRCh38
rs756015007 Chr.19: 7699931-7699932 on Build GRCh38
rs779874359 Chr.19: 7699746-7699746 on Build GRCh38
rs943135204 Chr.19: 7700000-7700000 on Build GRCh38
rs774989571 Chr.19: 7699759-7699759 on Build GRCh38
rs201613168 Chr.19: 7699787-7699787 on Build GRCh38
rs113187513 Chr.19: 7699933-7699933 on Build GRCh38
rs397713939 Chr.19: 7699663-7699664 on Build GRCh38
rs111370170 Chr.19: 7699715-7699715 on Build GRCh38
rs779331878 Chr.19: 7699794-7699794 on Build GRCh38
rs751651405 Chr.19: 7699705-7699705 on Build GRCh38
rs1051493654 Chr.19: 7699831-7699831 on Build GRCh38
rs778979534 Chr.19: 7699802-7699802 on Build GRCh38
rs140566353 Chr.19: 7699760-7699760 on Build GRCh38
rs779425156 Chr.19: 7699741-7699741 on Build GRCh38
rs750401440 Chr.19: 7699778-7699778 on Build GRCh38
rs537905897 Chr.19: 7699864-7699864 on Build GRCh38
rs778244821 Chr.19: 7699731-7699731 on Build GRCh38
rs897993363 Chr.19: 7699817-7699817 on Build GRCh38
rs778070903 Chr.19: 7699777-7699777 on Build GRCh38
rs755116276 Chr.19: 7699708-7699708 on Build GRCh38
rs1002181926 Chr.19: 7699886-7699886 on Build GRCh38
rs978405572 Chr.19: 7699975-7699975 on Build GRCh38
rs111846644 Chr.19: 7699926-7699926 on Build GRCh38
rs60818329 Chr.19: 7699940-7699940 on Build GRCh38
rs919275832 Chr.19: 7699727-7699727 on Build GRCh38
rs373421352 Chr.19: 7699714-7699714 on Build GRCh38
rs369245042 Chr.19: 7699875-7699875 on Build GRCh38
rs114566716 Chr.19: 7699806-7699806 on Build GRCh38
rs757927123 Chr.19: 7699633-7699633 on Build GRCh38

View Full Product Details