Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7960493-7960999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELAVL1
Entrez Gene ID
1994
Gene Name
ELAV like RNA binding protein 1
Gene Aliases
ELAV1, HUR, Hua, MelG
UniGene
Hs.184492
Gene Chromosome Location
Chr.19: 7958573-8005645 on Build GRCh38
Transcripts
NM_001419.2

SNP Detail

SNP ID Chromosome Location
rs765165606 Chr.19: 7960971-7960971 on Build GRCh38
rs143921406 Chr.19: 7960731-7960731 on Build GRCh38
rs958144838 Chr.19: 7960752-7960752 on Build GRCh38
rs1016089609 Chr.19: 7960633-7960633 on Build GRCh38
rs997112236 Chr.19: 7960591-7960591 on Build GRCh38
rs978073026 Chr.19: 7960713-7960713 on Build GRCh38
rs369819401 Chr.19: 7960542-7960542 on Build GRCh38
rs571393250 Chr.19: 7960923-7960923 on Build GRCh38
rs1047882482 Chr.19: 7960603-7960603 on Build GRCh38
rs972317162 Chr.19: 7960755-7960755 on Build GRCh38
rs527867670 Chr.19: 7960764-7960764 on Build GRCh38
rs557626641 Chr.19: 7960528-7960528 on Build GRCh38
rs1041633401 Chr.19: 7960921-7960921 on Build GRCh38
rs112011167 Chr.19: 7960831-7960831 on Build GRCh38
rs572838171 Chr.19: 7960604-7960604 on Build GRCh38
rs533374602 Chr.19: 7960866-7960866 on Build GRCh38
rs1041241388 Chr.19: 7960589-7960589 on Build GRCh38
rs758772658 Chr.19: 7960863-7960864 on Build GRCh38
rs189658634 Chr.19: 7960873-7960873 on Build GRCh38
rs949767960 Chr.19: 7960803-7960803 on Build GRCh38
rs563400250 Chr.19: 7960724-7960724 on Build GRCh38
rs930253842 Chr.19: 7960959-7960959 on Build GRCh38
rs937086429 Chr.19: 7960864-7960864 on Build GRCh38
rs909378078 Chr.19: 7960640-7960640 on Build GRCh38
rs1024872523 Chr.19: 7960699-7960699 on Build GRCh38
rs976463802 Chr.19: 7960535-7960535 on Build GRCh38
rs376389317 Chr.19: 7960720-7960720 on Build GRCh38
rs892979776 Chr.19: 7960876-7960876 on Build GRCh38
rs577606658 Chr.19: 7960566-7960566 on Build GRCh38
rs12983784 Chr.19: 7960607-7960607 on Build GRCh38
rs959432468 Chr.19: 7960681-7960681 on Build GRCh38
rs901452348 Chr.19: 7960590-7960590 on Build GRCh38
rs770421779 Chr.19: 7960723-7960723 on Build GRCh38
rs1045793987 Chr.19: 7960814-7960814 on Build GRCh38
rs776171497 Chr.19: 7960890-7960890 on Build GRCh38
rs562300297 Chr.19: 7960741-7960741 on Build GRCh38
rs542422643 Chr.19: 7960740-7960740 on Build GRCh38
rs73923951 Chr.19: 7960545-7960545 on Build GRCh38
rs940801056 Chr.19: 7960712-7960712 on Build GRCh38
rs911313506 Chr.19: 7960813-7960813 on Build GRCh38
rs889986597 Chr.19: 7960551-7960551 on Build GRCh38
rs927093999 Chr.19: 7960823-7960823 on Build GRCh38
rs548157433 Chr.19: 7960791-7960791 on Build GRCh38
rs1012086669 Chr.19: 7960697-7960697 on Build GRCh38
rs571152560 Chr.19: 7960838-7960838 on Build GRCh38

View Full Product Details