Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8333582-8334120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANK3
Entrez Gene ID
256949
Gene Name
KN motif and ankyrin repeat domains 3
Gene Aliases
ANKRD47
UniGene
Hs.745036
Gene Chromosome Location
Chr.19: 8322584-8343262 on Build GRCh38
Transcripts
NM_198471.2 XM_006722718.3 XM_011527884.2 XM_017026563.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6851285 c.1453G>A p.A485T Chr.19: 8334091-8334091 on Build GRCh38
3668390 c.1518C>A p.S506S Chr.19: 8334026-8334026 on Build GRCh38
4648361 c.1530C>T p.S510S Chr.19: 8334014-8334014 on Build GRCh38
3668391 c.1518C>A p.S506S Chr.19: 8334026-8334026 on Build GRCh38
5986033 c.1582C>T p.R528W Chr.19: 8333962-8333962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761324837 Chr.19: 8333860-8333862 on Build GRCh38
rs772939814 Chr.19: 8334097-8334097 on Build GRCh38
rs772637643 Chr.19: 8333925-8333925 on Build GRCh38
rs931925363 Chr.19: 8334030-8334030 on Build GRCh38
rs756180019 Chr.19: 8333974-8333974 on Build GRCh38
rs747587644 Chr.19: 8334048-8334048 on Build GRCh38
rs995515337 Chr.19: 8333729-8333729 on Build GRCh38
rs764965710 Chr.19: 8333951-8333951 on Build GRCh38
rs763661341 Chr.19: 8333821-8333821 on Build GRCh38
rs776260984 Chr.19: 8334071-8334071 on Build GRCh38
rs2009733 Chr.19: 8333830-8333830 on Build GRCh38
rs769486338 Chr.19: 8334072-8334083 on Build GRCh38
rs1027360062 Chr.19: 8333742-8333742 on Build GRCh38
rs771113678 Chr.19: 8333900-8333900 on Build GRCh38
rs1044076450 Chr.19: 8333670-8333670 on Build GRCh38
rs766432270 Chr.19: 8333959-8333959 on Build GRCh38
rs200669927 Chr.19: 8334075-8334075 on Build GRCh38
rs763238537 Chr.19: 8333763-8333763 on Build GRCh38
rs758464066 Chr.19: 8333845-8333845 on Build GRCh38
rs890939359 Chr.19: 8333696-8333696 on Build GRCh38
rs769017313 Chr.19: 8334061-8334061 on Build GRCh38
rs777879460 Chr.19: 8333882-8333882 on Build GRCh38
rs747787462 Chr.19: 8334068-8334079 on Build GRCh38
rs982971073 Chr.19: 8333646-8333646 on Build GRCh38
rs868269226 Chr.19: 8333675-8333675 on Build GRCh38
rs931791159 Chr.19: 8333710-8333710 on Build GRCh38
rs757533268 Chr.19: 8333987-8333987 on Build GRCh38
rs775969566 Chr.19: 8333943-8333943 on Build GRCh38
rs1052331811 Chr.19: 8333939-8333939 on Build GRCh38
rs761573919 Chr.19: 8333947-8333947 on Build GRCh38
rs951669387 Chr.19: 8333641-8333641 on Build GRCh38
rs746608015 Chr.19: 8334065-8334066 on Build GRCh38
rs773417305 Chr.19: 8333739-8333739 on Build GRCh38
rs535792407 Chr.19: 8333712-8333712 on Build GRCh38
rs753475201 Chr.19: 8333825-8333825 on Build GRCh38
rs2913955 Chr.19: 8334091-8334091 on Build GRCh38
rs1036459058 Chr.19: 8333808-8333808 on Build GRCh38
rs999816732 Chr.19: 8333688-8333688 on Build GRCh38
rs765161596 Chr.19: 8333836-8333836 on Build GRCh38
rs775913222 Chr.19: 8333928-8333928 on Build GRCh38
rs756423922 Chr.19: 8333607-8333607 on Build GRCh38
rs762518518 Chr.19: 8334077-8334088 on Build GRCh38
rs890992902 Chr.19: 8333940-8333940 on Build GRCh38
rs572205193 Chr.19: 8333756-8333756 on Build GRCh38
rs771280227 Chr.19: 8333775-8333775 on Build GRCh38
rs201862465 Chr.19: 8334076-8334076 on Build GRCh38
rs749487859 Chr.19: 8333897-8333897 on Build GRCh38
rs760232908 Chr.19: 8333820-8333820 on Build GRCh38
rs943933380 Chr.19: 8334005-8334005 on Build GRCh38
rs531314408 Chr.19: 8333937-8333937 on Build GRCh38
rs939115993 Chr.19: 8333649-8333649 on Build GRCh38
rs775042110 Chr.19: 8333818-8333818 on Build GRCh38
rs772100856 Chr.19: 8333701-8333701 on Build GRCh38
rs779106442 Chr.19: 8334015-8334015 on Build GRCh38
rs750217307 Chr.19: 8333837-8333837 on Build GRCh38
rs774872216 Chr.19: 8333776-8333776 on Build GRCh38
rs562990626 Chr.19: 8334026-8334026 on Build GRCh38
rs773122344 Chr.19: 8333880-8333880 on Build GRCh38
rs920181715 Chr.19: 8333618-8333618 on Build GRCh38
rs903772237 Chr.19: 8333681-8333681 on Build GRCh38
rs1003662179 Chr.19: 8333839-8333839 on Build GRCh38
rs375936820 Chr.19: 8333903-8333903 on Build GRCh38
rs760119579 Chr.19: 8333933-8333933 on Build GRCh38
rs890542970 Chr.19: 8333716-8333716 on Build GRCh38
rs751463795 Chr.19: 8333963-8333963 on Build GRCh38
rs1056125199 Chr.19: 8333656-8333656 on Build GRCh38
rs1039850569 Chr.19: 8333953-8333953 on Build GRCh38
rs577350856 Chr.19: 8333919-8333919 on Build GRCh38
rs940510744 Chr.19: 8334045-8334045 on Build GRCh38
rs781466759 Chr.19: 8333703-8333703 on Build GRCh38
rs2913954 Chr.19: 8333713-8333713 on Build GRCh38
rs772680384 Chr.19: 8334074-8334085 on Build GRCh38
rs748181101 Chr.19: 8333862-8333862 on Build GRCh38
rs2241589 Chr.19: 8333647-8333647 on Build GRCh38
rs951668848 Chr.19: 8333743-8333743 on Build GRCh38
rs1035845011 Chr.19: 8333844-8333844 on Build GRCh38
rs749305554 Chr.19: 8333983-8333983 on Build GRCh38
rs542991221 Chr.19: 8333971-8333971 on Build GRCh38
rs899124193 Chr.19: 8333814-8333814 on Build GRCh38
rs757832163 Chr.19: 8333695-8333695 on Build GRCh38
rs760077076 Chr.19: 8333794-8333794 on Build GRCh38
rs6146458 Chr.19: 8334076-8334087 on Build GRCh38
rs539828579 Chr.19: 8333660-8333660 on Build GRCh38
rs369037048 Chr.19: 8334036-8334036 on Build GRCh38
rs528922304 Chr.19: 8333662-8333662 on Build GRCh38
rs746218686 Chr.19: 8333920-8333920 on Build GRCh38
rs947740747 Chr.19: 8333664-8333664 on Build GRCh38
rs995960419 Chr.19: 8334032-8334032 on Build GRCh38
rs916331349 Chr.19: 8333892-8333892 on Build GRCh38
rs557847876 Chr.19: 8333895-8333895 on Build GRCh38
rs988426037 Chr.19: 8333659-8333659 on Build GRCh38
rs569964478 Chr.19: 8333689-8333689 on Build GRCh38
rs777950582 Chr.19: 8333979-8333979 on Build GRCh38
rs199822445 Chr.19: 8334077-8334077 on Build GRCh38
rs111751275 Chr.19: 8334066-8334077 on Build GRCh38
rs921884078 Chr.19: 8333708-8333708 on Build GRCh38
rs746378258 Chr.19: 8333722-8333722 on Build GRCh38
rs775887425 Chr.19: 8334042-8334042 on Build GRCh38
rs780134178 Chr.19: 8333859-8333859 on Build GRCh38
rs899914548 Chr.19: 8333988-8333988 on Build GRCh38
rs750307402 Chr.19: 8333955-8333955 on Build GRCh38
rs767991428 Chr.19: 8333799-8333799 on Build GRCh38
rs758199906 Chr.19: 8333956-8333956 on Build GRCh38

View Full Product Details