Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8334124-8334614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANK3
Entrez Gene ID
256949
Gene Name
KN motif and ankyrin repeat domains 3
Gene Aliases
ANKRD47
UniGene
Hs.745036
Gene Chromosome Location
Chr.19: 8322584-8343262 on Build GRCh38
Transcripts
NM_198471.2 XM_006722718.3 XM_011527884.2 XM_017026563.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4630555 c.1426G>T p.E476* Chr.19: 8334321-8334321 on Build GRCh38
3541583 c.1258G>A p.E420K Chr.19: 8334569-8334569 on Build GRCh38
3960718 c.1327+9C>G p.? Chr.19: 8334491-8334491 on Build GRCh38
2730229 c.1393A>G p.K465E Chr.19: 8334354-8334354 on Build GRCh38
4782212 c.1327+2T>A p.? Chr.19: 8334498-8334498 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775318392 Chr.19: 8334560-8334560 on Build GRCh38
rs142794933 Chr.19: 8334345-8334345 on Build GRCh38
rs772809333 Chr.19: 8334272-8334273 on Build GRCh38
rs767344916 Chr.19: 8334153-8334153 on Build GRCh38
rs924994555 Chr.19: 8334162-8334162 on Build GRCh38
rs781732916 Chr.19: 8334441-8334441 on Build GRCh38
rs768987295 Chr.19: 8334502-8334502 on Build GRCh38
rs774262181 Chr.19: 8334387-8334387 on Build GRCh38
rs760101852 Chr.19: 8334423-8334423 on Build GRCh38
rs765165253 Chr.19: 8334461-8334461 on Build GRCh38
rs745692465 Chr.19: 8334463-8334463 on Build GRCh38
rs969179844 Chr.19: 8334154-8334154 on Build GRCh38
rs201328334 Chr.19: 8334480-8334480 on Build GRCh38
rs759567371 Chr.19: 8334150-8334150 on Build GRCh38
rs767812564 Chr.19: 8334518-8334518 on Build GRCh38
rs770468433 Chr.19: 8334386-8334386 on Build GRCh38
rs1000648099 Chr.19: 8334412-8334412 on Build GRCh38
rs1056613033 Chr.19: 8334496-8334496 on Build GRCh38
rs765264510 Chr.19: 8334427-8334427 on Build GRCh38
rs778421405 Chr.19: 8334526-8334526 on Build GRCh38
rs771717450 Chr.19: 8334478-8334478 on Build GRCh38
rs934977039 Chr.19: 8334216-8334216 on Build GRCh38
rs371277106 Chr.19: 8334424-8334424 on Build GRCh38
rs570077993 Chr.19: 8334588-8334588 on Build GRCh38
rs905443281 Chr.19: 8334181-8334181 on Build GRCh38
rs931431662 Chr.19: 8334266-8334266 on Build GRCh38
rs920671249 Chr.19: 8334477-8334477 on Build GRCh38
rs754017042 Chr.19: 8334158-8334158 on Build GRCh38
rs758673221 Chr.19: 8334435-8334435 on Build GRCh38
rs8106699 Chr.19: 8334388-8334388 on Build GRCh38
rs779122979 Chr.19: 8334278-8334278 on Build GRCh38
rs762733520 Chr.19: 8334384-8334384 on Build GRCh38
rs1057024770 Chr.19: 8334355-8334355 on Build GRCh38
rs750476231 Chr.19: 8334430-8334430 on Build GRCh38
rs748813255 Chr.19: 8334543-8334543 on Build GRCh38
rs1009749823 Chr.19: 8334300-8334300 on Build GRCh38
rs758797647 Chr.19: 8334313-8334313 on Build GRCh38
rs748625414 Chr.19: 8334444-8334444 on Build GRCh38
rs756743213 Chr.19: 8334525-8334525 on Build GRCh38
rs373472364 Chr.19: 8334485-8334485 on Build GRCh38
rs766210140 Chr.19: 8334306-8334307 on Build GRCh38
rs969274500 Chr.19: 8334397-8334397 on Build GRCh38
rs374883316 Chr.19: 8334438-8334438 on Build GRCh38
rs769270108 Chr.19: 8334340-8334340 on Build GRCh38
rs1039563230 Chr.19: 8334257-8334257 on Build GRCh38
rs144654054 Chr.19: 8334192-8334193 on Build GRCh38
rs767497580 Chr.19: 8334392-8334392 on Build GRCh38
rs887151054 Chr.19: 8334307-8334307 on Build GRCh38
rs747672940 Chr.19: 8334359-8334359 on Build GRCh38
rs1020439649 Chr.19: 8334311-8334311 on Build GRCh38
rs773029634 Chr.19: 8334383-8334383 on Build GRCh38
rs757263409 Chr.19: 8334276-8334276 on Build GRCh38
rs761908569 Chr.19: 8334407-8334407 on Build GRCh38
rs753637463 Chr.19: 8334369-8334370 on Build GRCh38
rs781482388 Chr.19: 8334347-8334347 on Build GRCh38
rs943895363 Chr.19: 8334256-8334256 on Build GRCh38
rs533514076 Chr.19: 8334568-8334568 on Build GRCh38
rs148544896 Chr.19: 8334331-8334331 on Build GRCh38
rs868491681 Chr.19: 8334536-8334536 on Build GRCh38
rs766415077 Chr.19: 8334148-8334148 on Build GRCh38
rs778651841 Chr.19: 8334549-8334549 on Build GRCh38
rs939372692 Chr.19: 8334488-8334488 on Build GRCh38
rs759724793 Chr.19: 8334590-8334590 on Build GRCh38
rs773809499 Chr.19: 8334457-8334457 on Build GRCh38
rs767839100 Chr.19: 8334591-8334591 on Build GRCh38
rs376916341 Chr.19: 8334558-8334558 on Build GRCh38
rs760483926 Chr.19: 8334304-8334306 on Build GRCh38
rs780513093 Chr.19: 8334314-8334314 on Build GRCh38
rs1048945642 Chr.19: 8334269-8334269 on Build GRCh38
rs762872392 Chr.19: 8334492-8334492 on Build GRCh38
rs754937945 Chr.19: 8334428-8334429 on Build GRCh38
rs988089806 Chr.19: 8334226-8334226 on Build GRCh38
rs1023152551 Chr.19: 8334389-8334389 on Build GRCh38
rs770685641 Chr.19: 8334456-8334456 on Build GRCh38
rs752522041 Chr.19: 8334401-8334401 on Build GRCh38
rs200912318 Chr.19: 8334524-8334524 on Build GRCh38
rs372495490 Chr.19: 8334455-8334455 on Build GRCh38
rs562114233 Chr.19: 8334251-8334251 on Build GRCh38
rs746798805 Chr.19: 8334562-8334562 on Build GRCh38
rs766327411 Chr.19: 8334579-8334579 on Build GRCh38
rs771728143 Chr.19: 8334559-8334559 on Build GRCh38
rs151045287 Chr.19: 8334361-8334361 on Build GRCh38
rs750598901 Chr.19: 8334305-8334305 on Build GRCh38
rs975302281 Chr.19: 8334487-8334487 on Build GRCh38
rs899633296 Chr.19: 8334260-8334260 on Build GRCh38
rs185143796 Chr.19: 8334574-8334574 on Build GRCh38
rs1043939378 Chr.19: 8334529-8334529 on Build GRCh38
rs200557228 Chr.19: 8334506-8334506 on Build GRCh38
rs1003714628 Chr.19: 8334206-8334206 on Build GRCh38
rs755238642 Chr.19: 8334440-8334440 on Build GRCh38
rs766634780 Chr.19: 8334504-8334504 on Build GRCh38
rs773342563 Chr.19: 8334486-8334486 on Build GRCh38
rs756667715 Chr.19: 8334548-8334548 on Build GRCh38
rs904264384 Chr.19: 8334553-8334553 on Build GRCh38
rs780133358 Chr.19: 8334436-8334436 on Build GRCh38

View Full Product Details