Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10288108-10288640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICAM4
Entrez Gene ID
3386
Gene Name
intercellular adhesion molecule 4 (Landsteiner-Wiener blood group)
Gene Aliases
CD242, LW
UniGene
Hs.706750
Gene Chromosome Location
Chr.19: 10286967-10288584 on Build GRCh38
Transcripts
NM_001039132.2 NM_001544.4 NM_022377.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5800715 c.765C>T p.S255S Chr.19: 10288130-10288130 on Build GRCh38
5972670 c.776G>A p.R259K Chr.19: 10288141-10288141 on Build GRCh38
161652 c.791C>T p.S264F Chr.19: 10288156-10288156 on Build GRCh38
6033072 c.757C>T p.R253W Chr.19: 10288122-10288122 on Build GRCh38
5016610 c.787_788insA p.S264fs*>10 Chr.19: 10288152-10288153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004185068 Chr.19: 10288469-10288469 on Build GRCh38
rs991709497 Chr.19: 10288303-10288303 on Build GRCh38
rs55720142 Chr.19: 10288193-10288193 on Build GRCh38
rs927380669 Chr.19: 10288487-10288487 on Build GRCh38
rs3745267 Chr.19: 10288225-10288225 on Build GRCh38
rs781281480 Chr.19: 10288131-10288131 on Build GRCh38
rs55668681 Chr.19: 10288188-10288188 on Build GRCh38
rs191098583 Chr.19: 10288436-10288436 on Build GRCh38
rs750564722 Chr.19: 10288184-10288184 on Build GRCh38
rs747607311 Chr.19: 10288134-10288134 on Build GRCh38
rs767480804 Chr.19: 10288215-10288215 on Build GRCh38
rs934290202 Chr.19: 10288381-10288381 on Build GRCh38
rs56057687 Chr.19: 10288433-10288433 on Build GRCh38
rs55909127 Chr.19: 10288245-10288245 on Build GRCh38
rs201399464 Chr.19: 10288138-10288138 on Build GRCh38
rs55695233 Chr.19: 10288246-10288246 on Build GRCh38
rs568770787 Chr.19: 10288227-10288227 on Build GRCh38
rs766655632 Chr.19: 10288187-10288187 on Build GRCh38
rs971327891 Chr.19: 10288582-10288583 on Build GRCh38
rs928514016 Chr.19: 10288317-10288317 on Build GRCh38
rs755918962 Chr.19: 10288585-10288586 on Build GRCh38
rs55677215 Chr.19: 10288253-10288253 on Build GRCh38
rs376984546 Chr.19: 10288430-10288430 on Build GRCh38
rs1031516386 Chr.19: 10288594-10288594 on Build GRCh38
rs914282644 Chr.19: 10288412-10288412 on Build GRCh38
rs887112332 Chr.19: 10288466-10288466 on Build GRCh38
rs902947462 Chr.19: 10288464-10288464 on Build GRCh38
rs113892276 Chr.19: 10288137-10288137 on Build GRCh38
rs55761920 Chr.19: 10288205-10288205 on Build GRCh38
rs1022907092 Chr.19: 10288319-10288319 on Build GRCh38
rs762373199 Chr.19: 10288173-10288173 on Build GRCh38
rs1004301233 Chr.19: 10288265-10288265 on Build GRCh38
rs56081140 Chr.19: 10288247-10288247 on Build GRCh38
rs572585716 Chr.19: 10288272-10288273 on Build GRCh38
rs781559447 Chr.19: 10288424-10288424 on Build GRCh38
rs947280732 Chr.19: 10288379-10288379 on Build GRCh38
rs765979139 Chr.19: 10288276-10288276 on Build GRCh38
rs1023214195 Chr.19: 10288580-10288580 on Build GRCh38
rs769091807 Chr.19: 10288168-10288168 on Build GRCh38
rs891347520 Chr.19: 10288508-10288508 on Build GRCh38
rs56205873 Chr.19: 10288209-10288209 on Build GRCh38
rs989924070 Chr.19: 10288410-10288410 on Build GRCh38
rs55658286 Chr.19: 10288237-10288237 on Build GRCh38
rs900115410 Chr.19: 10288425-10288425 on Build GRCh38
rs946929083 Chr.19: 10288421-10288421 on Build GRCh38
rs937449491 Chr.19: 10288517-10288517 on Build GRCh38
rs534687123 Chr.19: 10288526-10288526 on Build GRCh38
rs1055744521 Chr.19: 10288355-10288355 on Build GRCh38
rs557626088 Chr.19: 10288449-10288449 on Build GRCh38
rs56087532 Chr.19: 10288249-10288249 on Build GRCh38
rs56219232 Chr.19: 10288211-10288211 on Build GRCh38
rs779541160 Chr.19: 10288423-10288423 on Build GRCh38
rs56392518 Chr.19: 10288242-10288242 on Build GRCh38
rs971277103 Chr.19: 10288352-10288352 on Build GRCh38
rs938612090 Chr.19: 10288327-10288327 on Build GRCh38
rs144913870 Chr.19: 10288166-10288166 on Build GRCh38
rs541877224 Chr.19: 10288558-10288558 on Build GRCh38
rs55900686 Chr.19: 10288199-10288199 on Build GRCh38
rs756380302 Chr.19: 10288583-10288585 on Build GRCh38
rs141101411 Chr.19: 10288151-10288151 on Build GRCh38
rs55675105 Chr.19: 10288241-10288241 on Build GRCh38
rs777108705 Chr.19: 10288171-10288171 on Build GRCh38
rs55805249 Chr.19: 10288248-10288248 on Build GRCh38
rs56350006 Chr.19: 10288244-10288244 on Build GRCh38
rs763036721 Chr.19: 10288185-10288185 on Build GRCh38
rs767071656 Chr.19: 10288160-10288160 on Build GRCh38
rs765268784 Chr.19: 10288179-10288179 on Build GRCh38
rs557694072 Chr.19: 10288278-10288278 on Build GRCh38
rs1008880702 Chr.19: 10288522-10288522 on Build GRCh38
rs893977191 Chr.19: 10288169-10288169 on Build GRCh38
rs981109870 Chr.19: 10288358-10288358 on Build GRCh38
rs751247769 Chr.19: 10288196-10288196 on Build GRCh38
rs146427583 Chr.19: 10288141-10288141 on Build GRCh38
rs536149845 Chr.19: 10288367-10288367 on Build GRCh38
rs56249494 Chr.19: 10288252-10288252 on Build GRCh38
rs770634773 Chr.19: 10288154-10288154 on Build GRCh38
rs974542661 Chr.19: 10288174-10288174 on Build GRCh38
rs998436286 Chr.19: 10288475-10288475 on Build GRCh38
rs55988724 Chr.19: 10288243-10288243 on Build GRCh38

View Full Product Details