Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 34366253-34366745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPI
Entrez Gene ID
2821
Gene Name
glucose-6-phosphate isomerase
Gene Aliases
AMF, GNPI, NLK, PGI, PHI, SA-36, SA36
UniGene
Hs.466471
Gene Chromosome Location
Chr.19: 34359592-34402413 on Build GRCh38
Transcripts
NM_000175.3 NM_001184722.1 NM_001289789.1 NM_001289790.1 XM_005258764.2 XM_006723148.2 XM_011526754.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3532305 c.127A>G p.T43A Chr.19: 34366349-34366349 on Build GRCh38
5429982 c.126G>C p.L42F Chr.19: 34366348-34366348 on Build GRCh38
3672841 c.156G>T p.L52L Chr.19: 34366378-34366378 on Build GRCh38
6673259 c.171G>T p.K57N Chr.19: 34366393-34366393 on Build GRCh38
6673275 c.314G>A p.R105Q Chr.19: 34366419-34366419 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568857075 Chr.19: 34366657-34366657 on Build GRCh38
rs769407863 Chr.19: 34366330-34366331 on Build GRCh38
rs775368109 Chr.19: 34366331-34366331 on Build GRCh38
rs1016342810 Chr.19: 34366317-34366317 on Build GRCh38
rs141473845 Chr.19: 34366405-34366405 on Build GRCh38
rs764508521 Chr.19: 34366416-34366416 on Build GRCh38
rs554714757 Chr.19: 34366693-34366693 on Build GRCh38
rs1039077693 Chr.19: 34366508-34366508 on Build GRCh38
rs966503214 Chr.19: 34366466-34366466 on Build GRCh38
rs777050721 Chr.19: 34366309-34366309 on Build GRCh38
rs779558324 Chr.19: 34366354-34366354 on Build GRCh38
rs1036060840 Chr.19: 34366671-34366673 on Build GRCh38
rs762109743 Chr.19: 34366419-34366419 on Build GRCh38
rs763215357 Chr.19: 34366477-34366488 on Build GRCh38
rs530935030 Chr.19: 34366350-34366350 on Build GRCh38
rs1016278690 Chr.19: 34366678-34366678 on Build GRCh38
rs369971788 Chr.19: 34366423-34366423 on Build GRCh38
rs772004540 Chr.19: 34366390-34366390 on Build GRCh38
rs755578867 Chr.19: 34366351-34366351 on Build GRCh38
rs1036216785 Chr.19: 34366611-34366611 on Build GRCh38
rs376682804 Chr.19: 34366458-34366458 on Build GRCh38
rs532119898 Chr.19: 34366585-34366585 on Build GRCh38
rs754835193 Chr.19: 34366360-34366360 on Build GRCh38
rs567812892 Chr.19: 34366715-34366715 on Build GRCh38
rs537510015 Chr.19: 34366659-34366659 on Build GRCh38
rs775411168 Chr.19: 34366412-34366412 on Build GRCh38
rs886519636 Chr.19: 34366617-34366617 on Build GRCh38
rs149486766 Chr.19: 34366336-34366336 on Build GRCh38
rs1044290895 Chr.19: 34366477-34366477 on Build GRCh38
rs747882262 Chr.19: 34366375-34366375 on Build GRCh38
rs773991318 Chr.19: 34366295-34366295 on Build GRCh38
rs886634169 Chr.19: 34366561-34366561 on Build GRCh38
rs933488928 Chr.19: 34366310-34366310 on Build GRCh38
rs748875055 Chr.19: 34366358-34366358 on Build GRCh38
rs1002695926 Chr.19: 34366607-34366607 on Build GRCh38
rs767146847 Chr.19: 34366342-34366342 on Build GRCh38
rs199803942 Chr.19: 34366415-34366415 on Build GRCh38
rs923552541 Chr.19: 34366518-34366518 on Build GRCh38
rs750026300 Chr.19: 34366470-34366470 on Build GRCh38
rs560712135 Chr.19: 34366418-34366418 on Build GRCh38
rs529844631 Chr.19: 34366421-34366421 on Build GRCh38
rs768446291 Chr.19: 34366294-34366294 on Build GRCh38
rs545112581 Chr.19: 34366318-34366318 on Build GRCh38
rs199673161 Chr.19: 34366447-34366447 on Build GRCh38
rs373498716 Chr.19: 34366480-34366480 on Build GRCh38
rs905934208 Chr.19: 34366630-34366630 on Build GRCh38
rs769947359 Chr.19: 34366411-34366411 on Build GRCh38
rs368757760 Chr.19: 34366471-34366471 on Build GRCh38
rs749211585 Chr.19: 34366287-34366287 on Build GRCh38
rs1047936795 Chr.19: 34366555-34366557 on Build GRCh38
rs780976808 Chr.19: 34366279-34366280 on Build GRCh38
rs551915593 Chr.19: 34366640-34366640 on Build GRCh38
rs933538748 Chr.19: 34366540-34366540 on Build GRCh38
rs779912076 Chr.19: 34366286-34366286 on Build GRCh38
rs764711659 Chr.19: 34366331-34366331 on Build GRCh38
rs778976371 Chr.19: 34366365-34366365 on Build GRCh38
rs1037827034 Chr.19: 34366586-34366586 on Build GRCh38
rs757710396 Chr.19: 34366339-34366339 on Build GRCh38
rs905714982 Chr.19: 34366479-34366479 on Build GRCh38
rs566472071 Chr.19: 34366503-34366503 on Build GRCh38
rs776102729 Chr.19: 34366323-34366323 on Build GRCh38
rs925022676 Chr.19: 34366488-34366488 on Build GRCh38
rs1006278450 Chr.19: 34366304-34366304 on Build GRCh38
rs771274643 Chr.19: 34366297-34366297 on Build GRCh38
rs763394513 Chr.19: 34366329-34366329 on Build GRCh38
rs745882863 Chr.19: 34366294-34366294 on Build GRCh38
rs986977053 Chr.19: 34366324-34366324 on Build GRCh38
rs1006373353 Chr.19: 34366563-34366563 on Build GRCh38
rs943084936 Chr.19: 34366496-34366496 on Build GRCh38
rs754574122 Chr.19: 34366484-34366484 on Build GRCh38
rs761704715 Chr.19: 34366296-34366296 on Build GRCh38
rs777756390 Chr.19: 34366685-34366685 on Build GRCh38
rs776925897 Chr.19: 34366404-34366404 on Build GRCh38

View Full Product Details