Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35118934-35119428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXYD3
Entrez Gene ID
5349
Gene Name
FXYD domain containing ion transport regulator 3
Gene Aliases
MAT8, PLML
UniGene
Hs.301350
Gene Chromosome Location
Chr.19: 35115821-35124324 on Build GRCh38
Transcripts
NM_001136007.1 NM_001136008.1 NM_001136009.1 NM_001136010.1 NM_001136011.1 NM_001136012.1 NM_005971.3 NM_021910.2 XM_005258996.4 XM_011527032.2 XM_017026877.1 XM_017026878.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6049530 c.179A>G p.K60R Chr.19: 35119384-35119384 on Build GRCh38
1161269 c.9G>T p.K3N Chr.19: 35119385-35119385 on Build GRCh38
6667991 c.15C>T p.T5T Chr.19: 35119391-35119391 on Build GRCh38
4154168 c.78G>T p.P26P Chr.19: 35119172-35119172 on Build GRCh38
6667992 c.15C>T p.T5T Chr.19: 35119391-35119391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756321989 Chr.19: 35119150-35119150 on Build GRCh38
rs1040363871 Chr.19: 35119193-35119193 on Build GRCh38
rs778251891 Chr.19: 35119279-35119279 on Build GRCh38
rs780778533 Chr.19: 35119354-35119354 on Build GRCh38
rs745372389 Chr.19: 35119357-35119357 on Build GRCh38
rs748156651 Chr.19: 35119138-35119141 on Build GRCh38
rs375912889 Chr.19: 35119228-35119228 on Build GRCh38
rs933047292 Chr.19: 35119340-35119340 on Build GRCh38
rs10543115 Chr.19: 35119069-35119071 on Build GRCh38
rs531609919 Chr.19: 35119136-35119136 on Build GRCh38
rs780735784 Chr.19: 35119272-35119272 on Build GRCh38
rs780295655 Chr.19: 35119157-35119157 on Build GRCh38
rs750618067 Chr.19: 35119225-35119225 on Build GRCh38
rs753887316 Chr.19: 35119247-35119247 on Build GRCh38
rs779622390 Chr.19: 35119363-35119363 on Build GRCh38
rs760932539 Chr.19: 35119322-35119322 on Build GRCh38
rs375704691 Chr.19: 35119089-35119089 on Build GRCh38
rs377069678 Chr.19: 35119146-35119146 on Build GRCh38
rs749200090 Chr.19: 35119364-35119364 on Build GRCh38
rs75407870 Chr.19: 35119107-35119107 on Build GRCh38
rs751966616 Chr.19: 35119350-35119350 on Build GRCh38
rs919425174 Chr.19: 35119339-35119339 on Build GRCh38
rs777049591 Chr.19: 35119294-35119294 on Build GRCh38
rs748531703 Chr.19: 35119250-35119250 on Build GRCh38
rs370586631 Chr.19: 35119349-35119349 on Build GRCh38
rs202178065 Chr.19: 35119069-35119069 on Build GRCh38
rs74977552 Chr.19: 35119269-35119269 on Build GRCh38
rs749738075 Chr.19: 35119160-35119160 on Build GRCh38
rs748858058 Chr.19: 35119405-35119405 on Build GRCh38
rs576060524 Chr.19: 35119330-35119330 on Build GRCh38
rs755471077 Chr.19: 35119161-35119161 on Build GRCh38
rs757679976 Chr.19: 35119391-35119391 on Build GRCh38
rs972095941 Chr.19: 35119313-35119313 on Build GRCh38
rs759465085 Chr.19: 35119200-35119200 on Build GRCh38
rs80021793 Chr.19: 35119133-35119133 on Build GRCh38
rs774319777 Chr.19: 35119315-35119315 on Build GRCh38
rs755478996 Chr.19: 35119249-35119249 on Build GRCh38
rs371019682 Chr.19: 35119343-35119343 on Build GRCh38
rs914147394 Chr.19: 35119056-35119056 on Build GRCh38
rs960744663 Chr.19: 35119207-35119207 on Build GRCh38
rs765909855 Chr.19: 35119068-35119069 on Build GRCh38
rs926689633 Chr.19: 35118962-35118962 on Build GRCh38
rs758533020 Chr.19: 35119214-35119215 on Build GRCh38
rs370026720 Chr.19: 35119098-35119098 on Build GRCh38
rs547252248 Chr.19: 35119227-35119227 on Build GRCh38
rs745414605 Chr.19: 35119280-35119280 on Build GRCh38
rs781632442 Chr.19: 35119396-35119396 on Build GRCh38
rs776146220 Chr.19: 35119198-35119198 on Build GRCh38
rs1032700179 Chr.19: 35119233-35119233 on Build GRCh38
rs544879179 Chr.19: 35118967-35118967 on Build GRCh38
rs199550224 Chr.19: 35119068-35119068 on Build GRCh38
rs776657302 Chr.19: 35119372-35119372 on Build GRCh38
rs752460954 Chr.19: 35119134-35119134 on Build GRCh38
rs901022948 Chr.19: 35119045-35119045 on Build GRCh38
rs1032648139 Chr.19: 35119101-35119101 on Build GRCh38
rs779383283 Chr.19: 35119403-35119403 on Build GRCh38
rs8113168 Chr.19: 35119323-35119323 on Build GRCh38
rs772906015 Chr.19: 35119186-35119186 on Build GRCh38
rs2278994 Chr.19: 35119341-35119341 on Build GRCh38
rs369284559 Chr.19: 35119171-35119171 on Build GRCh38
rs746593023 Chr.19: 35119369-35119369 on Build GRCh38
rs769131598 Chr.19: 35119359-35119359 on Build GRCh38
rs762962129 Chr.19: 35119284-35119284 on Build GRCh38
rs10426785 Chr.19: 35119077-35119077 on Build GRCh38
rs758701917 Chr.19: 35119259-35119259 on Build GRCh38
rs568277404 Chr.19: 35119196-35119196 on Build GRCh38
rs763423896 Chr.19: 35119380-35119380 on Build GRCh38
rs145627296 Chr.19: 35119398-35119398 on Build GRCh38
rs746621255 Chr.19: 35119261-35119261 on Build GRCh38
rs767841065 Chr.19: 35119224-35119224 on Build GRCh38
rs771706647 Chr.19: 35119177-35119177 on Build GRCh38
rs752841610 Chr.19: 35119377-35119377 on Build GRCh38
rs756875089 Chr.19: 35119352-35119352 on Build GRCh38
rs751921647 Chr.19: 35119388-35119388 on Build GRCh38
rs1033758383 Chr.19: 35119178-35119178 on Build GRCh38
rs768737517 Chr.19: 35119368-35119368 on Build GRCh38
rs762726782 Chr.19: 35119220-35119220 on Build GRCh38
rs1018174260 Chr.19: 35119040-35119040 on Build GRCh38
rs943326207 Chr.19: 35119187-35119187 on Build GRCh38
rs765187472 Chr.19: 35119376-35119376 on Build GRCh38
rs967574432 Chr.19: 35119096-35119096 on Build GRCh38
rs539530402 Chr.19: 35119282-35119282 on Build GRCh38
rs770357800 Chr.19: 35119197-35119197 on Build GRCh38
rs778113035 Chr.19: 35119260-35119260 on Build GRCh38
rs768501155 Chr.19: 35119314-35119314 on Build GRCh38
rs371344905 Chr.19: 35118960-35118960 on Build GRCh38
rs774659184 Chr.19: 35118976-35118976 on Build GRCh38
rs917511514 Chr.19: 35119109-35119109 on Build GRCh38
rs755824659 Chr.19: 35119399-35119399 on Build GRCh38
rs62120434 Chr.19: 35119384-35119384 on Build GRCh38
rs930706522 Chr.19: 35118959-35118959 on Build GRCh38
rs769769729 Chr.19: 35119281-35119281 on Build GRCh38
rs958337024 Chr.19: 35119104-35119104 on Build GRCh38
rs117797884 Chr.19: 35119084-35119084 on Build GRCh38
rs768386918 Chr.19: 35119370-35119370 on Build GRCh38
rs372776412 Chr.19: 35119203-35119203 on Build GRCh38
rs905726698 Chr.19: 35119164-35119164 on Build GRCh38
rs534126569 Chr.19: 35119208-35119208 on Build GRCh38

View Full Product Details