Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35123191-35123690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXYD3
Entrez Gene ID
5349
Gene Name
FXYD domain containing ion transport regulator 3
Gene Aliases
MAT8, PLML
UniGene
Hs.301350
Gene Chromosome Location
Chr.19: 35115821-35124324 on Build GRCh38
Transcripts
NM_001136007.1 NM_001136008.1 NM_001136011.1 NM_001136012.1 NM_005971.3 NM_021910.2 XM_005258996.4 XM_011527032.2 XM_017026877.1 XM_017026878.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5972870 c.388C>A p.P130T Chr.19: 35123278-35123278 on Build GRCh38
5972871 c.295C>A p.P99T Chr.19: 35123278-35123278 on Build GRCh38
4924535 c.333C>T p.A111A Chr.19: 35123448-35123448 on Build GRCh38
5888408 c.314T>A p.L105H Chr.19: 35123297-35123297 on Build GRCh38
5531522 c.414C>T p.T138T Chr.19: 35123304-35123304 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937088 Chr.19: 35123368-35123368 on Build GRCh38
rs1802804 Chr.19: 35123496-35123496 on Build GRCh38
rs777012208 Chr.19: 35123565-35123565 on Build GRCh38
rs904587900 Chr.19: 35123633-35123633 on Build GRCh38
rs369601420 Chr.19: 35123235-35123235 on Build GRCh38
rs369257217 Chr.19: 35123317-35123317 on Build GRCh38
rs565435089 Chr.19: 35123216-35123216 on Build GRCh38
rs1022215833 Chr.19: 35123584-35123584 on Build GRCh38
rs376419075 Chr.19: 35123341-35123341 on Build GRCh38
rs879151760 Chr.19: 35123434-35123435 on Build GRCh38
rs779225395 Chr.19: 35123220-35123220 on Build GRCh38
rs921936 Chr.19: 35123344-35123344 on Build GRCh38
rs113122676 Chr.19: 35123269-35123269 on Build GRCh38
rs373306109 Chr.19: 35123492-35123492 on Build GRCh38
rs756979211 Chr.19: 35123613-35123614 on Build GRCh38
rs752006247 Chr.19: 35123464-35123464 on Build GRCh38
rs3192433 Chr.19: 35123273-35123273 on Build GRCh38
rs201302981 Chr.19: 35123308-35123308 on Build GRCh38
rs377226344 Chr.19: 35123283-35123283 on Build GRCh38
rs773223877 Chr.19: 35123267-35123267 on Build GRCh38
rs201621694 Chr.19: 35123247-35123247 on Build GRCh38
rs749200117 Chr.19: 35123491-35123491 on Build GRCh38
rs558431476 Chr.19: 35123411-35123411 on Build GRCh38
rs770276554 Chr.19: 35123255-35123255 on Build GRCh38
rs890028195 Chr.19: 35123305-35123305 on Build GRCh38
rs73927930 Chr.19: 35123562-35123562 on Build GRCh38
rs111558644 Chr.19: 35123268-35123268 on Build GRCh38
rs370549538 Chr.19: 35123468-35123468 on Build GRCh38
rs538543270 Chr.19: 35123405-35123405 on Build GRCh38
rs1009517743 Chr.19: 35123574-35123574 on Build GRCh38
rs377297118 Chr.19: 35123527-35123527 on Build GRCh38
rs939749285 Chr.19: 35123218-35123218 on Build GRCh38
rs745747079 Chr.19: 35123263-35123263 on Build GRCh38
rs745427386 Chr.19: 35123342-35123342 on Build GRCh38
rs373469267 Chr.19: 35123334-35123334 on Build GRCh38
rs751129369 Chr.19: 35123330-35123330 on Build GRCh38
rs770130528 Chr.19: 35123369-35123370 on Build GRCh38
rs753031634 Chr.19: 35123350-35123353 on Build GRCh38
rs113577293 Chr.19: 35123369-35123369 on Build GRCh38
rs761401553 Chr.19: 35123233-35123233 on Build GRCh38
rs4835 Chr.19: 35123630-35123630 on Build GRCh38
rs891917946 Chr.19: 35123363-35123363 on Build GRCh38
rs760710551 Chr.19: 35123302-35123303 on Build GRCh38
rs763479134 Chr.19: 35123447-35123447 on Build GRCh38
rs979291000 Chr.19: 35123396-35123396 on Build GRCh38
rs111876747 Chr.19: 35123376-35123377 on Build GRCh38
rs760957927 Chr.19: 35123293-35123293 on Build GRCh38
rs374615281 Chr.19: 35123478-35123478 on Build GRCh38
rs911310014 Chr.19: 35123410-35123410 on Build GRCh38
rs771044986 Chr.19: 35123497-35123497 on Build GRCh38
rs764390697 Chr.19: 35123454-35123454 on Build GRCh38
rs904493252 Chr.19: 35123415-35123415 on Build GRCh38
rs764443826 Chr.19: 35123327-35123327 on Build GRCh38
rs781414043 Chr.19: 35123502-35123502 on Build GRCh38
rs946147879 Chr.19: 35123629-35123629 on Build GRCh38
rs543117183 Chr.19: 35123370-35123371 on Build GRCh38
rs1052400 Chr.19: 35123604-35123604 on Build GRCh38
rs201296423 Chr.19: 35123621-35123622 on Build GRCh38
rs961124869 Chr.19: 35123444-35123444 on Build GRCh38
rs1041198103 Chr.19: 35123599-35123599 on Build GRCh38
rs376261312 Chr.19: 35123573-35123573 on Build GRCh38
rs530242473 Chr.19: 35123343-35123343 on Build GRCh38
rs1672974 Chr.19: 35123432-35123432 on Build GRCh38
rs753099803 Chr.19: 35123225-35123225 on Build GRCh38
rs796872194 Chr.19: 35123368-35123370 on Build GRCh38
rs141165293 Chr.19: 35123246-35123246 on Build GRCh38
rs888643572 Chr.19: 35123452-35123452 on Build GRCh38
rs749253055 Chr.19: 35123360-35123360 on Build GRCh38
rs746414875 Chr.19: 35123215-35123215 on Build GRCh38
rs71167516 Chr.19: 35123349-35123350 on Build GRCh38
rs775905037 Chr.19: 35123437-35123437 on Build GRCh38
rs919559561 Chr.19: 35123239-35123239 on Build GRCh38
rs769904354 Chr.19: 35123264-35123264 on Build GRCh38
rs779716499 Chr.19: 35123481-35123481 on Build GRCh38
rs758701504 Chr.19: 35123324-35123324 on Build GRCh38
rs113610721 Chr.19: 35123310-35123310 on Build GRCh38
rs373271833 Chr.19: 35123509-35123509 on Build GRCh38
rs112343981 Chr.19: 35123355-35123356 on Build GRCh38
rs977176375 Chr.19: 35123647-35123647 on Build GRCh38
rs766157940 Chr.19: 35123342-35123343 on Build GRCh38
rs765800422 Chr.19: 35123304-35123304 on Build GRCh38
rs147918466 Chr.19: 35123351-35123352 on Build GRCh38
rs937089 Chr.19: 35123370-35123370 on Build GRCh38
rs780782970 Chr.19: 35123262-35123262 on Build GRCh38
rs747469345 Chr.19: 35123358-35123359 on Build GRCh38
rs1041186800 Chr.19: 35123325-35123325 on Build GRCh38
rs142898677 Chr.19: 35123301-35123301 on Build GRCh38
rs749166004 Chr.19: 35123275-35123275 on Build GRCh38
rs780198869 Chr.19: 35123354-35123354 on Build GRCh38
rs1023370052 Chr.19: 35123234-35123234 on Build GRCh38
rs776819772 Chr.19: 35123298-35123298 on Build GRCh38
rs537658851 Chr.19: 35123420-35123420 on Build GRCh38
rs567677161 Chr.19: 35123335-35123335 on Build GRCh38
rs749892028 Chr.19: 35123473-35123473 on Build GRCh38
rs565087214 Chr.19: 35123350-35123351 on Build GRCh38
rs567643585 Chr.19: 35123494-35123494 on Build GRCh38
rs1802802 Chr.19: 35123612-35123612 on Build GRCh38
rs149680357 Chr.19: 35123419-35123419 on Build GRCh38
rs182327242 Chr.19: 35123436-35123436 on Build GRCh38
rs967863016 Chr.19: 35123364-35123364 on Build GRCh38
rs370875872 Chr.19: 35123424-35123424 on Build GRCh38
rs766319137 Chr.19: 35123296-35123296 on Build GRCh38
rs374114255 Chr.19: 35123238-35123238 on Build GRCh38
rs746106514 Chr.19: 35123522-35123522 on Build GRCh38
rs746091219 Chr.19: 35123506-35123506 on Build GRCh38

View Full Product Details