Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35351443-35351933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FFAR1
Entrez Gene ID
2864
Gene Name
free fatty acid receptor 1
Gene Aliases
FFA1R, GPCR40, GPR40
UniGene
Hs.248127
Gene Chromosome Location
Chr.19: 35351542-35352464 on Build GRCh38
Transcripts
NM_005303.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1712162 c.357G>C p.R119S Chr.19: 35351908-35351908 on Build GRCh38
5706978 c.218C>T p.P73L Chr.19: 35351769-35351769 on Build GRCh38
3970919 c.361T>C p.C121R Chr.19: 35351912-35351912 on Build GRCh38
1392844 c.276C>A p.A92A Chr.19: 35351827-35351827 on Build GRCh38
4076994 c.338G>A p.G113D Chr.19: 35351889-35351889 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572675476 Chr.19: 35351821-35351821 on Build GRCh38
rs745577342 Chr.19: 35351701-35351701 on Build GRCh38
rs758874978 Chr.19: 35351669-35351669 on Build GRCh38
rs770590725 Chr.19: 35351563-35351563 on Build GRCh38
rs769406381 Chr.19: 35351716-35351716 on Build GRCh38
rs182374629 Chr.19: 35351482-35351482 on Build GRCh38
rs757549371 Chr.19: 35351692-35351692 on Build GRCh38
rs779719073 Chr.19: 35351791-35351791 on Build GRCh38
rs747746804 Chr.19: 35351596-35351596 on Build GRCh38
rs759823354 Chr.19: 35351654-35351654 on Build GRCh38
rs994395390 Chr.19: 35351545-35351545 on Build GRCh38
rs868171432 Chr.19: 35351726-35351726 on Build GRCh38
rs777330282 Chr.19: 35351677-35351677 on Build GRCh38
rs780726156 Chr.19: 35351788-35351788 on Build GRCh38
rs776224934 Chr.19: 35351566-35351566 on Build GRCh38
rs143820035 Chr.19: 35351861-35351861 on Build GRCh38
rs780470492 Chr.19: 35351462-35351462 on Build GRCh38
rs769938346 Chr.19: 35351822-35351822 on Build GRCh38
rs956591250 Chr.19: 35351903-35351903 on Build GRCh38
rs758935200 Chr.19: 35351514-35351514 on Build GRCh38
rs200829130 Chr.19: 35351889-35351889 on Build GRCh38
rs765530284 Chr.19: 35351759-35351759 on Build GRCh38
rs867765877 Chr.19: 35351639-35351639 on Build GRCh38
rs919482801 Chr.19: 35351792-35351792 on Build GRCh38
rs754058898 Chr.19: 35351643-35351643 on Build GRCh38
rs372221350 Chr.19: 35351781-35351781 on Build GRCh38
rs772053681 Chr.19: 35351745-35351745 on Build GRCh38
rs200173092 Chr.19: 35351661-35351661 on Build GRCh38
rs368886663 Chr.19: 35351671-35351671 on Build GRCh38
rs778529179 Chr.19: 35351885-35351885 on Build GRCh38
rs780407446 Chr.19: 35351486-35351486 on Build GRCh38
rs765475198 Chr.19: 35351660-35351660 on Build GRCh38
rs984383484 Chr.19: 35351831-35351831 on Build GRCh38
rs1040577492 Chr.19: 35351656-35351656 on Build GRCh38
rs773140188 Chr.19: 35351748-35351748 on Build GRCh38
rs375966117 Chr.19: 35351582-35351582 on Build GRCh38
rs75455784 Chr.19: 35351587-35351587 on Build GRCh38
rs774289904 Chr.19: 35351841-35351841 on Build GRCh38
rs368171672 Chr.19: 35351827-35351827 on Build GRCh38
rs753588393 Chr.19: 35351862-35351862 on Build GRCh38
rs761631316 Chr.19: 35351636-35351636 on Build GRCh38
rs1034662709 Chr.19: 35351893-35351893 on Build GRCh38
rs866089391 Chr.19: 35351714-35351714 on Build GRCh38
rs879039227 Chr.19: 35351504-35351504 on Build GRCh38
rs774976696 Chr.19: 35351594-35351594 on Build GRCh38
rs774038954 Chr.19: 35351634-35351634 on Build GRCh38
rs868401114 Chr.19: 35351708-35351708 on Build GRCh38
rs766715388 Chr.19: 35351641-35351641 on Build GRCh38
rs767964895 Chr.19: 35351844-35351844 on Build GRCh38
rs762954324 Chr.19: 35351828-35351828 on Build GRCh38
rs536139569 Chr.19: 35351655-35351655 on Build GRCh38
rs374114764 Chr.19: 35351539-35351539 on Build GRCh38
rs768508851 Chr.19: 35351732-35351732 on Build GRCh38
rs773186856 Chr.19: 35351806-35351806 on Build GRCh38
rs763045854 Chr.19: 35351763-35351763 on Build GRCh38
rs541591995 Chr.19: 35351845-35351845 on Build GRCh38
rs1000167582 Chr.19: 35351730-35351730 on Build GRCh38
rs780834782 Chr.19: 35351696-35351696 on Build GRCh38
rs981781717 Chr.19: 35351625-35351625 on Build GRCh38
rs138336486 Chr.19: 35351623-35351623 on Build GRCh38
rs959003243 Chr.19: 35351742-35351742 on Build GRCh38
rs762839741 Chr.19: 35351622-35351622 on Build GRCh38
rs865873341 Chr.19: 35351729-35351729 on Build GRCh38
rs943105017 Chr.19: 35351905-35351905 on Build GRCh38
rs148545938 Chr.19: 35351546-35351546 on Build GRCh38
rs762356918 Chr.19: 35351842-35351842 on Build GRCh38
rs750765669 Chr.19: 35351788-35351788 on Build GRCh38
rs746505975 Chr.19: 35351683-35351683 on Build GRCh38
rs375191494 Chr.19: 35351782-35351782 on Build GRCh38
rs764512579 Chr.19: 35351768-35351768 on Build GRCh38
rs756775454 Chr.19: 35351671-35351671 on Build GRCh38
rs777685773 Chr.19: 35351562-35351563 on Build GRCh38
rs369386290 Chr.19: 35351468-35351468 on Build GRCh38
rs774280566 Chr.19: 35351734-35351734 on Build GRCh38
rs770404949 Chr.19: 35351800-35351800 on Build GRCh38
rs903170523 Chr.19: 35351705-35351705 on Build GRCh38
rs778277031 Chr.19: 35351799-35351799 on Build GRCh38
rs779479930 Chr.19: 35351721-35351721 on Build GRCh38
rs772100372 Chr.19: 35351804-35351804 on Build GRCh38
rs765964806 Chr.19: 35351856-35351856 on Build GRCh38
rs748755425 Chr.19: 35351797-35351797 on Build GRCh38
rs748785019 Chr.19: 35351722-35351722 on Build GRCh38
rs1019708893 Chr.19: 35351469-35351469 on Build GRCh38
rs947544833 Chr.19: 35351704-35351704 on Build GRCh38
rs866102651 Chr.19: 35351777-35351777 on Build GRCh38
rs768158175 Chr.19: 35351798-35351798 on Build GRCh38
rs373180810 Chr.19: 35351914-35351914 on Build GRCh38
rs952318388 Chr.19: 35351824-35351824 on Build GRCh38
rs1048730488 Chr.19: 35351613-35351613 on Build GRCh38
rs761529668 Chr.19: 35351739-35351739 on Build GRCh38
rs1009896341 Chr.19: 35351667-35351667 on Build GRCh38
rs558342219 Chr.19: 35351897-35351897 on Build GRCh38
rs777810859 Chr.19: 35351898-35351898 on Build GRCh38
rs759873868 Chr.19: 35351754-35351754 on Build GRCh38
rs981729343 Chr.19: 35351883-35351883 on Build GRCh38

View Full Product Details