Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35352146-35352635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FFAR1
Entrez Gene ID
2864
Gene Name
free fatty acid receptor 1
Gene Aliases
FFA1R, GPCR40, GPR40
UniGene
Hs.248127
Gene Chromosome Location
Chr.19: 35351542-35352464 on Build GRCh38
Transcripts
NM_005303.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5804647 c.770G>T p.W257L Chr.19: 35352321-35352321 on Build GRCh38
1612069 c.818C>T p.P273L Chr.19: 35352369-35352369 on Build GRCh38
5792233 c.872C>T p.A291V Chr.19: 35352423-35352423 on Build GRCh38
158734 c.696G>A p.T232T Chr.19: 35352247-35352247 on Build GRCh38
1641129 c.603C>A p.C201* Chr.19: 35352154-35352154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767645426 Chr.19: 35352506-35352506 on Build GRCh38
rs775171662 Chr.19: 35352471-35352471 on Build GRCh38
rs780266784 Chr.19: 35352413-35352413 on Build GRCh38
rs766312605 Chr.19: 35352302-35352302 on Build GRCh38
rs767730676 Chr.19: 35352370-35352370 on Build GRCh38
rs866758889 Chr.19: 35352228-35352228 on Build GRCh38
rs772745297 Chr.19: 35352203-35352203 on Build GRCh38
rs747120023 Chr.19: 35352166-35352166 on Build GRCh38
rs41275830 Chr.19: 35352468-35352468 on Build GRCh38
rs543539181 Chr.19: 35352438-35352438 on Build GRCh38
rs376911636 Chr.19: 35352529-35352529 on Build GRCh38
rs1006412186 Chr.19: 35352355-35352355 on Build GRCh38
rs907246345 Chr.19: 35352306-35352306 on Build GRCh38
rs991130581 Chr.19: 35352495-35352495 on Build GRCh38
rs755476796 Chr.19: 35352421-35352421 on Build GRCh38
rs538021112 Chr.19: 35352609-35352609 on Build GRCh38
rs749630680 Chr.19: 35352174-35352174 on Build GRCh38
rs376074468 Chr.19: 35352212-35352212 on Build GRCh38
rs1021941516 Chr.19: 35352432-35352432 on Build GRCh38
rs2301151 Chr.19: 35352183-35352183 on Build GRCh38
rs975749544 Chr.19: 35352430-35352430 on Build GRCh38
rs1030411793 Chr.19: 35352440-35352440 on Build GRCh38
rs772617515 Chr.19: 35352285-35352285 on Build GRCh38
rs748492841 Chr.19: 35352436-35352436 on Build GRCh38
rs74530614 Chr.19: 35352444-35352444 on Build GRCh38
rs977361371 Chr.19: 35352507-35352507 on Build GRCh38
rs762769438 Chr.19: 35352486-35352486 on Build GRCh38
rs746431371 Chr.19: 35352169-35352169 on Build GRCh38
rs1047527532 Chr.19: 35352266-35352266 on Build GRCh38
rs780640960 Chr.19: 35352229-35352229 on Build GRCh38
rs374423505 Chr.19: 35352223-35352223 on Build GRCh38
rs776047449 Chr.19: 35352171-35352171 on Build GRCh38
rs1045143569 Chr.19: 35352315-35352315 on Build GRCh38
rs749683890 Chr.19: 35352233-35352233 on Build GRCh38
rs993550748 Chr.19: 35352308-35352308 on Build GRCh38
rs542569546 Chr.19: 35352593-35352593 on Build GRCh38
rs746539345 Chr.19: 35352447-35352447 on Build GRCh38
rs747794205 Chr.19: 35352274-35352274 on Build GRCh38
rs757499169 Chr.19: 35352218-35352218 on Build GRCh38
rs768405979 Chr.19: 35352293-35352293 on Build GRCh38
rs756710530 Chr.19: 35352224-35352224 on Build GRCh38
rs755702794 Chr.19: 35352466-35352466 on Build GRCh38
rs578146950 Chr.19: 35352323-35352323 on Build GRCh38
rs781171288 Chr.19: 35352222-35352222 on Build GRCh38
rs779191290 Chr.19: 35352422-35352422 on Build GRCh38
rs766999539 Chr.19: 35352195-35352195 on Build GRCh38
rs574019963 Chr.19: 35352204-35352204 on Build GRCh38
rs770395673 Chr.19: 35352170-35352170 on Build GRCh38
rs865840644 Chr.19: 35352335-35352335 on Build GRCh38
rs965347513 Chr.19: 35352389-35352389 on Build GRCh38
rs147071299 Chr.19: 35352288-35352288 on Build GRCh38
rs750562924 Chr.19: 35352379-35352379 on Build GRCh38
rs553007692 Chr.19: 35352168-35352168 on Build GRCh38
rs35923096 Chr.19: 35352494-35352495 on Build GRCh38
rs371520895 Chr.19: 35352343-35352343 on Build GRCh38
rs976225990 Chr.19: 35352199-35352199 on Build GRCh38
rs949331866 Chr.19: 35352534-35352534 on Build GRCh38
rs748369472 Chr.19: 35352484-35352492 on Build GRCh38
rs1017887146 Chr.19: 35352385-35352385 on Build GRCh38
rs904995059 Chr.19: 35352320-35352320 on Build GRCh38
rs761512453 Chr.19: 35352187-35352187 on Build GRCh38
rs929393185 Chr.19: 35352602-35352602 on Build GRCh38
rs753667981 Chr.19: 35352214-35352214 on Build GRCh38
rs1026433053 Chr.19: 35352339-35352339 on Build GRCh38
rs1045699787 Chr.19: 35352575-35352575 on Build GRCh38
rs780428323 Chr.19: 35352330-35352330 on Build GRCh38
rs754705434 Chr.19: 35352216-35352216 on Build GRCh38
rs756203531 Chr.19: 35352396-35352396 on Build GRCh38
rs1029894009 Chr.19: 35352528-35352528 on Build GRCh38
rs988192134 Chr.19: 35352467-35352467 on Build GRCh38
rs865782329 Chr.19: 35352500-35352500 on Build GRCh38
rs985892634 Chr.19: 35352188-35352188 on Build GRCh38
rs776117423 Chr.19: 35352466-35352466 on Build GRCh38
rs929988063 Chr.19: 35352249-35352249 on Build GRCh38
rs372542737 Chr.19: 35352492-35352492 on Build GRCh38
rs757472012 Chr.19: 35352361-35352361 on Build GRCh38
rs770861016 Chr.19: 35352167-35352167 on Build GRCh38
rs774772392 Chr.19: 35352247-35352247 on Build GRCh38
rs763046473 Chr.19: 35352455-35352455 on Build GRCh38
rs765011486 Chr.19: 35352331-35352331 on Build GRCh38
rs201401516 Chr.19: 35352217-35352217 on Build GRCh38
rs894448487 Chr.19: 35352255-35352255 on Build GRCh38
rs776496904 Chr.19: 35352314-35352314 on Build GRCh38
rs376032484 Chr.19: 35352277-35352277 on Build GRCh38
rs768427411 Chr.19: 35352180-35352180 on Build GRCh38
rs749874342 Chr.19: 35352417-35352417 on Build GRCh38
rs769199724 Chr.19: 35352237-35352237 on Build GRCh38

View Full Product Details