Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35554843-35555334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP4A
Entrez Gene ID
495
Gene Name
ATPase H+/K+ transporting alpha subunit
Gene Aliases
ATP6A
UniGene
Hs.36992
Gene Chromosome Location
Chr.19: 35550193-35563658 on Build GRCh38
Transcripts
NM_000704.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6084526 c.2214G>A p.K738K Chr.19: 35555278-35555278 on Build GRCh38
4843395 c.2379G>A p.K793K Chr.19: 35555024-35555024 on Build GRCh38
6607913 c.2453T>C p.I818T Chr.19: 35554950-35554950 on Build GRCh38
5616968 c.2223C>T p.I741I Chr.19: 35555269-35555269 on Build GRCh38
3198057 c.2179G>A p.G727R Chr.19: 35555313-35555313 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141280378 Chr.19: 35554979-35554979 on Build GRCh38
rs992593504 Chr.19: 35554903-35554903 on Build GRCh38
rs576925308 Chr.19: 35554940-35554940 on Build GRCh38
rs375301898 Chr.19: 35554984-35554984 on Build GRCh38
rs749693841 Chr.19: 35555313-35555313 on Build GRCh38
rs761437112 Chr.19: 35555175-35555175 on Build GRCh38
rs142843033 Chr.19: 35554963-35554963 on Build GRCh38
rs778443222 Chr.19: 35554978-35554978 on Build GRCh38
rs940519441 Chr.19: 35554915-35554915 on Build GRCh38
rs961811189 Chr.19: 35555164-35555164 on Build GRCh38
rs777249685 Chr.19: 35555146-35555146 on Build GRCh38
rs770151145 Chr.19: 35555152-35555152 on Build GRCh38
rs775868034 Chr.19: 35555291-35555291 on Build GRCh38
rs780872582 Chr.19: 35554976-35554976 on Build GRCh38
rs376393633 Chr.19: 35555154-35555154 on Build GRCh38
rs931919781 Chr.19: 35555260-35555260 on Build GRCh38
rs768875718 Chr.19: 35554873-35554873 on Build GRCh38
rs907599890 Chr.19: 35554906-35554906 on Build GRCh38
rs748375907 Chr.19: 35554995-35554995 on Build GRCh38
rs761106663 Chr.19: 35555032-35555032 on Build GRCh38
rs779298671 Chr.19: 35555146-35555146 on Build GRCh38
rs541790101 Chr.19: 35554985-35554985 on Build GRCh38
rs750306708 Chr.19: 35555094-35555094 on Build GRCh38
rs746402089 Chr.19: 35555271-35555271 on Build GRCh38
rs773923127 Chr.19: 35554899-35554899 on Build GRCh38
rs766665156 Chr.19: 35554916-35554916 on Build GRCh38
rs766299190 Chr.19: 35555197-35555197 on Build GRCh38
rs370361513 Chr.19: 35555162-35555162 on Build GRCh38
rs770659863 Chr.19: 35555268-35555268 on Build GRCh38
rs746796959 Chr.19: 35555256-35555256 on Build GRCh38
rs1041113616 Chr.19: 35555030-35555030 on Build GRCh38
rs1031210028 Chr.19: 35555003-35555003 on Build GRCh38
rs1045465356 Chr.19: 35555172-35555172 on Build GRCh38
rs146535206 Chr.19: 35555269-35555269 on Build GRCh38
rs777723789 Chr.19: 35555043-35555043 on Build GRCh38
rs776970403 Chr.19: 35555062-35555062 on Build GRCh38
rs779926969 Chr.19: 35555112-35555112 on Build GRCh38
rs757964467 Chr.19: 35554961-35554961 on Build GRCh38
rs150844981 Chr.19: 35555027-35555027 on Build GRCh38
rs763697225 Chr.19: 35555161-35555161 on Build GRCh38
rs201082348 Chr.19: 35555272-35555272 on Build GRCh38
rs747181945 Chr.19: 35555210-35555210 on Build GRCh38
rs768987341 Chr.19: 35555314-35555314 on Build GRCh38
rs772282579 Chr.19: 35555005-35555005 on Build GRCh38
rs754178516 Chr.19: 35554925-35554925 on Build GRCh38
rs750377482 Chr.19: 35555177-35555177 on Build GRCh38
rs373242882 Chr.19: 35555135-35555135 on Build GRCh38
rs765503046 Chr.19: 35555069-35555069 on Build GRCh38
rs753645038 Chr.19: 35555203-35555203 on Build GRCh38
rs774606141 Chr.19: 35554877-35554877 on Build GRCh38
rs774113274 Chr.19: 35555315-35555315 on Build GRCh38
rs199650617 Chr.19: 35555155-35555155 on Build GRCh38
rs766862710 Chr.19: 35554873-35554876 on Build GRCh38
rs372209900 Chr.19: 35554946-35554946 on Build GRCh38
rs930609548 Chr.19: 35554885-35554885 on Build GRCh38
rs770891533 Chr.19: 35555141-35555141 on Build GRCh38
rs1001775978 Chr.19: 35555133-35555133 on Build GRCh38
rs778185949 Chr.19: 35555209-35555209 on Build GRCh38
rs757172998 Chr.19: 35555084-35555084 on Build GRCh38
rs762376654 Chr.19: 35555121-35555121 on Build GRCh38
rs369099819 Chr.19: 35555073-35555073 on Build GRCh38
rs898999962 Chr.19: 35554874-35554874 on Build GRCh38
rs368295347 Chr.19: 35554876-35554876 on Build GRCh38
rs2733739 Chr.19: 35554870-35554870 on Build GRCh38
rs781230902 Chr.19: 35555089-35555089 on Build GRCh38
rs61746955 Chr.19: 35554973-35554973 on Build GRCh38
rs752417012 Chr.19: 35555072-35555072 on Build GRCh38
rs147416424 Chr.19: 35554964-35554964 on Build GRCh38
rs755944544 Chr.19: 35555188-35555188 on Build GRCh38
rs772264272 Chr.19: 35554891-35554891 on Build GRCh38
rs761368270 Chr.19: 35555316-35555316 on Build GRCh38
rs754394674 Chr.19: 35555069-35555069 on Build GRCh38
rs754784580 Chr.19: 35555204-35555204 on Build GRCh38
rs200604186 Chr.19: 35555125-35555125 on Build GRCh38
rs748394516 Chr.19: 35555114-35555114 on Build GRCh38
rs986024114 Chr.19: 35555266-35555266 on Build GRCh38
rs756003053 Chr.19: 35555096-35555096 on Build GRCh38
rs761195011 Chr.19: 35554909-35554909 on Build GRCh38
rs763611116 Chr.19: 35555077-35555077 on Build GRCh38
rs973681577 Chr.19: 35555217-35555217 on Build GRCh38
rs532736050 Chr.19: 35555176-35555176 on Build GRCh38
rs751024992 Chr.19: 35555081-35555081 on Build GRCh38
rs755307645 Chr.19: 35555132-35555132 on Build GRCh38
rs940816969 Chr.19: 35555103-35555103 on Build GRCh38
rs150792532 Chr.19: 35555063-35555063 on Build GRCh38
rs778026828 Chr.19: 35555129-35555129 on Build GRCh38
rs771425685 Chr.19: 35555054-35555054 on Build GRCh38
rs759139986 Chr.19: 35554934-35554934 on Build GRCh38
rs762024796 Chr.19: 35554882-35554882 on Build GRCh38

View Full Product Details