Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35555486-35555926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP4A
Entrez Gene ID
495
Gene Name
ATPase H+/K+ transporting alpha subunit
Gene Aliases
ATP6A
UniGene
Hs.36992
Gene Chromosome Location
Chr.19: 35550193-35563658 on Build GRCh38
Transcripts
NM_000704.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6607918 c.1928G>A p.G643D Chr.19: 35555754-35555754 on Build GRCh38
4565341 c.1992_1993CC>TT p.Q665* Chr.19: 35555689-35555690 on Build GRCh38
3404134 c.1948G>A p.E650K Chr.19: 35555734-35555734 on Build GRCh38
4580959 c.2080C>T p.R694C Chr.19: 35555517-35555517 on Build GRCh38
3198060 c.2060C>T p.S687L Chr.19: 35555537-35555537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749198983 Chr.19: 35555634-35555634 on Build GRCh38
rs766625990 Chr.19: 35555594-35555594 on Build GRCh38
rs371581853 Chr.19: 35555626-35555626 on Build GRCh38
rs780882384 Chr.19: 35555561-35555561 on Build GRCh38
rs769329382 Chr.19: 35555574-35555574 on Build GRCh38
rs372060721 Chr.19: 35555780-35555780 on Build GRCh38
rs760821367 Chr.19: 35555512-35555512 on Build GRCh38
rs867542713 Chr.19: 35555585-35555585 on Build GRCh38
rs780708990 Chr.19: 35555593-35555593 on Build GRCh38
rs80198561 Chr.19: 35555665-35555665 on Build GRCh38
rs753426264 Chr.19: 35555735-35555735 on Build GRCh38
rs764843139 Chr.19: 35555873-35555873 on Build GRCh38
rs570707673 Chr.19: 35555874-35555874 on Build GRCh38
rs61736770 Chr.19: 35555696-35555696 on Build GRCh38
rs147129403 Chr.19: 35555612-35555612 on Build GRCh38
rs778214581 Chr.19: 35555538-35555538 on Build GRCh38
rs570878591 Chr.19: 35555624-35555624 on Build GRCh38
rs768008534 Chr.19: 35555829-35555829 on Build GRCh38
rs759588356 Chr.19: 35555605-35555605 on Build GRCh38
rs749245477 Chr.19: 35555582-35555582 on Build GRCh38
rs150964145 Chr.19: 35555527-35555527 on Build GRCh38
rs1008984874 Chr.19: 35555529-35555529 on Build GRCh38
rs767990060 Chr.19: 35555710-35555710 on Build GRCh38
rs373762221 Chr.19: 35555614-35555614 on Build GRCh38
rs145767701 Chr.19: 35555679-35555679 on Build GRCh38
rs752094325 Chr.19: 35555703-35555703 on Build GRCh38
rs1051070657 Chr.19: 35555520-35555520 on Build GRCh38
rs200522604 Chr.19: 35555506-35555506 on Build GRCh38
rs770021144 Chr.19: 35555795-35555795 on Build GRCh38
rs748068530 Chr.19: 35555777-35555777 on Build GRCh38
rs1023068872 Chr.19: 35555798-35555798 on Build GRCh38
rs754606914 Chr.19: 35555825-35555825 on Build GRCh38
rs368721895 Chr.19: 35555728-35555728 on Build GRCh38
rs888887677 Chr.19: 35555770-35555770 on Build GRCh38
rs149961054 Chr.19: 35555517-35555517 on Build GRCh38
rs75318424 Chr.19: 35555625-35555625 on Build GRCh38
rs1028106478 Chr.19: 35555755-35555755 on Build GRCh38
rs745415480 Chr.19: 35555567-35555567 on Build GRCh38
rs138654635 Chr.19: 35555680-35555680 on Build GRCh38
rs768595874 Chr.19: 35555653-35555653 on Build GRCh38
rs774239291 Chr.19: 35555654-35555654 on Build GRCh38
rs201830604 Chr.19: 35555717-35555717 on Build GRCh38
rs775347977 Chr.19: 35555578-35555578 on Build GRCh38
rs370554663 Chr.19: 35555670-35555670 on Build GRCh38
rs779336531 Chr.19: 35555526-35555526 on Build GRCh38
rs779455265 Chr.19: 35555734-35555734 on Build GRCh38
rs560720583 Chr.19: 35555611-35555611 on Build GRCh38
rs769485930 Chr.19: 35555663-35555664 on Build GRCh38
rs1020876573 Chr.19: 35555895-35555895 on Build GRCh38
rs375470351 Chr.19: 35555817-35555817 on Build GRCh38
rs527680506 Chr.19: 35555851-35555851 on Build GRCh38
rs766467929 Chr.19: 35555513-35555513 on Build GRCh38
rs61729956 Chr.19: 35555558-35555558 on Build GRCh38
rs267605431 Chr.19: 35555692-35555692 on Build GRCh38
rs757758386 Chr.19: 35555704-35555704 on Build GRCh38
rs539328184 Chr.19: 35555884-35555884 on Build GRCh38
rs778879380 Chr.19: 35555773-35555773 on Build GRCh38
rs1007249420 Chr.19: 35555778-35555778 on Build GRCh38
rs755734977 Chr.19: 35555726-35555726 on Build GRCh38
rs981302194 Chr.19: 35555896-35555896 on Build GRCh38
rs199693426 Chr.19: 35555535-35555535 on Build GRCh38
rs775065728 Chr.19: 35555824-35555824 on Build GRCh38
rs746669377 Chr.19: 35555546-35555546 on Build GRCh38
rs1014497874 Chr.19: 35555539-35555539 on Build GRCh38
rs776811961 Chr.19: 35555603-35555603 on Build GRCh38
rs571661299 Chr.19: 35555584-35555584 on Build GRCh38
rs775546496 Chr.19: 35555814-35555814 on Build GRCh38
rs755053833 Chr.19: 35555524-35555524 on Build GRCh38
rs267605430 Chr.19: 35555590-35555590 on Build GRCh38
rs967405578 Chr.19: 35555657-35555657 on Build GRCh38
rs561359329 Chr.19: 35555583-35555583 on Build GRCh38
rs989838077 Chr.19: 35555661-35555661 on Build GRCh38
rs745362587 Chr.19: 35555611-35555611 on Build GRCh38
rs763145432 Chr.19: 35555815-35555815 on Build GRCh38
rs898108443 Chr.19: 35555709-35555709 on Build GRCh38
rs547752227 Chr.19: 35555866-35555866 on Build GRCh38
rs750752202 Chr.19: 35555848-35555848 on Build GRCh38
rs763403043 Chr.19: 35555687-35555687 on Build GRCh38
rs779551044 Chr.19: 35555631-35555631 on Build GRCh38
rs760913997 Chr.19: 35555849-35555849 on Build GRCh38
rs1036467168 Chr.19: 35555547-35555547 on Build GRCh38
rs753419217 Chr.19: 35555864-35555864 on Build GRCh38
rs763936814 Chr.19: 35555505-35555505 on Build GRCh38
rs904563816 Chr.19: 35555516-35555516 on Build GRCh38
rs751936807 Chr.19: 35555615-35555615 on Build GRCh38
rs746098310 Chr.19: 35555786-35555786 on Build GRCh38
rs777563260 Chr.19: 35555783-35555783 on Build GRCh38
rs150371575 Chr.19: 35555745-35555745 on Build GRCh38
rs896110909 Chr.19: 35555536-35555536 on Build GRCh38
rs775544857 Chr.19: 35555664-35555664 on Build GRCh38

View Full Product Details