Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35754863-35755370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPB6
Entrez Gene ID
126393
Gene Name
heat shock protein family B (small) member 6
Gene Aliases
HEL55, Hsp20, PPP1R91
UniGene
Hs.534538
Gene Chromosome Location
Chr.19: 35754566-35757029 on Build GRCh38
Transcripts
NM_144617.2

SNP Detail

SNP ID Chromosome Location
rs149052927 Chr.19: 35755331-35755331 on Build GRCh38
rs889385354 Chr.19: 35755136-35755136 on Build GRCh38
rs942433916 Chr.19: 35754898-35754898 on Build GRCh38
rs796908971 Chr.19: 35754925-35754925 on Build GRCh38
rs997129988 Chr.19: 35754954-35754954 on Build GRCh38
rs932318411 Chr.19: 35755064-35755064 on Build GRCh38
rs951611601 Chr.19: 35755321-35755321 on Build GRCh38
rs866994188 Chr.19: 35754894-35754894 on Build GRCh38
rs1050660947 Chr.19: 35755100-35755100 on Build GRCh38
rs985127967 Chr.19: 35755091-35755091 on Build GRCh38
rs1003786289 Chr.19: 35755299-35755299 on Build GRCh38
rs906552073 Chr.19: 35755279-35755279 on Build GRCh38
rs765719966 Chr.19: 35754893-35754893 on Build GRCh38
rs867395643 Chr.19: 35754931-35754931 on Build GRCh38
rs1029112773 Chr.19: 35755061-35755061 on Build GRCh38
rs540472813 Chr.19: 35755036-35755037 on Build GRCh38
rs137940999 Chr.19: 35755101-35755101 on Build GRCh38
rs953280226 Chr.19: 35755085-35755085 on Build GRCh38
rs775220917 Chr.19: 35754910-35754910 on Build GRCh38
rs924341726 Chr.19: 35754985-35754985 on Build GRCh38
rs893859924 Chr.19: 35755236-35755236 on Build GRCh38
rs190135278 Chr.19: 35755271-35755271 on Build GRCh38
rs1012322839 Chr.19: 35755249-35755249 on Build GRCh38
rs1022857093 Chr.19: 35755327-35755327 on Build GRCh38
rs762629966 Chr.19: 35755343-35755343 on Build GRCh38
rs370292442 Chr.19: 35755322-35755322 on Build GRCh38
rs1026288991 Chr.19: 35755317-35755317 on Build GRCh38
rs571906073 Chr.19: 35755058-35755058 on Build GRCh38
rs903111498 Chr.19: 35755157-35755157 on Build GRCh38
rs1054221925 Chr.19: 35755235-35755235 on Build GRCh38
rs554528232 Chr.19: 35755288-35755288 on Build GRCh38
rs935814933 Chr.19: 35755195-35755195 on Build GRCh38
rs897910588 Chr.19: 35755263-35755264 on Build GRCh38
rs952038875 Chr.19: 35755304-35755304 on Build GRCh38
rs991915216 Chr.19: 35755309-35755309 on Build GRCh38
rs1008991593 Chr.19: 35755190-35755190 on Build GRCh38
rs762851078 Chr.19: 35754937-35754937 on Build GRCh38
rs550100049 Chr.19: 35755026-35755026 on Build GRCh38
rs933903307 Chr.19: 35754936-35754936 on Build GRCh38
rs1054529461 Chr.19: 35755056-35755056 on Build GRCh38

View Full Product Details