Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37344531-37345044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HKR1
Entrez Gene ID
284459
Gene Name
HKR1, GLI-Kruppel zinc finger family member
Gene Aliases
ZNF875
UniGene
Hs.657027
Gene Chromosome Location
Chr.19: 37317900-37364455 on Build GRCh38
Transcripts
NM_181786.2 XM_017026672.1 XM_017026673.1 XM_017026674.1 XM_017026675.1 XM_017026676.1 XM_017026677.1 XM_017026678.1 XM_017026679.1 XM_017026680.1 XM_017026681.1 XM_017026682.1 XM_017026683.1 XM_017026684.1 XM_017026685.1 XM_017026686.1 XM_017026687.1 XM_017026688.1 XM_017026689.1 XM_017026690.1 XM_017026691.1 XM_017026692.1 XM_017026693.1 XM_017026694.1 XM_017026695.1 XM_017026696.1 XM_017026697.1 XM_017026698.1 XM_017026699.1 XM_017026700.1 XM_017026701.1 XM_017026702.1 XM_017026703.1 XM_017026704.1 XM_017026706.1 XM_017026707.1 XM_017026708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
123609 c.53G>C p.R18T Chr.19: 37344737-37344737 on Build GRCh38
6441030 c.69T>G p.C23W Chr.19: 37344753-37344753 on Build GRCh38
6427016 c.23C>A p.S8Y Chr.19: 37344707-37344707 on Build GRCh38
6034547 c.80G>A p.G27E Chr.19: 37344764-37344764 on Build GRCh38
71076 c.3G>T p.M1I Chr.19: 37344687-37344687 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768777642 Chr.19: 37344979-37344979 on Build GRCh38
rs190214110 Chr.19: 37345003-37345003 on Build GRCh38
rs1020791590 Chr.19: 37344876-37344876 on Build GRCh38
rs768991575 Chr.19: 37344744-37344744 on Build GRCh38
rs753740155 Chr.19: 37344692-37344692 on Build GRCh38
rs756908992 Chr.19: 37344721-37344721 on Build GRCh38
rs2305115 Chr.19: 37344756-37344756 on Build GRCh38
rs139271093 Chr.19: 37344739-37344739 on Build GRCh38
rs141591406 Chr.19: 37344999-37344999 on Build GRCh38
rs759376920 Chr.19: 37344696-37344696 on Build GRCh38
rs542031923 Chr.19: 37344774-37344774 on Build GRCh38
rs181114217 Chr.19: 37344827-37344827 on Build GRCh38
rs747227334 Chr.19: 37344746-37344750 on Build GRCh38
rs751221765 Chr.19: 37344669-37344669 on Build GRCh38
rs777105569 Chr.19: 37344712-37344712 on Build GRCh38
rs752668786 Chr.19: 37344710-37344710 on Build GRCh38
rs1024085420 Chr.19: 37344786-37344786 on Build GRCh38
rs925915176 Chr.19: 37344573-37344573 on Build GRCh38
rs970191779 Chr.19: 37344789-37344789 on Build GRCh38
rs755338258 Chr.19: 37344828-37344828 on Build GRCh38
rs920059800 Chr.19: 37344858-37344858 on Build GRCh38
rs1045806471 Chr.19: 37344942-37344942 on Build GRCh38
rs749716233 Chr.19: 37344743-37344743 on Build GRCh38
rs930227377 Chr.19: 37344862-37344862 on Build GRCh38
rs934080706 Chr.19: 37344632-37344632 on Build GRCh38
rs116402786 Chr.19: 37344766-37344766 on Build GRCh38
rs767212237 Chr.19: 37344822-37344822 on Build GRCh38
rs773484901 Chr.19: 37344641-37344641 on Build GRCh38
rs1020188397 Chr.19: 37345016-37345016 on Build GRCh38
rs373423139 Chr.19: 37344820-37344820 on Build GRCh38
rs566183834 Chr.19: 37344890-37344890 on Build GRCh38
rs762508577 Chr.19: 37344919-37344919 on Build GRCh38
rs771985100 Chr.19: 37344636-37344636 on Build GRCh38
rs748715498 Chr.19: 37344831-37344831 on Build GRCh38
rs139866643 Chr.19: 37344812-37344812 on Build GRCh38
rs758880987 Chr.19: 37344841-37344841 on Build GRCh38
rs567330497 Chr.19: 37344882-37344882 on Build GRCh38
rs760770927 Chr.19: 37344657-37344657 on Build GRCh38
rs1032809582 Chr.19: 37344981-37344981 on Build GRCh38
rs186060791 Chr.19: 37344846-37344846 on Build GRCh38
rs771330202 Chr.19: 37344755-37344755 on Build GRCh38
rs759462491 Chr.19: 37344780-37344780 on Build GRCh38
rs34322506 Chr.19: 37344631-37344632 on Build GRCh38
rs757941352 Chr.19: 37344711-37344711 on Build GRCh38
rs1000604727 Chr.19: 37344910-37344910 on Build GRCh38
rs889965403 Chr.19: 37344693-37344693 on Build GRCh38
rs913200868 Chr.19: 37344621-37344621 on Build GRCh38
rs777483166 Chr.19: 37344697-37344698 on Build GRCh38
rs748585314 Chr.19: 37344755-37344755 on Build GRCh38
rs751071684 Chr.19: 37344817-37344817 on Build GRCh38
rs542642959 Chr.19: 37344595-37344595 on Build GRCh38
rs765333386 Chr.19: 37344704-37344704 on Build GRCh38
rs957292260 Chr.19: 37344826-37344826 on Build GRCh38
rs528780940 Chr.19: 37344848-37344848 on Build GRCh38
rs898789140 Chr.19: 37344716-37344716 on Build GRCh38
rs62109240 Chr.19: 37344640-37344640 on Build GRCh38
rs749822463 Chr.19: 37344897-37344897 on Build GRCh38
rs766221969 Chr.19: 37344690-37344690 on Build GRCh38
rs964925061 Chr.19: 37344760-37344760 on Build GRCh38
rs143856884 Chr.19: 37344735-37344735 on Build GRCh38
rs776989432 Chr.19: 37344775-37344775 on Build GRCh38
rs79386354 Chr.19: 37344871-37344871 on Build GRCh38
rs762749324 Chr.19: 37344804-37344804 on Build GRCh38
rs1038570885 Chr.19: 37344850-37344850 on Build GRCh38
rs751245115 Chr.19: 37344717-37344717 on Build GRCh38
rs933966137 Chr.19: 37344843-37344843 on Build GRCh38
rs935916120 Chr.19: 37344583-37344583 on Build GRCh38
rs761935051 Chr.19: 37344634-37344634 on Build GRCh38
rs765067932 Chr.19: 37344787-37344787 on Build GRCh38
rs547557353 Chr.19: 37344845-37344845 on Build GRCh38
rs1017212019 Chr.19: 37344958-37344958 on Build GRCh38
rs374880884 Chr.19: 37344749-37344749 on Build GRCh38
rs752687215 Chr.19: 37344792-37344792 on Build GRCh38
rs905645698 Chr.19: 37344971-37344971 on Build GRCh38
rs746198897 Chr.19: 37344773-37344773 on Build GRCh38
rs1038927833 Chr.19: 37344715-37344715 on Build GRCh38

View Full Product Details