Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37347697-37348192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HKR1
Entrez Gene ID
284459
Gene Name
HKR1, GLI-Kruppel zinc finger family member
Gene Aliases
ZNF875
UniGene
Hs.657027
Gene Chromosome Location
Chr.19: 37317900-37364455 on Build GRCh38
Transcripts
NM_181786.2 XM_017026672.1 XM_017026673.1 XM_017026674.1 XM_017026675.1 XM_017026676.1 XM_017026677.1 XM_017026678.1 XM_017026679.1 XM_017026680.1 XM_017026681.1 XM_017026682.1 XM_017026683.1 XM_017026684.1 XM_017026685.1 XM_017026686.1 XM_017026687.1 XM_017026688.1 XM_017026689.1 XM_017026690.1 XM_017026691.1 XM_017026692.1 XM_017026693.1 XM_017026694.1 XM_017026695.1 XM_017026696.1 XM_017026697.1 XM_017026698.1 XM_017026699.1 XM_017026700.1 XM_017026701.1 XM_017026702.1 XM_017026703.1 XM_017026704.1 XM_017026706.1 XM_017026707.1 XM_017026708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3822870 c.287G>T p.R96I Chr.19: 37347846-37347846 on Build GRCh38
3822871 c.230G>T p.R77I Chr.19: 37347846-37347846 on Build GRCh38
6557820 c.264G>T p.G88G Chr.19: 37347823-37347823 on Build GRCh38
3533143 c.207G>A p.G69G Chr.19: 37347823-37347823 on Build GRCh38
4282296 c.213G>A p.A71A Chr.19: 37347829-37347829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764516411 Chr.19: 37347774-37347775 on Build GRCh38
rs369012450 Chr.19: 37347794-37347794 on Build GRCh38
rs765910142 Chr.19: 37347829-37347829 on Build GRCh38
rs1034874492 Chr.19: 37347925-37347925 on Build GRCh38
rs907059721 Chr.19: 37348083-37348083 on Build GRCh38
rs1049243381 Chr.19: 37348105-37348105 on Build GRCh38
rs772648218 Chr.19: 37347737-37347737 on Build GRCh38
rs752862578 Chr.19: 37347850-37347850 on Build GRCh38
rs888009034 Chr.19: 37347857-37347857 on Build GRCh38
rs374229854 Chr.19: 37347822-37347822 on Build GRCh38
rs748006800 Chr.19: 37347753-37347753 on Build GRCh38
rs765966427 Chr.19: 37347897-37347903 on Build GRCh38
rs954324297 Chr.19: 37347879-37347879 on Build GRCh38
rs915262547 Chr.19: 37347762-37347762 on Build GRCh38
rs748905319 Chr.19: 37347909-37347909 on Build GRCh38
rs555477570 Chr.19: 37348014-37348014 on Build GRCh38
rs940086053 Chr.19: 37347916-37347916 on Build GRCh38
rs775286986 Chr.19: 37347904-37347904 on Build GRCh38
rs972833700 Chr.19: 37347920-37347920 on Build GRCh38
rs1008506946 Chr.19: 37348160-37348160 on Build GRCh38
rs377635462 Chr.19: 37347883-37347883 on Build GRCh38
rs777705309 Chr.19: 37347769-37347769 on Build GRCh38
rs745439766 Chr.19: 37347897-37347897 on Build GRCh38
rs765557594 Chr.19: 37347828-37347828 on Build GRCh38
rs534422645 Chr.19: 37347770-37347770 on Build GRCh38
rs999114457 Chr.19: 37348088-37348088 on Build GRCh38
rs867674145 Chr.19: 37348013-37348013 on Build GRCh38
rs139266128 Chr.19: 37347811-37347811 on Build GRCh38
rs370613197 Chr.19: 37347749-37347749 on Build GRCh38
rs753992404 Chr.19: 37347821-37347821 on Build GRCh38
rs976543819 Chr.19: 37348066-37348066 on Build GRCh38
rs369600300 Chr.19: 37347896-37347896 on Build GRCh38
rs966162014 Chr.19: 37348065-37348065 on Build GRCh38
rs773757893 Chr.19: 37347919-37347919 on Build GRCh38
rs780137346 Chr.19: 37347736-37347736 on Build GRCh38
rs998645780 Chr.19: 37347817-37347817 on Build GRCh38
rs780984915 Chr.19: 37347890-37347890 on Build GRCh38
rs756262724 Chr.19: 37347728-37347728 on Build GRCh38
rs60405678 Chr.19: 37347989-37347989 on Build GRCh38
rs746972248 Chr.19: 37347867-37347867 on Build GRCh38
rs1373991 Chr.19: 37348009-37348009 on Build GRCh38
rs757270217 Chr.19: 37347888-37347888 on Build GRCh38
rs940160711 Chr.19: 37347743-37347743 on Build GRCh38
rs894357349 Chr.19: 37348000-37348000 on Build GRCh38
rs909495851 Chr.19: 37348124-37348124 on Build GRCh38
rs923898993 Chr.19: 37348021-37348021 on Build GRCh38
rs771845395 Chr.19: 37347754-37347754 on Build GRCh38
rs544328921 Chr.19: 37348161-37348161 on Build GRCh38
rs954434726 Chr.19: 37348153-37348153 on Build GRCh38
rs776159140 Chr.19: 37347786-37347786 on Build GRCh38
rs372968263 Chr.19: 37347799-37347799 on Build GRCh38
rs112714662 Chr.19: 37347735-37347735 on Build GRCh38
rs765657216 Chr.19: 37347971-37347971 on Build GRCh38
rs932159492 Chr.19: 37348104-37348104 on Build GRCh38
rs1005248187 Chr.19: 37347869-37347869 on Build GRCh38
rs185216417 Chr.19: 37347973-37347973 on Build GRCh38
rs769554350 Chr.19: 37347791-37347791 on Build GRCh38
rs751962724 Chr.19: 37347830-37347830 on Build GRCh38
rs931272774 Chr.19: 37347932-37347932 on Build GRCh38
rs1020180704 Chr.19: 37348168-37348168 on Build GRCh38
rs753540564 Chr.19: 37347845-37347845 on Build GRCh38
rs377264603 Chr.19: 37347814-37347814 on Build GRCh38
rs200176071 Chr.19: 37347789-37347789 on Build GRCh38
rs746882660 Chr.19: 37347784-37347784 on Build GRCh38
rs1011160076 Chr.19: 37348127-37348127 on Build GRCh38
rs777504944 Chr.19: 37347855-37347855 on Build GRCh38
rs201744812 Chr.19: 37347819-37347819 on Build GRCh38
rs755274380 Chr.19: 37347745-37347745 on Build GRCh38
rs182413666 Chr.19: 37347921-37347921 on Build GRCh38
rs922140070 Chr.19: 37348093-37348093 on Build GRCh38
rs372874887 Chr.19: 37347899-37347899 on Build GRCh38
rs940886206 Chr.19: 37348125-37348125 on Build GRCh38
rs770380410 Chr.19: 37347785-37347785 on Build GRCh38
rs760296210 Chr.19: 37347832-37347833 on Build GRCh38
rs574072796 Chr.19: 37348138-37348138 on Build GRCh38
rs1028500140 Chr.19: 37347854-37347854 on Build GRCh38
rs1012018422 Chr.19: 37347984-37347984 on Build GRCh38
rs369823853 Chr.19: 37347818-37347818 on Build GRCh38
rs959445956 Chr.19: 37347957-37347957 on Build GRCh38

View Full Product Details