Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37362412-37362921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HKR1
Entrez Gene ID
284459
Gene Name
HKR1, GLI-Kruppel zinc finger family member
Gene Aliases
ZNF875
UniGene
Hs.657027
Gene Chromosome Location
Chr.19: 37317900-37364455 on Build GRCh38
Transcripts
NM_181786.2 XM_017026672.1 XM_017026673.1 XM_017026674.1 XM_017026675.1 XM_017026676.1 XM_017026677.1 XM_017026678.1 XM_017026679.1 XM_017026680.1 XM_017026681.1 XM_017026682.1 XM_017026683.1 XM_017026684.1 XM_017026685.1 XM_017026686.1 XM_017026687.1 XM_017026688.1 XM_017026689.1 XM_017026690.1 XM_017026691.1 XM_017026692.1 XM_017026693.1 XM_017026694.1 XM_017026695.1 XM_017026696.1 XM_017026697.1 XM_017026698.1 XM_017026699.1 XM_017026700.1 XM_017026701.1 XM_017026702.1 XM_017026703.1 XM_017026704.1 XM_017026705.1 XM_017026707.1 XM_017026708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6557822 c.834G>T p.W278C Chr.19: 37362629-37362629 on Build GRCh38
4690589 c.864delC p.I289fs*28 Chr.19: 37362659-37362659 on Build GRCh38
3362825 c.904A>G p.R302G Chr.19: 37362756-37362756 on Build GRCh38
5972901 c.763A>C p.M255L Chr.19: 37362615-37362615 on Build GRCh38
1680952 c.628G>A p.E210K Chr.19: 37362423-37362423 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745719513 Chr.19: 37362562-37362562 on Build GRCh38
rs758992658 Chr.19: 37362624-37362624 on Build GRCh38
rs755355980 Chr.19: 37362480-37362480 on Build GRCh38
rs560065326 Chr.19: 37362832-37362832 on Build GRCh38
rs753167850 Chr.19: 37362618-37362618 on Build GRCh38
rs777568613 Chr.19: 37362841-37362841 on Build GRCh38
rs374853630 Chr.19: 37362717-37362717 on Build GRCh38
rs767872971 Chr.19: 37362851-37362852 on Build GRCh38
rs372430557 Chr.19: 37362865-37362865 on Build GRCh38
rs1043870684 Chr.19: 37362661-37362661 on Build GRCh38
rs762124057 Chr.19: 37362834-37362836 on Build GRCh38
rs201417310 Chr.19: 37362767-37362768 on Build GRCh38
rs779299590 Chr.19: 37362492-37362492 on Build GRCh38
rs146265235 Chr.19: 37362524-37362524 on Build GRCh38
rs373966528 Chr.19: 37362709-37362709 on Build GRCh38
rs201241071 Chr.19: 37362850-37362850 on Build GRCh38
rs145202068 Chr.19: 37362708-37362708 on Build GRCh38
rs368976173 Chr.19: 37362718-37362718 on Build GRCh38
rs999709068 Chr.19: 37362674-37362674 on Build GRCh38
rs771323767 Chr.19: 37362542-37362542 on Build GRCh38
rs144076342 Chr.19: 37362629-37362629 on Build GRCh38
rs763959744 Chr.19: 37362509-37362509 on Build GRCh38
rs772884785 Chr.19: 37362697-37362697 on Build GRCh38
rs776352754 Chr.19: 37362860-37362860 on Build GRCh38
rs549222752 Chr.19: 37362874-37362874 on Build GRCh38
rs938573006 Chr.19: 37362601-37362601 on Build GRCh38
rs763226157 Chr.19: 37362713-37362713 on Build GRCh38
rs762371703 Chr.19: 37362458-37362458 on Build GRCh38
rs761324142 Chr.19: 37362692-37362692 on Build GRCh38
rs769546404 Chr.19: 37362879-37362879 on Build GRCh38
rs144928795 Chr.19: 37362771-37362771 on Build GRCh38
rs561529070 Chr.19: 37362646-37362646 on Build GRCh38
rs746060051 Chr.19: 37362655-37362655 on Build GRCh38
rs757420510 Chr.19: 37362722-37362722 on Build GRCh38
rs981209055 Chr.19: 37362548-37362548 on Build GRCh38
rs141791100 Chr.19: 37362848-37362848 on Build GRCh38
rs200258398 Chr.19: 37362768-37362768 on Build GRCh38
rs891602072 Chr.19: 37362634-37362634 on Build GRCh38
rs781564734 Chr.19: 37362551-37362551 on Build GRCh38
rs762456237 Chr.19: 37362804-37362804 on Build GRCh38
rs113383966 Chr.19: 37362680-37362680 on Build GRCh38
rs761307891 Chr.19: 37362599-37362599 on Build GRCh38
rs769904875 Chr.19: 37362680-37362681 on Build GRCh38
rs763616903 Chr.19: 37362462-37362462 on Build GRCh38
rs776621335 Chr.19: 37362438-37362438 on Build GRCh38
rs199921655 Chr.19: 37362650-37362650 on Build GRCh38
rs771594404 Chr.19: 37362775-37362775 on Build GRCh38
rs770500910 Chr.19: 37362831-37362831 on Build GRCh38
rs61741499 Chr.19: 37362473-37362473 on Build GRCh38
rs554059074 Chr.19: 37362699-37362699 on Build GRCh38
rs556902010 Chr.19: 37362598-37362598 on Build GRCh38
rs771502234 Chr.19: 37362696-37362696 on Build GRCh38
rs757402237 Chr.19: 37362641-37362641 on Build GRCh38
rs775319262 Chr.19: 37362582-37362582 on Build GRCh38
rs772827496 Chr.19: 37362778-37362778 on Build GRCh38
rs769495658 Chr.19: 37362664-37362664 on Build GRCh38
rs200823279 Chr.19: 37362801-37362801 on Build GRCh38
rs540658729 Chr.19: 37362730-37362730 on Build GRCh38
rs143859791 Chr.19: 37362454-37362454 on Build GRCh38
rs770687735 Chr.19: 37362787-37362787 on Build GRCh38
rs769576840 Chr.19: 37362572-37362572 on Build GRCh38
rs571969598 Chr.19: 37362669-37362669 on Build GRCh38
rs774859990 Chr.19: 37362787-37362792 on Build GRCh38
rs1056364092 Chr.19: 37362573-37362573 on Build GRCh38
rs765156846 Chr.19: 37362448-37362448 on Build GRCh38
rs867496971 Chr.19: 37362472-37362472 on Build GRCh38
rs1043715536 Chr.19: 37362849-37362849 on Build GRCh38
rs759825105 Chr.19: 37362615-37362615 on Build GRCh38
rs969900944 Chr.19: 37362830-37362830 on Build GRCh38
rs945866007 Chr.19: 37362656-37362656 on Build GRCh38
rs750018165 Chr.19: 37362611-37362611 on Build GRCh38
rs149130534 Chr.19: 37362763-37362763 on Build GRCh38
rs751759669 Chr.19: 37362631-37362631 on Build GRCh38
rs775501206 Chr.19: 37362684-37362684 on Build GRCh38
rs200269969 Chr.19: 37362740-37362746 on Build GRCh38
rs768452915 Chr.19: 37362594-37362594 on Build GRCh38
rs539977617 Chr.19: 37362721-37362721 on Build GRCh38
rs759376928 Chr.19: 37362445-37362445 on Build GRCh38
rs925373660 Chr.19: 37362877-37362877 on Build GRCh38
rs200183559 Chr.19: 37362802-37362802 on Build GRCh38
rs543758955 Chr.19: 37362791-37362791 on Build GRCh38
rs559715244 Chr.19: 37362781-37362781 on Build GRCh38
rs984129638 Chr.19: 37362490-37362490 on Build GRCh38
rs35764998 Chr.19: 37362811-37362812 on Build GRCh38
rs754267556 Chr.19: 37362476-37362476 on Build GRCh38
rs951555549 Chr.19: 37362489-37362489 on Build GRCh38
rs748188125 Chr.19: 37362543-37362543 on Build GRCh38
rs756394172 Chr.19: 37362741-37362741 on Build GRCh38
rs376405953 Chr.19: 37362698-37362698 on Build GRCh38
rs766637109 Chr.19: 37362825-37362825 on Build GRCh38
rs778632069 Chr.19: 37362835-37362835 on Build GRCh38
rs750579055 Chr.19: 37362820-37362820 on Build GRCh38
rs767214467 Chr.19: 37362602-37362602 on Build GRCh38
rs762896488 Chr.19: 37362586-37362586 on Build GRCh38
rs747063730 Chr.19: 37362532-37362532 on Build GRCh38
rs368148904 Chr.19: 37362575-37362575 on Build GRCh38
rs749000957 Chr.19: 37362679-37362679 on Build GRCh38
rs758372853 Chr.19: 37362840-37362840 on Build GRCh38
rs878933973 Chr.19: 37362719-37362719 on Build GRCh38
rs139305805 Chr.19: 37362684-37362684 on Build GRCh38
rs749773137 Chr.19: 37362762-37362765 on Build GRCh38
rs774894163 Chr.19: 37362883-37362883 on Build GRCh38
rs918522614 Chr.19: 37362555-37362555 on Build GRCh38
rs767652525 Chr.19: 37362727-37362727 on Build GRCh38
rs927213615 Chr.19: 37362593-37362593 on Build GRCh38
rs772831183 Chr.19: 37362737-37362737 on Build GRCh38
rs34809375 Chr.19: 37362464-37362464 on Build GRCh38
rs149990980 Chr.19: 37362703-37362703 on Build GRCh38
rs147945768 Chr.19: 37362812-37362812 on Build GRCh38
rs746472194 Chr.19: 37362785-37362785 on Build GRCh38
rs758440171 Chr.19: 37362506-37362506 on Build GRCh38
rs771455653 Chr.19: 37362712-37362712 on Build GRCh38
rs770292858 Chr.19: 37362855-37362855 on Build GRCh38
rs112164866 Chr.19: 37362463-37362463 on Build GRCh38
rs754709756 Chr.19: 37362770-37362770 on Build GRCh38
rs202210305 Chr.19: 37362869-37362869 on Build GRCh38
rs780131454 Chr.19: 37362758-37362758 on Build GRCh38
rs778396892 Chr.19: 37362628-37362628 on Build GRCh38
rs778981164 Chr.19: 37362500-37362500 on Build GRCh38
rs193293988 Chr.19: 37362821-37362821 on Build GRCh38
rs777952106 Chr.19: 37362514-37362514 on Build GRCh38
rs555011193 Chr.19: 37362728-37362728 on Build GRCh38
rs754797536 Chr.19: 37362834-37362834 on Build GRCh38

View Full Product Details