Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38705960-38706418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTN4
Entrez Gene ID
81
Gene Name
actinin alpha 4
Gene Aliases
ACTININ-4, FSGS, FSGS1
UniGene
Hs.270291
Gene Chromosome Location
Chr.19: 38647616-38730532 on Build GRCh38
Transcripts
NM_001322033.1 NM_004924.5 XM_005259281.4 XM_006723406.2 XM_017027331.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5748054 c.530C>T p.T177I Chr.19: 38706089-38706089 on Build GRCh38
711562 c.508C>T p.L170F Chr.19: 38706067-38706067 on Build GRCh38
439432 c.536C>G p.P179R Chr.19: 38706095-38706095 on Build GRCh38
3756761 c.546C>T p.N182N Chr.19: 38706105-38706105 on Build GRCh38
5813745 c.496A>T p.K166* Chr.19: 38706055-38706055 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138744742 Chr.19: 38706298-38706298 on Build GRCh38
rs750397674 Chr.19: 38706023-38706023 on Build GRCh38
rs202126611 Chr.19: 38706110-38706110 on Build GRCh38
rs1023903731 Chr.19: 38706195-38706195 on Build GRCh38
rs994626689 Chr.19: 38706098-38706098 on Build GRCh38
rs753425567 Chr.19: 38706036-38706036 on Build GRCh38
rs1054585756 Chr.19: 38706385-38706385 on Build GRCh38
rs145730643 Chr.19: 38706136-38706136 on Build GRCh38
rs908094639 Chr.19: 38706238-38706238 on Build GRCh38
rs953246618 Chr.19: 38706372-38706372 on Build GRCh38
rs751578535 Chr.19: 38706004-38706004 on Build GRCh38
rs890338538 Chr.19: 38706035-38706035 on Build GRCh38
rs377717555 Chr.19: 38706051-38706051 on Build GRCh38
rs11553600 Chr.19: 38706096-38706096 on Build GRCh38
rs374307321 Chr.19: 38706015-38706015 on Build GRCh38
rs537646804 Chr.19: 38706263-38706263 on Build GRCh38
rs956915113 Chr.19: 38706336-38706336 on Build GRCh38
rs774586070 Chr.19: 38706137-38706137 on Build GRCh38
rs964772051 Chr.19: 38706141-38706141 on Build GRCh38
rs374345491 Chr.19: 38706377-38706377 on Build GRCh38
rs754477143 Chr.19: 38706056-38706056 on Build GRCh38
rs761831106 Chr.19: 38706138-38706138 on Build GRCh38
rs749800666 Chr.19: 38706384-38706384 on Build GRCh38
rs1046014063 Chr.19: 38706362-38706362 on Build GRCh38
rs763322376 Chr.19: 38706134-38706134 on Build GRCh38
rs553380979 Chr.19: 38706345-38706345 on Build GRCh38
rs3745859 Chr.19: 38706105-38706105 on Build GRCh38
rs1003779231 Chr.19: 38706273-38706273 on Build GRCh38
rs753626944 Chr.19: 38706176-38706176 on Build GRCh38
rs750292320 Chr.19: 38706152-38706152 on Build GRCh38
rs112614234 Chr.19: 38706378-38706378 on Build GRCh38
rs202069899 Chr.19: 38706030-38706030 on Build GRCh38
rs905512450 Chr.19: 38706364-38706364 on Build GRCh38
rs779779723 Chr.19: 38706365-38706365 on Build GRCh38
rs567413755 Chr.19: 38706218-38706218 on Build GRCh38
rs892900081 Chr.19: 38706390-38706390 on Build GRCh38
rs1020179411 Chr.19: 38706100-38706100 on Build GRCh38
rs754733256 Chr.19: 38706048-38706048 on Build GRCh38
rs555388589 Chr.19: 38706181-38706181 on Build GRCh38
rs571733186 Chr.19: 38706346-38706346 on Build GRCh38
rs767316355 Chr.19: 38706022-38706022 on Build GRCh38
rs779935779 Chr.19: 38706031-38706031 on Build GRCh38
rs200442845 Chr.19: 38706154-38706154 on Build GRCh38
rs560404625 Chr.19: 38705986-38705986 on Build GRCh38
rs60710743 Chr.19: 38706142-38706142 on Build GRCh38
rs577359710 Chr.19: 38706281-38706281 on Build GRCh38
rs745566266 Chr.19: 38706221-38706221 on Build GRCh38
rs766071828 Chr.19: 38706160-38706160 on Build GRCh38
rs149027682 Chr.19: 38706095-38706095 on Build GRCh38
rs1031036950 Chr.19: 38706310-38706310 on Build GRCh38
rs556220518 Chr.19: 38706280-38706280 on Build GRCh38
rs571812757 Chr.19: 38706106-38706106 on Build GRCh38
rs769397885 Chr.19: 38706242-38706242 on Build GRCh38

View Full Product Details