Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38838809-38839310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPL
Entrez Gene ID
3191
Gene Name
heterogeneous nuclear ribonucleoprotein L
Gene Aliases
HNRPL, P/OKcl.14, hnRNP-L
UniGene
Hs.644906
Gene Chromosome Location
Chr.19: 38836378-38852339 on Build GRCh38
Transcripts
NM_001005335.1 NM_001533.2 XM_011526886.1 XM_011526887.1 XM_011526888.2 XM_011526889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4769044 c.918C>T p.L306L Chr.19: 38838932-38838932 on Build GRCh38
1393462 c.856C>T p.P286S Chr.19: 38838994-38838994 on Build GRCh38
3223059 c.1256C>T p.P419L Chr.19: 38838993-38838993 on Build GRCh38
5743990 c.863C>G p.A288G Chr.19: 38838987-38838987 on Build GRCh38
5743989 c.1262C>G p.A421G Chr.19: 38838987-38838987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746172881 Chr.19: 38838938-38838938 on Build GRCh38
rs748513036 Chr.19: 38839049-38839050 on Build GRCh38
rs1001410739 Chr.19: 38839183-38839183 on Build GRCh38
rs111990928 Chr.19: 38839218-38839218 on Build GRCh38
rs753436899 Chr.19: 38839027-38839027 on Build GRCh38
rs775543327 Chr.19: 38838842-38838842 on Build GRCh38
rs138853456 Chr.19: 38839254-38839254 on Build GRCh38
rs761627186 Chr.19: 38838857-38838857 on Build GRCh38
rs752038266 Chr.19: 38839032-38839032 on Build GRCh38
rs1013670714 Chr.19: 38839142-38839142 on Build GRCh38
rs755510224 Chr.19: 38838866-38838867 on Build GRCh38
rs761523358 Chr.19: 38838977-38838977 on Build GRCh38
rs781665231 Chr.19: 38839047-38839047 on Build GRCh38
rs756619213 Chr.19: 38839093-38839093 on Build GRCh38
rs532376599 Chr.19: 38838831-38838831 on Build GRCh38
rs941887334 Chr.19: 38839033-38839033 on Build GRCh38
rs377469393 Chr.19: 38838845-38838847 on Build GRCh38
rs577232775 Chr.19: 38839242-38839242 on Build GRCh38
rs75870432 Chr.19: 38838876-38838876 on Build GRCh38
rs1033035356 Chr.19: 38838958-38838958 on Build GRCh38
rs994141826 Chr.19: 38838840-38838840 on Build GRCh38
rs979686914 Chr.19: 38838950-38838950 on Build GRCh38
rs919152764 Chr.19: 38839084-38839084 on Build GRCh38
rs748179974 Chr.19: 38839063-38839063 on Build GRCh38
rs922941829 Chr.19: 38839018-38839018 on Build GRCh38
rs368341400 Chr.19: 38838853-38838853 on Build GRCh38
rs8111670 Chr.19: 38838877-38838877 on Build GRCh38
rs375580983 Chr.19: 38838885-38838885 on Build GRCh38
rs192696991 Chr.19: 38838861-38838861 on Build GRCh38
rs749389416 Chr.19: 38839059-38839059 on Build GRCh38
rs778490307 Chr.19: 38839200-38839200 on Build GRCh38
rs750030160 Chr.19: 38838868-38838868 on Build GRCh38
rs75450845 Chr.19: 38839077-38839077 on Build GRCh38
rs774056652 Chr.19: 38838854-38838854 on Build GRCh38
rs113878198 Chr.19: 38839028-38839028 on Build GRCh38
rs781275659 Chr.19: 38838927-38838927 on Build GRCh38
rs541796197 Chr.19: 38838829-38838829 on Build GRCh38
rs764523189 Chr.19: 38839029-38839029 on Build GRCh38
rs757701406 Chr.19: 38839037-38839037 on Build GRCh38
rs1045582869 Chr.19: 38839158-38839158 on Build GRCh38
rs765425372 Chr.19: 38839030-38839030 on Build GRCh38
rs887291879 Chr.19: 38838859-38838859 on Build GRCh38
rs1009781014 Chr.19: 38839220-38839220 on Build GRCh38
rs964785031 Chr.19: 38839269-38839269 on Build GRCh38
rs142725492 Chr.19: 38838959-38838959 on Build GRCh38
rs779634455 Chr.19: 38839041-38839041 on Build GRCh38
rs778716649 Chr.19: 38839061-38839061 on Build GRCh38
rs201176574 Chr.19: 38838878-38838878 on Build GRCh38
rs976139287 Chr.19: 38839128-38839128 on Build GRCh38
rs941500989 Chr.19: 38839160-38839160 on Build GRCh38
rs535615136 Chr.19: 38839021-38839021 on Build GRCh38
rs771828498 Chr.19: 38838985-38838985 on Build GRCh38
rs758635649 Chr.19: 38839050-38839051 on Build GRCh38
rs757585525 Chr.19: 38838919-38838919 on Build GRCh38
rs751892855 Chr.19: 38838886-38838886 on Build GRCh38
rs905258811 Chr.19: 38839185-38839185 on Build GRCh38
rs1047346765 Chr.19: 38839152-38839152 on Build GRCh38
rs544280273 Chr.19: 38839133-38839133 on Build GRCh38
rs550948499 Chr.19: 38839090-38839090 on Build GRCh38
rs896512302 Chr.19: 38839204-38839204 on Build GRCh38
rs780211831 Chr.19: 38839055-38839055 on Build GRCh38
rs1038574254 Chr.19: 38839162-38839162 on Build GRCh38
rs1056840353 Chr.19: 38839203-38839203 on Build GRCh38
rs201860193 Chr.19: 38838848-38838848 on Build GRCh38
rs1047063547 Chr.19: 38839099-38839099 on Build GRCh38
rs768720452 Chr.19: 38839060-38839060 on Build GRCh38
rs778306000 Chr.19: 38838884-38838884 on Build GRCh38
rs200248378 Chr.19: 38838880-38838880 on Build GRCh38
rs200815731 Chr.19: 38838944-38838944 on Build GRCh38
rs760460749 Chr.19: 38838989-38838989 on Build GRCh38
rs1017465527 Chr.19: 38839259-38839259 on Build GRCh38
rs756370705 Chr.19: 38839051-38839051 on Build GRCh38
rs780160907 Chr.19: 38838945-38838945 on Build GRCh38
rs539852555 Chr.19: 38839134-38839134 on Build GRCh38
rs774292826 Chr.19: 38838960-38838960 on Build GRCh38
rs148046358 Chr.19: 38838986-38838986 on Build GRCh38
rs749338445 Chr.19: 38838949-38838949 on Build GRCh38
rs1020836437 Chr.19: 38839230-38839230 on Build GRCh38
rs372302259 Chr.19: 38838992-38838992 on Build GRCh38
rs370003343 Chr.19: 38838939-38838939 on Build GRCh38
rs375728875 Chr.19: 38839048-38839048 on Build GRCh38
rs868857187 Chr.19: 38839222-38839222 on Build GRCh38
rs550065375 Chr.19: 38839261-38839261 on Build GRCh38
rs905344804 Chr.19: 38839168-38839168 on Build GRCh38

View Full Product Details