Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38840397-38840891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPL
Entrez Gene ID
3191
Gene Name
heterogeneous nuclear ribonucleoprotein L
Gene Aliases
HNRPL, P/OKcl.14, hnRNP-L
UniGene
Hs.644906
Gene Chromosome Location
Chr.19: 38836378-38852339 on Build GRCh38
Transcripts
NM_001005335.1 NM_001533.2 XM_011526886.1 XM_011526887.1 XM_011526888.2 XM_011526889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
188884 c.908G>A p.R303H Chr.19: 38840532-38840532 on Build GRCh38
4779563 c.490G>A p.G164S Chr.19: 38840551-38840551 on Build GRCh38
6038201 c.515G>T p.R172M Chr.19: 38840526-38840526 on Build GRCh38
188885 c.509G>A p.R170H Chr.19: 38840532-38840532 on Build GRCh38
6038200 c.914G>T p.R305M Chr.19: 38840526-38840526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1041549450 Chr.19: 38840716-38840716 on Build GRCh38
rs138342994 Chr.19: 38840537-38840537 on Build GRCh38
rs181891806 Chr.19: 38840763-38840763 on Build GRCh38
rs749703277 Chr.19: 38840737-38840737 on Build GRCh38
rs115459451 Chr.19: 38840679-38840679 on Build GRCh38
rs527655440 Chr.19: 38840565-38840565 on Build GRCh38
rs755171573 Chr.19: 38840439-38840439 on Build GRCh38
rs1041335412 Chr.19: 38840521-38840521 on Build GRCh38
rs190189566 Chr.19: 38840761-38840761 on Build GRCh38
rs373422201 Chr.19: 38840718-38840718 on Build GRCh38
rs375258554 Chr.19: 38840426-38840426 on Build GRCh38
rs991268339 Chr.19: 38840620-38840620 on Build GRCh38
rs756615008 Chr.19: 38840571-38840571 on Build GRCh38
rs952215326 Chr.19: 38840482-38840482 on Build GRCh38
rs1008873464 Chr.19: 38840434-38840434 on Build GRCh38
rs917093102 Chr.19: 38840655-38840655 on Build GRCh38
rs201026923 Chr.19: 38840455-38840455 on Build GRCh38
rs1054287486 Chr.19: 38840423-38840423 on Build GRCh38
rs773325531 Chr.19: 38840474-38840474 on Build GRCh38
rs753605911 Chr.19: 38840479-38840479 on Build GRCh38
rs572104032 Chr.19: 38840741-38840741 on Build GRCh38
rs578144612 Chr.19: 38840448-38840448 on Build GRCh38
rs780566220 Chr.19: 38840572-38840572 on Build GRCh38
rs769078974 Chr.19: 38840599-38840599 on Build GRCh38
rs983692286 Chr.19: 38840553-38840553 on Build GRCh38
rs544465888 Chr.19: 38840431-38840431 on Build GRCh38
rs926985447 Chr.19: 38840601-38840601 on Build GRCh38
rs1015103660 Chr.19: 38840800-38840800 on Build GRCh38
rs11083481 Chr.19: 38840656-38840656 on Build GRCh38
rs574304705 Chr.19: 38840427-38840427 on Build GRCh38
rs764881166 Chr.19: 38840498-38840498 on Build GRCh38
rs1008573044 Chr.19: 38840730-38840730 on Build GRCh38
rs891052555 Chr.19: 38840432-38840432 on Build GRCh38
rs950367372 Chr.19: 38840573-38840573 on Build GRCh38
rs937030222 Chr.19: 38840870-38840870 on Build GRCh38
rs775758796 Chr.19: 38840454-38840454 on Build GRCh38
rs766305798 Chr.19: 38840477-38840477 on Build GRCh38
rs753964012 Chr.19: 38840422-38840428 on Build GRCh38
rs767557213 Chr.19: 38840470-38840470 on Build GRCh38
rs368946663 Chr.19: 38840678-38840678 on Build GRCh38
rs563117450 Chr.19: 38840441-38840441 on Build GRCh38
rs1027771511 Chr.19: 38840773-38840773 on Build GRCh38
rs890648908 Chr.19: 38840738-38840738 on Build GRCh38
rs949831531 Chr.19: 38840519-38840519 on Build GRCh38
rs754664760 Chr.19: 38840422-38840422 on Build GRCh38
rs754718124 Chr.19: 38840481-38840481 on Build GRCh38
rs572466486 Chr.19: 38840421-38840421 on Build GRCh38
rs1027683253 Chr.19: 38840863-38840863 on Build GRCh38
rs760573689 Chr.19: 38840476-38840476 on Build GRCh38
rs749686243 Chr.19: 38840586-38840586 on Build GRCh38
rs915462051 Chr.19: 38840725-38840725 on Build GRCh38
rs553628126 Chr.19: 38840698-38840698 on Build GRCh38
rs900543042 Chr.19: 38840745-38840745 on Build GRCh38
rs574181669 Chr.19: 38840568-38840568 on Build GRCh38
rs891003922 Chr.19: 38840717-38840717 on Build GRCh38
rs995226475 Chr.19: 38840830-38840830 on Build GRCh38
rs555614813 Chr.19: 38840531-38840531 on Build GRCh38
rs769625150 Chr.19: 38840607-38840607 on Build GRCh38
rs1054154772 Chr.19: 38840666-38840666 on Build GRCh38
rs958672758 Chr.19: 38840602-38840602 on Build GRCh38
rs1054168146 Chr.19: 38840715-38840715 on Build GRCh38
rs369462452 Chr.19: 38840567-38840567 on Build GRCh38
rs777899404 Chr.19: 38840533-38840533 on Build GRCh38
rs566584297 Chr.19: 38840786-38840786 on Build GRCh38
rs561817674 Chr.19: 38840653-38840653 on Build GRCh38
rs768878394 Chr.19: 38840760-38840760 on Build GRCh38
rs745525449 Chr.19: 38840577-38840580 on Build GRCh38
rs971665310 Chr.19: 38840860-38840860 on Build GRCh38
rs777199339 Chr.19: 38840562-38840562 on Build GRCh38
rs372961507 Chr.19: 38840576-38840576 on Build GRCh38
rs568663477 Chr.19: 38840690-38840690 on Build GRCh38
rs147384264 Chr.19: 38840555-38840555 on Build GRCh38
rs982139602 Chr.19: 38840865-38840865 on Build GRCh38
rs761991974 Chr.19: 38840469-38840469 on Build GRCh38
rs774659308 Chr.19: 38840468-38840468 on Build GRCh38
rs769026015 Chr.19: 38840464-38840464 on Build GRCh38
rs748437040 Chr.19: 38840610-38840610 on Build GRCh38
rs952121872 Chr.19: 38840778-38840778 on Build GRCh38
rs758978115 Chr.19: 38840599-38840599 on Build GRCh38
rs1054455114 Chr.19: 38840557-38840557 on Build GRCh38
rs977144622 Chr.19: 38840675-38840675 on Build GRCh38
rs765424872 Chr.19: 38840575-38840576 on Build GRCh38
rs973728733 Chr.19: 38840430-38840430 on Build GRCh38
rs945852374 Chr.19: 38840699-38840699 on Build GRCh38
rs926960395 Chr.19: 38840867-38840867 on Build GRCh38
rs983595742 Chr.19: 38840799-38840799 on Build GRCh38
rs774849215 Chr.19: 38840603-38840603 on Build GRCh38
rs372108768 Chr.19: 38840424-38840424 on Build GRCh38
rs777333488 Chr.19: 38840439-38840439 on Build GRCh38

View Full Product Details