Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38843759-38844266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPL
Entrez Gene ID
3191
Gene Name
heterogeneous nuclear ribonucleoprotein L
Gene Aliases
HNRPL, P/OKcl.14, hnRNP-L
UniGene
Hs.644906
Gene Chromosome Location
Chr.19: 38836378-38852339 on Build GRCh38
Transcripts
NM_001005335.1 NM_001533.2 XM_011526886.1 XM_011526887.1 XM_011526888.2 XM_011526889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1750875 c.719C>T p.S240L Chr.19: 38844096-38844096 on Build GRCh38
3822988 c.789G>A p.L263L Chr.19: 38844026-38844026 on Build GRCh38
6209684 c.771T>G p.Y257* Chr.19: 38844044-38844044 on Build GRCh38
6209685 c.372T>G p.Y124* Chr.19: 38844044-38844044 on Build GRCh38
3533586 c.469C>T p.L157F Chr.19: 38843854-38843854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757501860 Chr.19: 38844056-38844056 on Build GRCh38
rs374481287 Chr.19: 38843831-38843831 on Build GRCh38
rs777860885 Chr.19: 38843972-38843972 on Build GRCh38
rs190537736 Chr.19: 38844195-38844195 on Build GRCh38
rs751839176 Chr.19: 38844053-38844053 on Build GRCh38
rs754465629 Chr.19: 38843821-38843821 on Build GRCh38
rs764577444 Chr.19: 38844038-38844038 on Build GRCh38
rs781000802 Chr.19: 38844220-38844220 on Build GRCh38
rs763671581 Chr.19: 38843924-38843924 on Build GRCh38
rs2278011 Chr.19: 38844171-38844171 on Build GRCh38
rs1017286943 Chr.19: 38843780-38843780 on Build GRCh38
rs768565086 Chr.19: 38843988-38843988 on Build GRCh38
rs755688211 Chr.19: 38843968-38843968 on Build GRCh38
rs768874041 Chr.19: 38843785-38843785 on Build GRCh38
rs543663859 Chr.19: 38844212-38844212 on Build GRCh38
rs779744645 Chr.19: 38844105-38844106 on Build GRCh38
rs559981281 Chr.19: 38843825-38843825 on Build GRCh38
rs889830408 Chr.19: 38844192-38844192 on Build GRCh38
rs113254301 Chr.19: 38843985-38843985 on Build GRCh38
rs762763265 Chr.19: 38843923-38843923 on Build GRCh38
rs776687180 Chr.19: 38843858-38843858 on Build GRCh38
rs926644067 Chr.19: 38843885-38843885 on Build GRCh38
rs116924024 Chr.19: 38843807-38843807 on Build GRCh38
rs758826050 Chr.19: 38844028-38844028 on Build GRCh38
rs561801215 Chr.19: 38844014-38844014 on Build GRCh38
rs772211840 Chr.19: 38843830-38843830 on Build GRCh38
rs371371816 Chr.19: 38844060-38844060 on Build GRCh38
rs769746428 Chr.19: 38843983-38843983 on Build GRCh38
rs369335148 Chr.19: 38843945-38843945 on Build GRCh38
rs74970157 Chr.19: 38844033-38844033 on Build GRCh38
rs751159349 Chr.19: 38843928-38843928 on Build GRCh38
rs768096000 Chr.19: 38844144-38844172 on Build GRCh38
rs367888491 Chr.19: 38843809-38843809 on Build GRCh38
rs753025004 Chr.19: 38843958-38843958 on Build GRCh38
rs1052809286 Chr.19: 38843804-38843804 on Build GRCh38
rs530794820 Chr.19: 38844083-38844083 on Build GRCh38
rs769689876 Chr.19: 38843882-38843882 on Build GRCh38
rs780432896 Chr.19: 38843813-38843813 on Build GRCh38
rs373027792 Chr.19: 38843980-38843980 on Build GRCh38
rs573847733 Chr.19: 38843808-38843808 on Build GRCh38
rs974141132 Chr.19: 38844206-38844206 on Build GRCh38
rs747855653 Chr.19: 38844120-38844120 on Build GRCh38
rs767794858 Chr.19: 38843856-38843856 on Build GRCh38
rs576269771 Chr.19: 38844201-38844201 on Build GRCh38
rs1052292731 Chr.19: 38844153-38844153 on Build GRCh38
rs779517962 Chr.19: 38843955-38843955 on Build GRCh38
rs1056905591 Chr.19: 38844234-38844234 on Build GRCh38
rs745908371 Chr.19: 38843981-38843981 on Build GRCh38
rs148949374 Chr.19: 38844174-38844174 on Build GRCh38
rs763780238 Chr.19: 38844173-38844173 on Build GRCh38
rs184836083 Chr.19: 38843926-38843926 on Build GRCh38
rs774237727 Chr.19: 38844114-38844114 on Build GRCh38
rs982936814 Chr.19: 38844180-38844180 on Build GRCh38
rs376505507 Chr.19: 38843947-38843947 on Build GRCh38
rs762665336 Chr.19: 38843987-38843987 on Build GRCh38
rs1014401519 Chr.19: 38844236-38844236 on Build GRCh38
rs997210556 Chr.19: 38844168-38844168 on Build GRCh38
rs745962318 Chr.19: 38844068-38844068 on Build GRCh38
rs759329224 Chr.19: 38843792-38843792 on Build GRCh38
rs765076257 Chr.19: 38843794-38843794 on Build GRCh38
rs112519570 Chr.19: 38843791-38843791 on Build GRCh38
rs780107935 Chr.19: 38844104-38844104 on Build GRCh38
rs756738326 Chr.19: 38843811-38843811 on Build GRCh38
rs755577550 Chr.19: 38843949-38843949 on Build GRCh38
rs749309091 Chr.19: 38844105-38844105 on Build GRCh38
rs564952220 Chr.19: 38844222-38844222 on Build GRCh38
rs755526210 Chr.19: 38843824-38843824 on Build GRCh38
rs994702972 Chr.19: 38844217-38844217 on Build GRCh38
rs552220975 Chr.19: 38844003-38844003 on Build GRCh38
rs753178935 Chr.19: 38844020-38844020 on Build GRCh38
rs377118943 Chr.19: 38844142-38844143 on Build GRCh38
rs199628884 Chr.19: 38843993-38843993 on Build GRCh38
rs767243516 Chr.19: 38843998-38843998 on Build GRCh38
rs761370201 Chr.19: 38843942-38843942 on Build GRCh38
rs191573929 Chr.19: 38843826-38843826 on Build GRCh38
rs766053087 Chr.19: 38844152-38844152 on Build GRCh38
rs1026288923 Chr.19: 38843954-38843954 on Build GRCh38
rs534999499 Chr.19: 38844154-38844154 on Build GRCh38
rs752468189 Chr.19: 38843796-38843796 on Build GRCh38
rs979172247 Chr.19: 38844098-38844098 on Build GRCh38
rs759521118 Chr.19: 38843861-38843861 on Build GRCh38
rs745325877 Chr.19: 38844238-38844238 on Build GRCh38
rs752544577 Chr.19: 38844138-38844138 on Build GRCh38
rs377566147 Chr.19: 38844111-38844111 on Build GRCh38
rs774829809 Chr.19: 38843787-38843787 on Build GRCh38
rs903566016 Chr.19: 38844062-38844062 on Build GRCh38
rs775352906 Chr.19: 38843984-38843984 on Build GRCh38
rs751057618 Chr.19: 38843810-38843810 on Build GRCh38
rs200635144 Chr.19: 38843969-38843969 on Build GRCh38
rs368215965 Chr.19: 38843935-38843939 on Build GRCh38
rs767190542 Chr.19: 38843944-38843944 on Build GRCh38
rs775295811 Chr.19: 38843922-38843922 on Build GRCh38
rs771871476 Chr.19: 38844124-38844124 on Build GRCh38
rs771038536 Chr.19: 38843973-38843973 on Build GRCh38
rs765851962 Chr.19: 38844017-38844017 on Build GRCh38
rs935129474 Chr.19: 38844145-38844145 on Build GRCh38
rs2278012 Chr.19: 38844190-38844190 on Build GRCh38
rs756123113 Chr.19: 38844071-38844071 on Build GRCh38
rs747073174 Chr.19: 38843849-38843849 on Build GRCh38
rs994417839 Chr.19: 38843943-38843943 on Build GRCh38
rs534954873 Chr.19: 38844224-38844224 on Build GRCh38
rs747446901 Chr.19: 38843927-38843931 on Build GRCh38
rs375903656 Chr.19: 38843907-38843907 on Build GRCh38

View Full Product Details