Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46671733-46672236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX38
Entrez Gene ID
374973
Gene Name
testis expressed 38
Gene Aliases
ATPAF1-AS1, C1orf223, THEG4
UniGene
Hs.443299
Gene Chromosome Location
Chr.1: 46669344-46673603 on Build GRCh38
Transcripts
NM_001145474.3 NM_001300863.1 NM_001300864.1 XM_011541421.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4239001 c.139G>A p.G47S Chr.1: 46671911-46671911 on Build GRCh38
4144049 c.78G>C p.L26L Chr.1: 46671850-46671850 on Build GRCh38
6412788 c.172C>T p.Q58* Chr.1: 46671944-46671944 on Build GRCh38
3977707 c.135C>T p.A45A Chr.1: 46671907-46671907 on Build GRCh38
6318834 c.158G>T p.S53I Chr.1: 46671930-46671930 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs594891 Chr.1: 46671753-46671753 on Build GRCh38
rs563814218 Chr.1: 46671760-46671760 on Build GRCh38
rs746832224 Chr.1: 46671865-46671865 on Build GRCh38
rs772036779 Chr.1: 46671845-46671865 on Build GRCh38
rs1003121525 Chr.1: 46671857-46671857 on Build GRCh38
rs760152690 Chr.1: 46671910-46671910 on Build GRCh38
rs547104348 Chr.1: 46672094-46672094 on Build GRCh38
rs770465669 Chr.1: 46671959-46671959 on Build GRCh38
rs3795317 Chr.1: 46672149-46672149 on Build GRCh38
rs777680365 Chr.1: 46671820-46671820 on Build GRCh38
rs185707183 Chr.1: 46671799-46671799 on Build GRCh38
rs561535715 Chr.1: 46671931-46671931 on Build GRCh38
rs987869593 Chr.1: 46672133-46672133 on Build GRCh38
rs944255406 Chr.1: 46671852-46671852 on Build GRCh38
rs1767602 Chr.1: 46671844-46671844 on Build GRCh38
rs745897014 Chr.1: 46671886-46671886 on Build GRCh38
rs900097052 Chr.1: 46671950-46671950 on Build GRCh38
rs919304336 Chr.1: 46672121-46672121 on Build GRCh38
rs781060431 Chr.1: 46671882-46671882 on Build GRCh38
rs972234061 Chr.1: 46672075-46672077 on Build GRCh38
rs545447967 Chr.1: 46671756-46671756 on Build GRCh38
rs991783839 Chr.1: 46671954-46671954 on Build GRCh38
rs963515211 Chr.1: 46671986-46671986 on Build GRCh38
rs528915381 Chr.1: 46671933-46671933 on Build GRCh38
rs959204198 Chr.1: 46671942-46671942 on Build GRCh38
rs903566363 Chr.1: 46672091-46672091 on Build GRCh38
rs1047500857 Chr.1: 46671819-46671819 on Build GRCh38
rs774756134 Chr.1: 46672000-46672000 on Build GRCh38
rs893665792 Chr.1: 46671762-46671762 on Build GRCh38
rs771332301 Chr.1: 46672023-46672026 on Build GRCh38
rs931421954 Chr.1: 46671766-46671766 on Build GRCh38
rs190616662 Chr.1: 46672141-46672141 on Build GRCh38
rs1033387821 Chr.1: 46671888-46671888 on Build GRCh38
rs758226697 Chr.1: 46671759-46671759 on Build GRCh38
rs933430926 Chr.1: 46672130-46672130 on Build GRCh38
rs374061457 Chr.1: 46672179-46672179 on Build GRCh38
rs768905853 Chr.1: 46671983-46671983 on Build GRCh38
rs770892144 Chr.1: 46671874-46671874 on Build GRCh38
rs943265204 Chr.1: 46672181-46672181 on Build GRCh38

View Full Product Details