Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46814130-46814601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4B1
Entrez Gene ID
1580
Gene Name
cytochrome P450 family 4 subfamily B member 1
Gene Aliases
CYPIVB1, P-450HP
UniGene
Hs.436317
Gene Chromosome Location
Chr.1: 46798995-46819349 on Build GRCh38
Transcripts
NM_000779.3 NM_001099772.1 NM_001319161.1 NM_001319162.1 NM_001319163.1 NR_135003.1 XM_011540832.1 XM_011540833.1 XM_017000466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6639346 c.813G>A p.E271E Chr.1: 46814249-46814249 on Build GRCh38
126643 c.811G>T p.E271* Chr.1: 46814247-46814247 on Build GRCh38
79715 c.867C>G p.L289L Chr.1: 46814303-46814303 on Build GRCh38
6639358 c.806A>G p.Q269R Chr.1: 46814242-46814242 on Build GRCh38
5529538 c.879G>A p.R293R Chr.1: 46814315-46814315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs889967456 Chr.1: 46814532-46814532 on Build GRCh38
rs367726647 Chr.1: 46814328-46814328 on Build GRCh38
rs372926949 Chr.1: 46814504-46814504 on Build GRCh38
rs368259053 Chr.1: 46814246-46814246 on Build GRCh38
rs571442654 Chr.1: 46814486-46814486 on Build GRCh38
rs141452762 Chr.1: 46814275-46814275 on Build GRCh38
rs775371411 Chr.1: 46814203-46814203 on Build GRCh38
rs759345617 Chr.1: 46814182-46814182 on Build GRCh38
rs770558605 Chr.1: 46814173-46814173 on Build GRCh38
rs45581132 Chr.1: 46814359-46814359 on Build GRCh38
rs111807240 Chr.1: 46814256-46814256 on Build GRCh38
rs559516007 Chr.1: 46814273-46814273 on Build GRCh38
rs953983730 Chr.1: 46814432-46814432 on Build GRCh38
rs200462383 Chr.1: 46814214-46814214 on Build GRCh38
rs777081248 Chr.1: 46814363-46814363 on Build GRCh38
rs934297858 Chr.1: 46814267-46814267 on Build GRCh38
rs202136460 Chr.1: 46814177-46814177 on Build GRCh38
rs376567483 Chr.1: 46814530-46814530 on Build GRCh38
rs536939727 Chr.1: 46814557-46814557 on Build GRCh38
rs535076825 Chr.1: 46814162-46814162 on Build GRCh38
rs138537119 Chr.1: 46814227-46814227 on Build GRCh38
rs770562848 Chr.1: 46814259-46814259 on Build GRCh38
rs762025915 Chr.1: 46814228-46814228 on Build GRCh38
rs376093889 Chr.1: 46814298-46814298 on Build GRCh38
rs771919855 Chr.1: 46814190-46814190 on Build GRCh38
rs756632732 Chr.1: 46814161-46814161 on Build GRCh38
rs140074148 Chr.1: 46814170-46814170 on Build GRCh38
rs746509037 Chr.1: 46814258-46814258 on Build GRCh38
rs888414581 Chr.1: 46814212-46814212 on Build GRCh38
rs4646490 Chr.1: 46814414-46814415 on Build GRCh38
rs998490328 Chr.1: 46814529-46814529 on Build GRCh38
rs33942115 Chr.1: 46814413-46814414 on Build GRCh38
rs780919702 Chr.1: 46814485-46814485 on Build GRCh38
rs755830475 Chr.1: 46814340-46814340 on Build GRCh38
rs139611523 Chr.1: 46814313-46814313 on Build GRCh38
rs774429812 Chr.1: 46814278-46814278 on Build GRCh38
rs370689909 Chr.1: 46814237-46814237 on Build GRCh38
rs748159614 Chr.1: 46814348-46814348 on Build GRCh38
rs749048055 Chr.1: 46814344-46814344 on Build GRCh38
rs754324596 Chr.1: 46814303-46814303 on Build GRCh38
rs528324009 Chr.1: 46814436-46814436 on Build GRCh38
rs767645233 Chr.1: 46814233-46814233 on Build GRCh38
rs547157011 Chr.1: 46814385-46814385 on Build GRCh38
rs772865894 Chr.1: 46814359-46814360 on Build GRCh38
rs113702184 Chr.1: 46814417-46814417 on Build GRCh38
rs78619059 Chr.1: 46814413-46814413 on Build GRCh38
rs756549059 Chr.1: 46814546-46814546 on Build GRCh38
rs762922694 Chr.1: 46814209-46814209 on Build GRCh38
rs45446505 Chr.1: 46814226-46814226 on Build GRCh38
rs45520436 Chr.1: 46814165-46814165 on Build GRCh38
rs773484108 Chr.1: 46814354-46814354 on Build GRCh38
rs776321886 Chr.1: 46814190-46814191 on Build GRCh38
rs756688395 Chr.1: 46814314-46814314 on Build GRCh38
rs745503494 Chr.1: 46814336-46814336 on Build GRCh38
rs547051882 Chr.1: 46814443-46814443 on Build GRCh38
rs45592032 Chr.1: 46814415-46814416 on Build GRCh38
rs555010161 Chr.1: 46814561-46814561 on Build GRCh38
rs750642727 Chr.1: 46814286-46814286 on Build GRCh38
rs371917763 Chr.1: 46814480-46814480 on Build GRCh38
rs561129739 Chr.1: 46814382-46814382 on Build GRCh38
rs780606115 Chr.1: 46814250-46814250 on Build GRCh38
rs760978452 Chr.1: 46814358-46814358 on Build GRCh38
rs955714908 Chr.1: 46814450-46814450 on Build GRCh38
rs78212764 Chr.1: 46814416-46814416 on Build GRCh38
rs938141213 Chr.1: 46814342-46814342 on Build GRCh38
rs768587657 Chr.1: 46814346-46814346 on Build GRCh38
rs754375547 Chr.1: 46814255-46814255 on Build GRCh38
rs371702253 Chr.1: 46814341-46814341 on Build GRCh38
rs45578838 Chr.1: 46814257-46814257 on Build GRCh38
rs761368513 Chr.1: 46814193-46814194 on Build GRCh38
rs777201380 Chr.1: 46814168-46814168 on Build GRCh38
rs765004551 Chr.1: 46814356-46814357 on Build GRCh38
rs561327765 Chr.1: 46814152-46814152 on Build GRCh38
rs530787349 Chr.1: 46814351-46814351 on Build GRCh38
rs1048467253 Chr.1: 46814374-46814374 on Build GRCh38
rs532490055 Chr.1: 46814488-46814488 on Build GRCh38
rs58741295 Chr.1: 46814545-46814545 on Build GRCh38
rs755448268 Chr.1: 46814304-46814304 on Build GRCh38
rs769677293 Chr.1: 46814194-46814194 on Build GRCh38
rs377170127 Chr.1: 46814274-46814274 on Build GRCh38
rs45463299 Chr.1: 46814282-46814282 on Build GRCh38
rs200389917 Chr.1: 46814163-46814163 on Build GRCh38
rs372020140 Chr.1: 46814261-46814261 on Build GRCh38
rs1000340639 Chr.1: 46814542-46814542 on Build GRCh38
rs761027364 Chr.1: 46814288-46814288 on Build GRCh38
rs957959104 Chr.1: 46814316-46814316 on Build GRCh38
rs373949660 Chr.1: 46814357-46814357 on Build GRCh38

View Full Product Details