Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46817859-46818358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4B1
Entrez Gene ID
1580
Gene Name
cytochrome P450 family 4 subfamily B member 1
Gene Aliases
CYPIVB1, P-450HP
UniGene
Hs.436317
Gene Chromosome Location
Chr.1: 46798995-46819349 on Build GRCh38
Transcripts
NM_000779.3 NM_001099772.1 NM_001319161.1 NM_001319162.1 NM_001319163.1 NR_135003.1 XM_011540833.1 XM_017000466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3490374 c.1242G>A p.R414R Chr.1: 46818002-46818002 on Build GRCh38
3490375 c.1326C>T p.A442A Chr.1: 46818187-46818187 on Build GRCh38
681881 c.1316A>G p.H439R Chr.1: 46818177-46818177 on Build GRCh38
6063779 c.1260C>G p.P420P Chr.1: 46818020-46818020 on Build GRCh38
3490372 c.1205G>A p.G402E Chr.1: 46817965-46817965 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766475753 Chr.1: 46817947-46817947 on Build GRCh38
rs983897751 Chr.1: 46817897-46817897 on Build GRCh38
rs777324104 Chr.1: 46818222-46818222 on Build GRCh38
rs771709446 Chr.1: 46818218-46818218 on Build GRCh38
rs758763794 Chr.1: 46818006-46818006 on Build GRCh38
rs752801234 Chr.1: 46818077-46818079 on Build GRCh38
rs539676662 Chr.1: 46818096-46818096 on Build GRCh38
rs551914469 Chr.1: 46818165-46818165 on Build GRCh38
rs377503842 Chr.1: 46818237-46818237 on Build GRCh38
rs911890450 Chr.1: 46817904-46817904 on Build GRCh38
rs765344321 Chr.1: 46818069-46818069 on Build GRCh38
rs771474053 Chr.1: 46818279-46818279 on Build GRCh38
rs115133540 Chr.1: 46818146-46818146 on Build GRCh38
rs944648127 Chr.1: 46818282-46818282 on Build GRCh38
rs775080256 Chr.1: 46818025-46818025 on Build GRCh38
rs561533017 Chr.1: 46818023-46818023 on Build GRCh38
rs773003953 Chr.1: 46818147-46818147 on Build GRCh38
rs563755426 Chr.1: 46818335-46818335 on Build GRCh38
rs999227114 Chr.1: 46818032-46818032 on Build GRCh38
rs754957856 Chr.1: 46817982-46817983 on Build GRCh38
rs753868251 Chr.1: 46818065-46818065 on Build GRCh38
rs4646497 Chr.1: 46818062-46818062 on Build GRCh38
rs756081544 Chr.1: 46817974-46817974 on Build GRCh38
rs74072926 Chr.1: 46818142-46818142 on Build GRCh38
rs767406668 Chr.1: 46818065-46818065 on Build GRCh38
rs764241104 Chr.1: 46818176-46818176 on Build GRCh38
rs749594050 Chr.1: 46818213-46818213 on Build GRCh38
rs767989995 Chr.1: 46818259-46818259 on Build GRCh38
rs545470964 Chr.1: 46817910-46817910 on Build GRCh38
rs761696782 Chr.1: 46817924-46817924 on Build GRCh38
rs562588511 Chr.1: 46817901-46817901 on Build GRCh38
rs761419425 Chr.1: 46818041-46818041 on Build GRCh38
rs770817630 Chr.1: 46818153-46818153 on Build GRCh38
rs781771938 Chr.1: 46818129-46818129 on Build GRCh38
rs373023642 Chr.1: 46818101-46818101 on Build GRCh38
rs774412624 Chr.1: 46818252-46818252 on Build GRCh38
rs577801403 Chr.1: 46818288-46818288 on Build GRCh38
rs183384583 Chr.1: 46817996-46817996 on Build GRCh38
rs759646194 Chr.1: 46818067-46818067 on Build GRCh38
rs866561413 Chr.1: 46818181-46818181 on Build GRCh38
rs929337870 Chr.1: 46817950-46817950 on Build GRCh38
rs1039319486 Chr.1: 46818283-46818283 on Build GRCh38
rs896155857 Chr.1: 46818299-46818299 on Build GRCh38
rs760411857 Chr.1: 46818148-46818148 on Build GRCh38
rs4646496 Chr.1: 46817955-46817955 on Build GRCh38
rs553280407 Chr.1: 46818275-46818275 on Build GRCh38
rs998596745 Chr.1: 46818102-46818102 on Build GRCh38
rs199759345 Chr.1: 46818173-46818173 on Build GRCh38
rs775588500 Chr.1: 46818249-46818249 on Build GRCh38
rs973515479 Chr.1: 46817906-46817906 on Build GRCh38
rs778052251 Chr.1: 46818011-46818011 on Build GRCh38
rs778420761 Chr.1: 46818217-46818217 on Build GRCh38
rs754586930 Chr.1: 46818209-46818209 on Build GRCh38
rs944627666 Chr.1: 46817942-46817942 on Build GRCh38
rs555691883 Chr.1: 46818169-46818169 on Build GRCh38
rs769733939 Chr.1: 46818247-46818247 on Build GRCh38
rs764273055 Chr.1: 46818251-46818251 on Build GRCh38
rs776473025 Chr.1: 46818235-46818235 on Build GRCh38
rs148403337 Chr.1: 46818131-46818131 on Build GRCh38
rs777869173 Chr.1: 46818206-46818206 on Build GRCh38
rs369562814 Chr.1: 46818083-46818083 on Build GRCh38
rs747847112 Chr.1: 46818250-46818250 on Build GRCh38
rs777411655 Chr.1: 46818320-46818320 on Build GRCh38
rs975078150 Chr.1: 46818216-46818216 on Build GRCh38
rs756753020 Chr.1: 46818214-46818214 on Build GRCh38
rs757410886 Chr.1: 46818225-46818242 on Build GRCh38
rs757730950 Chr.1: 46818103-46818103 on Build GRCh38
rs45482605 Chr.1: 46818033-46818033 on Build GRCh38
rs545165074 Chr.1: 46818292-46818292 on Build GRCh38
rs201095720 Chr.1: 46818225-46818225 on Build GRCh38
rs565857118 Chr.1: 46818095-46818095 on Build GRCh38
rs370193617 Chr.1: 46818137-46818137 on Build GRCh38
rs1043525777 Chr.1: 46818012-46818012 on Build GRCh38
rs771887648 Chr.1: 46818052-46818052 on Build GRCh38
rs772948119 Chr.1: 46818059-46818059 on Build GRCh38
rs1045107543 Chr.1: 46818334-46818334 on Build GRCh38
rs1011945487 Chr.1: 46818327-46818327 on Build GRCh38
rs186785862 Chr.1: 46818020-46818020 on Build GRCh38
rs368867656 Chr.1: 46818174-46818174 on Build GRCh38
rs751909883 Chr.1: 46818179-46818179 on Build GRCh38
rs756065009 Chr.1: 46817943-46817943 on Build GRCh38
rs757662475 Chr.1: 46818195-46818195 on Build GRCh38
rs767256630 Chr.1: 46817933-46817933 on Build GRCh38
rs911882102 Chr.1: 46818201-46818201 on Build GRCh38
rs143337507 Chr.1: 46818154-46818154 on Build GRCh38
rs750351122 Chr.1: 46817940-46817940 on Build GRCh38
rs766318612 Chr.1: 46818063-46818063 on Build GRCh38
rs922147640 Chr.1: 46818285-46818285 on Build GRCh38
rs753657629 Chr.1: 46818040-46818040 on Build GRCh38
rs755136107 Chr.1: 46817966-46817966 on Build GRCh38
rs748827485 Chr.1: 46818030-46818030 on Build GRCh38
rs779131973 Chr.1: 46817983-46817983 on Build GRCh38
rs770694733 Chr.1: 46818232-46818232 on Build GRCh38
rs948592425 Chr.1: 46817894-46817894 on Build GRCh38
rs190875616 Chr.1: 46817884-46817884 on Build GRCh38
rs762183993 Chr.1: 46818254-46818254 on Build GRCh38
rs141281141 Chr.1: 46818206-46818206 on Build GRCh38
rs756135384 Chr.1: 46818139-46818140 on Build GRCh38

View Full Product Details