Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 50491676-50492202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAF1
Entrez Gene ID
11124
Gene Name
Fas associated factor 1
Gene Aliases
CGI-03, HFAF1s, UBXD12, UBXN3A, hFAF1
UniGene
Hs.530402
Gene Chromosome Location
Chr.1: 50441263-50960264 on Build GRCh38
Transcripts
NM_007051.2 XM_017000136.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6203272 c.802G>C p.E268Q Chr.1: 50491768-50491768 on Build GRCh38
6657963 c.778G>A p.E260K Chr.1: 50491792-50491792 on Build GRCh38
6657962 c.1504G>A p.E502K Chr.1: 50491792-50491792 on Build GRCh38
6265768 c.783C>A p.A261A Chr.1: 50491787-50491787 on Build GRCh38
6265767 c.1509C>A p.A503A Chr.1: 50491787-50491787 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769925871 Chr.1: 50491848-50491849 on Build GRCh38
rs773421330 Chr.1: 50491849-50491849 on Build GRCh38
rs370213986 Chr.1: 50492138-50492138 on Build GRCh38
rs772979864 Chr.1: 50491814-50491814 on Build GRCh38
rs76736434 Chr.1: 50491795-50491795 on Build GRCh38
rs565913348 Chr.1: 50491965-50491965 on Build GRCh38
rs1149790 Chr.1: 50491837-50491837 on Build GRCh38
rs547701776 Chr.1: 50491866-50491866 on Build GRCh38
rs190662129 Chr.1: 50492142-50492142 on Build GRCh38
rs768680457 Chr.1: 50491840-50491840 on Build GRCh38
rs760655656 Chr.1: 50491816-50491816 on Build GRCh38
rs71651146 Chr.1: 50491955-50491955 on Build GRCh38
rs746714010 Chr.1: 50492116-50492116 on Build GRCh38
rs935302663 Chr.1: 50492055-50492055 on Build GRCh38
rs1004236654 Chr.1: 50491715-50491715 on Build GRCh38
rs748774659 Chr.1: 50491727-50491727 on Build GRCh38
rs748185846 Chr.1: 50491839-50491840 on Build GRCh38
rs777475188 Chr.1: 50492054-50492054 on Build GRCh38
rs879038678 Chr.1: 50491857-50491857 on Build GRCh38
rs533395343 Chr.1: 50492021-50492021 on Build GRCh38
rs781448547 Chr.1: 50491840-50491843 on Build GRCh38
rs954995277 Chr.1: 50491710-50491710 on Build GRCh38
rs942264481 Chr.1: 50492043-50492043 on Build GRCh38
rs537867308 Chr.1: 50491859-50491860 on Build GRCh38
rs756800491 Chr.1: 50492171-50492171 on Build GRCh38
rs1028008988 Chr.1: 50491706-50491706 on Build GRCh38
rs774004965 Chr.1: 50491735-50491735 on Build GRCh38
rs1048017166 Chr.1: 50491952-50491952 on Build GRCh38
rs996609303 Chr.1: 50492093-50492093 on Build GRCh38
rs938666481 Chr.1: 50491931-50491931 on Build GRCh38
rs529517331 Chr.1: 50491848-50491848 on Build GRCh38
rs928668261 Chr.1: 50491930-50491930 on Build GRCh38
rs771971245 Chr.1: 50491749-50491749 on Build GRCh38
rs878922768 Chr.1: 50491859-50491859 on Build GRCh38
rs886685275 Chr.1: 50491974-50491974 on Build GRCh38
rs1038279947 Chr.1: 50492045-50492045 on Build GRCh38
rs921791651 Chr.1: 50491909-50491909 on Build GRCh38
rs59743675 Chr.1: 50491713-50491713 on Build GRCh38
rs747902598 Chr.1: 50491739-50491739 on Build GRCh38
rs61781013 Chr.1: 50491726-50491726 on Build GRCh38
rs766277658 Chr.1: 50491817-50491817 on Build GRCh38
rs35112243 Chr.1: 50492069-50492069 on Build GRCh38
rs914377064 Chr.1: 50491846-50491846 on Build GRCh38
rs1045229236 Chr.1: 50492137-50492137 on Build GRCh38
rs746989957 Chr.1: 50491805-50491810 on Build GRCh38
rs933248346 Chr.1: 50491935-50491935 on Build GRCh38
rs776726405 Chr.1: 50491827-50491827 on Build GRCh38
rs893872074 Chr.1: 50492107-50492107 on Build GRCh38
rs900646768 Chr.1: 50492051-50492051 on Build GRCh38
rs543413026 Chr.1: 50491730-50491730 on Build GRCh38

View Full Product Details