Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 50584583-50585124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAF1
Entrez Gene ID
11124
Gene Name
Fas associated factor 1
Gene Aliases
CGI-03, HFAF1s, UBXD12, UBXN3A, hFAF1
UniGene
Hs.530402
Gene Chromosome Location
Chr.1: 50441263-50960264 on Build GRCh38
Transcripts
NM_007051.2 XM_017000136.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5658094 c.241+3_241+6delGAGT p.? Chr.1: 50584679-50584682 on Build GRCh38
6657956 c.873C>T p.S291S Chr.1: 50584779-50584779 on Build GRCh38
5189190 c.938C>T p.A313V Chr.1: 50584714-50584714 on Build GRCh38
1602520 c.895G>A p.A299T Chr.1: 50584757-50584757 on Build GRCh38
6657957 c.147C>T p.S49S Chr.1: 50584779-50584779 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750708658 Chr.1: 50584642-50584642 on Build GRCh38
rs201015521 Chr.1: 50584713-50584713 on Build GRCh38
rs750480004 Chr.1: 50584779-50584779 on Build GRCh38
rs769588515 Chr.1: 50584860-50584860 on Build GRCh38
rs976289404 Chr.1: 50584867-50584867 on Build GRCh38
rs779144190 Chr.1: 50584841-50584841 on Build GRCh38
rs200561819 Chr.1: 50584678-50584678 on Build GRCh38
rs71574409 Chr.1: 50585080-50585081 on Build GRCh38
rs935690941 Chr.1: 50584900-50584900 on Build GRCh38
rs767641721 Chr.1: 50584742-50584742 on Build GRCh38
rs200052941 Chr.1: 50584662-50584662 on Build GRCh38
rs756360430 Chr.1: 50584803-50584803 on Build GRCh38
rs1026228697 Chr.1: 50585057-50585057 on Build GRCh38
rs779420030 Chr.1: 50584669-50584669 on Build GRCh38
rs973624349 Chr.1: 50584835-50584835 on Build GRCh38
rs952081587 Chr.1: 50584743-50584743 on Build GRCh38
rs762757210 Chr.1: 50584719-50584719 on Build GRCh38
rs763948352 Chr.1: 50584724-50584724 on Build GRCh38
rs1054489419 Chr.1: 50584901-50584901 on Build GRCh38
rs1042687972 Chr.1: 50584890-50584890 on Build GRCh38
rs775452819 Chr.1: 50584717-50584717 on Build GRCh38
rs371330134 Chr.1: 50584714-50584714 on Build GRCh38
rs931972363 Chr.1: 50584885-50584885 on Build GRCh38
rs774729432 Chr.1: 50584722-50584724 on Build GRCh38
rs13376044 Chr.1: 50584864-50584864 on Build GRCh38
rs373303991 Chr.1: 50584852-50584852 on Build GRCh38
rs557223304 Chr.1: 50584620-50584620 on Build GRCh38
rs1035408825 Chr.1: 50584658-50584658 on Build GRCh38
rs755777064 Chr.1: 50585001-50585001 on Build GRCh38
rs113363749 Chr.1: 50584995-50584995 on Build GRCh38
rs779696655 Chr.1: 50584861-50584861 on Build GRCh38
rs17106299 Chr.1: 50584849-50584849 on Build GRCh38
rs201818355 Chr.1: 50584633-50584633 on Build GRCh38
rs17106296 Chr.1: 50584828-50584828 on Build GRCh38
rs541555206 Chr.1: 50584796-50584796 on Build GRCh38
rs755304910 Chr.1: 50584837-50584837 on Build GRCh38
rs35819162 Chr.1: 50585076-50585076 on Build GRCh38
rs945508444 Chr.1: 50584882-50584882 on Build GRCh38
rs756315303 Chr.1: 50584648-50584648 on Build GRCh38
rs1013060151 Chr.1: 50584989-50584989 on Build GRCh38
rs779360606 Chr.1: 50584802-50584802 on Build GRCh38
rs766539923 Chr.1: 50584821-50584821 on Build GRCh38
rs541804534 Chr.1: 50585048-50585048 on Build GRCh38
rs913053357 Chr.1: 50585015-50585015 on Build GRCh38
rs577174710 Chr.1: 50584992-50584992 on Build GRCh38
rs911941154 Chr.1: 50584910-50584910 on Build GRCh38
rs774276657 Chr.1: 50584727-50584727 on Build GRCh38
rs757689865 Chr.1: 50584795-50584795 on Build GRCh38
rs563691620 Chr.1: 50584968-50584968 on Build GRCh38
rs1012761786 Chr.1: 50584626-50584626 on Build GRCh38
rs183436561 Chr.1: 50584664-50584664 on Build GRCh38
rs758954819 Chr.1: 50584848-50584848 on Build GRCh38
rs561096847 Chr.1: 50584675-50584675 on Build GRCh38
rs748750319 Chr.1: 50584845-50584845 on Build GRCh38
rs905790905 Chr.1: 50585061-50585061 on Build GRCh38
rs923410001 Chr.1: 50584881-50584881 on Build GRCh38
rs573049422 Chr.1: 50584685-50584685 on Build GRCh38
rs1024042699 Chr.1: 50585023-50585023 on Build GRCh38
rs779523601 Chr.1: 50585037-50585037 on Build GRCh38
rs776272769 Chr.1: 50584711-50584711 on Build GRCh38
rs575493791 Chr.1: 50584623-50584623 on Build GRCh38
rs542895560 Chr.1: 50584660-50584660 on Build GRCh38
rs770635020 Chr.1: 50584701-50584701 on Build GRCh38
rs1052565850 Chr.1: 50584996-50584996 on Build GRCh38
rs74526277 Chr.1: 50584687-50584687 on Build GRCh38
rs72899002 Chr.1: 50585087-50585087 on Build GRCh38
rs187976565 Chr.1: 50584899-50584899 on Build GRCh38

View Full Product Details