Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54146495-54146994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT3
Entrez Gene ID
4849
Gene Name
CCR4-NOT transcription complex subunit 3
Gene Aliases
LENG2, NOT3, NOT3H
UniGene
Hs.343571
Gene Chromosome Location
Chr.19: 54137689-54155708 on Build GRCh38
Transcripts
NM_014516.3 XM_005278279.1 XM_005278280.2 XM_005278281.1 XM_005278282.2 XM_011526992.1 XM_011526993.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7147037 c.304G>C p.E102Q Chr.19: 54146610-54146610 on Build GRCh38
4673193 c.859A>G p.K287E Chr.19: 54146622-54146622 on Build GRCh38
4673194 c.316A>G p.K106E Chr.19: 54146622-54146622 on Build GRCh38
7416940 c.884A>C p.E295A Chr.19: 54146647-54146647 on Build GRCh38
5476114 c.868C>T p.R290C Chr.19: 54146631-54146631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049512525 Chr.19: 54146819-54146819 on Build GRCh38
rs902340630 Chr.19: 54146851-54146851 on Build GRCh38
rs587600619 Chr.19: 54146947-54146947 on Build GRCh38
rs758483098 Chr.19: 54146690-54146690 on Build GRCh38
rs1010762309 Chr.19: 54146841-54146841 on Build GRCh38
rs922331349 Chr.19: 54146773-54146773 on Build GRCh38
rs748387536 Chr.19: 54146560-54146560 on Build GRCh38
rs755080158 Chr.19: 54146686-54146686 on Build GRCh38
rs587735637 Chr.19: 54146891-54146891 on Build GRCh38
rs201797045 Chr.19: 54146676-54146676 on Build GRCh38
rs192978934 Chr.19: 54146734-54146734 on Build GRCh38
rs374160737 Chr.19: 54146571-54146571 on Build GRCh38
rs370716584 Chr.19: 54146737-54146737 on Build GRCh38
rs185667295 Chr.19: 54146888-54146888 on Build GRCh38
rs748274382 Chr.19: 54146689-54146689 on Build GRCh38
rs748066954 Chr.19: 54146695-54146697 on Build GRCh38
rs370365731 Chr.19: 54146623-54146623 on Build GRCh38
rs762533463 Chr.19: 54146608-54146608 on Build GRCh38
rs1035062119 Chr.19: 54146540-54146540 on Build GRCh38
rs999836741 Chr.19: 54146528-54146528 on Build GRCh38
rs201212915 Chr.19: 54146591-54146591 on Build GRCh38
rs761311036 Chr.19: 54146632-54146632 on Build GRCh38
rs111776800 Chr.19: 54146658-54146658 on Build GRCh38
rs751169999 Chr.19: 54146762-54146762 on Build GRCh38
rs778089289 Chr.19: 54146705-54146705 on Build GRCh38
rs1035310992 Chr.19: 54146874-54146874 on Build GRCh38
rs587687699 Chr.19: 54146556-54146556 on Build GRCh38
rs367641242 Chr.19: 54146677-54146677 on Build GRCh38
rs144315558 Chr.19: 54146783-54146783 on Build GRCh38
rs908083001 Chr.19: 54146919-54146919 on Build GRCh38
rs768770937 Chr.19: 54146550-54146550 on Build GRCh38
rs966641520 Chr.19: 54146522-54146522 on Build GRCh38
rs766057798 Chr.19: 54146749-54146749 on Build GRCh38
rs999489539 Chr.19: 54146858-54146858 on Build GRCh38
rs762308435 Chr.19: 54146687-54146687 on Build GRCh38
rs773392238 Chr.19: 54146587-54146587 on Build GRCh38
rs79832574 Chr.19: 54146831-54146831 on Build GRCh38
rs370320885 Chr.19: 54146684-54146684 on Build GRCh38
rs751208319 Chr.19: 54146631-54146631 on Build GRCh38
rs35764464 Chr.19: 54146786-54146786 on Build GRCh38
rs940979751 Chr.19: 54146736-54146736 on Build GRCh38
rs587619299 Chr.19: 54146881-54146881 on Build GRCh38
rs984903818 Chr.19: 54146657-54146657 on Build GRCh38
rs1043609572 Chr.19: 54146848-54146848 on Build GRCh38
rs984825327 Chr.19: 54146900-54146900 on Build GRCh38
rs990925041 Chr.19: 54146579-54146579 on Build GRCh38
rs750176011 Chr.19: 54146664-54146664 on Build GRCh38
rs767281755 Chr.19: 54146934-54146934 on Build GRCh38
rs772018529 Chr.19: 54146705-54146705 on Build GRCh38
rs767199085 Chr.19: 54146648-54146648 on Build GRCh38
rs777304192 Chr.19: 54146603-54146603 on Build GRCh38
rs952156300 Chr.19: 54146896-54146896 on Build GRCh38
rs375898768 Chr.19: 54146541-54146541 on Build GRCh38
rs761127163 Chr.19: 54146589-54146589 on Build GRCh38
rs113310524 Chr.19: 54146798-54146798 on Build GRCh38
rs767231364 Chr.19: 54146886-54146888 on Build GRCh38
rs78341745 Chr.19: 54146744-54146744 on Build GRCh38

View Full Product Details