Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 52927150-52927640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECHDC2
Entrez Gene ID
55268
Gene Name
enoyl-CoA hydratase domain containing 2
Gene Aliases
-
UniGene
Hs.476319
Gene Chromosome Location
Chr.1: 52895910-52927212 on Build GRCh38
Transcripts
XM_011541709.2 XM_011541710.2 XM_011541711.2 XM_011541712.2 XM_011541721.2 XM_011541724.2 XM_011541725.2 XM_017001638.1 XM_017001641.1 NM_001007098.2 NM_001193599.1 NM_001193600.1 NM_001193617.1 NM_002979.4 XM_005271103.4 XM_011541935.2 XM_017002045.1

Gene Symbol
SCP2
Entrez Gene ID
6342
Gene Name
sterol carrier protein 2
Gene Aliases
NLTP, NSL-TP, SCP-2, SCP-CHI, SCP-X, SCPX
UniGene
Hs.476365
Gene Chromosome Location
Chr.1: 52927229-53051617 on Build GRCh38
Transcripts
XM_011541709.2 XM_011541710.2 XM_011541711.2 XM_011541712.2 XM_011541721.2 XM_011541724.2 XM_011541725.2 XM_017001638.1 XM_017001641.1 NM_001007098.2 NM_001193599.1 NM_001193600.1 NM_001193617.1 NM_002979.4 XM_005271103.4 XM_011541935.2 XM_017002045.1

SNP Detail

SNP ID Chromosome Location
rs373753458 Chr.1: 52927478-52927478 on Build GRCh38
rs954324941 Chr.1: 52927566-52927566 on Build GRCh38
rs184943760 Chr.1: 52927310-52927310 on Build GRCh38
rs1046867819 Chr.1: 52927260-52927260 on Build GRCh38
rs754517763 Chr.1: 52927373-52927373 on Build GRCh38
rs995900168 Chr.1: 52927533-52927533 on Build GRCh38
rs768084891 Chr.1: 52927371-52927371 on Build GRCh38
rs752248876 Chr.1: 52927495-52927495 on Build GRCh38
rs1026165429 Chr.1: 52927241-52927241 on Build GRCh38
rs542278215 Chr.1: 52927488-52927488 on Build GRCh38
rs773753117 Chr.1: 52927483-52927483 on Build GRCh38
rs1031845989 Chr.1: 52927507-52927507 on Build GRCh38
rs1005977610 Chr.1: 52927291-52927291 on Build GRCh38
rs115810571 Chr.1: 52927317-52927317 on Build GRCh38
rs899493265 Chr.1: 52927309-52927309 on Build GRCh38
rs1002715928 Chr.1: 52927211-52927211 on Build GRCh38
rs566646314 Chr.1: 52927300-52927300 on Build GRCh38
rs754235728 Chr.1: 52927351-52927351 on Build GRCh38
rs190197851 Chr.1: 52927430-52927430 on Build GRCh38
rs964455273 Chr.1: 52927305-52927305 on Build GRCh38
rs144609947 Chr.1: 52927564-52927564 on Build GRCh38
rs774762362 Chr.1: 52927349-52927349 on Build GRCh38
rs547050230 Chr.1: 52927296-52927296 on Build GRCh38
rs1044388788 Chr.1: 52927409-52927409 on Build GRCh38
rs938604219 Chr.1: 52927449-52927449 on Build GRCh38
rs900283647 Chr.1: 52927510-52927510 on Build GRCh38
rs990329848 Chr.1: 52927337-52927337 on Build GRCh38
rs924809433 Chr.1: 52927335-52927335 on Build GRCh38
rs147697594 Chr.1: 52927411-52927411 on Build GRCh38
rs140387282 Chr.1: 52927451-52927451 on Build GRCh38
rs746822872 Chr.1: 52927386-52927386 on Build GRCh38
rs777497403 Chr.1: 52927385-52927385 on Build GRCh38
rs1242331 Chr.1: 52927236-52927236 on Build GRCh38
rs149958725 Chr.1: 52927463-52927463 on Build GRCh38
rs772675472 Chr.1: 52927615-52927615 on Build GRCh38
rs1005429552 Chr.1: 52927298-52927298 on Build GRCh38
rs369597903 Chr.1: 52927453-52927453 on Build GRCh38
rs886232412 Chr.1: 52927277-52927277 on Build GRCh38
rs137902950 Chr.1: 52927201-52927201 on Build GRCh38
rs1044355817 Chr.1: 52927205-52927205 on Build GRCh38
rs148709071 Chr.1: 52927413-52927413 on Build GRCh38
rs907157057 Chr.1: 52927209-52927209 on Build GRCh38
rs762306060 Chr.1: 52927504-52927504 on Build GRCh38
rs990752444 Chr.1: 52927619-52927619 on Build GRCh38
rs867378228 Chr.1: 52927295-52927295 on Build GRCh38
rs763818441 Chr.1: 52927513-52927513 on Build GRCh38
rs772389056 Chr.1: 52927480-52927480 on Build GRCh38
rs755353816 Chr.1: 52927431-52927431 on Build GRCh38
rs915526566 Chr.1: 52927342-52927342 on Build GRCh38
rs764562702 Chr.1: 52927571-52927571 on Build GRCh38
rs1032378019 Chr.1: 52927606-52927606 on Build GRCh38
rs745760488 Chr.1: 52927403-52927403 on Build GRCh38
rs867761855 Chr.1: 52927173-52927173 on Build GRCh38
rs927949529 Chr.1: 52927605-52927605 on Build GRCh38
rs867975186 Chr.1: 52927455-52927455 on Build GRCh38
rs927968551 Chr.1: 52927358-52927358 on Build GRCh38
rs750884412 Chr.1: 52927372-52927372 on Build GRCh38
rs772379139 Chr.1: 52927318-52927318 on Build GRCh38
rs906643489 Chr.1: 52927238-52927238 on Build GRCh38
rs762708645 Chr.1: 52927354-52927354 on Build GRCh38
rs939427728 Chr.1: 52927397-52927397 on Build GRCh38
rs987438106 Chr.1: 52927323-52927323 on Build GRCh38
rs573557498 Chr.1: 52927436-52927436 on Build GRCh38
rs948792228 Chr.1: 52927405-52927405 on Build GRCh38
rs756943613 Chr.1: 52927387-52927387 on Build GRCh38
rs764696541 Chr.1: 52927376-52927376 on Build GRCh38
rs956437477 Chr.1: 52927336-52927336 on Build GRCh38
rs761148476 Chr.1: 52927486-52927486 on Build GRCh38
rs1011055309 Chr.1: 52927556-52927556 on Build GRCh38
rs941210130 Chr.1: 52927418-52927418 on Build GRCh38
rs748713155 Chr.1: 52927586-52927586 on Build GRCh38
rs532306192 Chr.1: 52927600-52927600 on Build GRCh38
rs781299267 Chr.1: 52927400-52927400 on Build GRCh38
rs981008602 Chr.1: 52927579-52927579 on Build GRCh38
rs999190474 Chr.1: 52927598-52927598 on Build GRCh38
rs779585406 Chr.1: 52927475-52927475 on Build GRCh38
rs780256464 Chr.1: 52927417-52927417 on Build GRCh38
rs140752743 Chr.1: 52927340-52927340 on Build GRCh38
rs1048072495 Chr.1: 52927458-52927458 on Build GRCh38
rs758057676 Chr.1: 52927377-52927377 on Build GRCh38

View Full Product Details