Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53594008-53594500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLIS1
Entrez Gene ID
148979
Gene Name
GLIS family zinc finger 1
Gene Aliases
-
UniGene
Hs.306691
Gene Chromosome Location
Chr.1: 53506233-53739171 on Build GRCh38
Transcripts
NM_147193.2 XM_017000408.1 XM_017000409.1 XM_017000410.1 XM_017000411.1 XM_017000412.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4988592 c.593G>A p.R198H Chr.1: 53594310-53594310 on Build GRCh38
6671612 c.416G>A p.C139Y Chr.1: 53594487-53594487 on Build GRCh38
5901033 c.772G>A p.G258S Chr.1: 53594131-53594131 on Build GRCh38
291390 c.670C>A p.R224S Chr.1: 53594233-53594233 on Build GRCh38
4008896 c.761G>A p.R254Q Chr.1: 53594142-53594142 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1032416709 Chr.1: 53594213-53594213 on Build GRCh38
rs748404280 Chr.1: 53594299-53594299 on Build GRCh38
rs766095000 Chr.1: 53594455-53594455 on Build GRCh38
rs376755747 Chr.1: 53594403-53594403 on Build GRCh38
rs142216816 Chr.1: 53594171-53594171 on Build GRCh38
rs372329639 Chr.1: 53594429-53594429 on Build GRCh38
rs111547294 Chr.1: 53594376-53594376 on Build GRCh38
rs867195754 Chr.1: 53594398-53594398 on Build GRCh38
rs759207828 Chr.1: 53594091-53594091 on Build GRCh38
rs779059013 Chr.1: 53594414-53594414 on Build GRCh38
rs779399927 Chr.1: 53594478-53594478 on Build GRCh38
rs756693688 Chr.1: 53594255-53594255 on Build GRCh38
rs752683116 Chr.1: 53594469-53594469 on Build GRCh38
rs745827457 Chr.1: 53594147-53594147 on Build GRCh38
rs201526886 Chr.1: 53594386-53594386 on Build GRCh38
rs1007822292 Chr.1: 53594339-53594339 on Build GRCh38
rs897287635 Chr.1: 53594096-53594096 on Build GRCh38
rs1005828459 Chr.1: 53594172-53594172 on Build GRCh38
rs763221218 Chr.1: 53594110-53594110 on Build GRCh38
rs775513632 Chr.1: 53594109-53594109 on Build GRCh38
rs777253978 Chr.1: 53594071-53594071 on Build GRCh38
rs375635801 Chr.1: 53594381-53594381 on Build GRCh38
rs193201988 Chr.1: 53594323-53594323 on Build GRCh38
rs57378215 Chr.1: 53594184-53594184 on Build GRCh38
rs774675510 Chr.1: 53594286-53594286 on Build GRCh38
rs370468534 Chr.1: 53594254-53594254 on Build GRCh38
rs761232530 Chr.1: 53594232-53594232 on Build GRCh38
rs749938080 Chr.1: 53594330-53594330 on Build GRCh38
rs370494270 Chr.1: 53594382-53594382 on Build GRCh38
rs777772667 Chr.1: 53594095-53594095 on Build GRCh38
rs140752554 Chr.1: 53594470-53594470 on Build GRCh38
rs747050302 Chr.1: 53594380-53594380 on Build GRCh38
rs550452713 Chr.1: 53594091-53594097 on Build GRCh38
rs201030759 Chr.1: 53594081-53594081 on Build GRCh38
rs35227000 Chr.1: 53594343-53594343 on Build GRCh38
rs766894332 Chr.1: 53594315-53594315 on Build GRCh38
rs374386897 Chr.1: 53594118-53594118 on Build GRCh38
rs750997565 Chr.1: 53594126-53594126 on Build GRCh38
rs765206704 Chr.1: 53594463-53594463 on Build GRCh38
rs878859840 Chr.1: 53594194-53594194 on Build GRCh38
rs1051840380 Chr.1: 53594459-53594459 on Build GRCh38
rs200512169 Chr.1: 53594430-53594430 on Build GRCh38
rs184554996 Chr.1: 53594454-53594454 on Build GRCh38
rs36070063 Chr.1: 53594409-53594409 on Build GRCh38
rs368627108 Chr.1: 53594458-53594458 on Build GRCh38
rs917416018 Chr.1: 53594240-53594240 on Build GRCh38
rs751657312 Chr.1: 53594116-53594116 on Build GRCh38
rs767709665 Chr.1: 53594226-53594226 on Build GRCh38
rs77195406 Chr.1: 53594141-53594141 on Build GRCh38
rs562792631 Chr.1: 53594453-53594453 on Build GRCh38
rs147448341 Chr.1: 53594202-53594202 on Build GRCh38
rs1030668665 Chr.1: 53594195-53594195 on Build GRCh38
rs773361849 Chr.1: 53594379-53594379 on Build GRCh38
rs771427108 Chr.1: 53594066-53594066 on Build GRCh38
rs751276401 Chr.1: 53594373-53594373 on Build GRCh38
rs755560525 Chr.1: 53594428-53594428 on Build GRCh38
rs754934554 Chr.1: 53594410-53594410 on Build GRCh38
rs755594736 Chr.1: 53594338-53594338 on Build GRCh38
rs201038184 Chr.1: 53594283-53594283 on Build GRCh38
rs772471069 Chr.1: 53594310-53594310 on Build GRCh38
rs139516969 Chr.1: 53594436-53594436 on Build GRCh38
rs770827651 Chr.1: 53594083-53594083 on Build GRCh38
rs150235436 Chr.1: 53594456-53594456 on Build GRCh38
rs763211371 Chr.1: 53594054-53594054 on Build GRCh38
rs764287713 Chr.1: 53594111-53594111 on Build GRCh38
rs745412773 Chr.1: 53594263-53594263 on Build GRCh38
rs1048147750 Chr.1: 53594334-53594334 on Build GRCh38
rs193059626 Chr.1: 53594029-53594029 on Build GRCh38
rs145199173 Chr.1: 53594142-53594142 on Build GRCh38
rs778828067 Chr.1: 53594368-53594368 on Build GRCh38
rs760192516 Chr.1: 53594407-53594407 on Build GRCh38
rs902458303 Chr.1: 53594375-53594375 on Build GRCh38
rs539985102 Chr.1: 53594239-53594239 on Build GRCh38
rs780480693 Chr.1: 53594133-53594133 on Build GRCh38
rs773798489 Chr.1: 53594227-53594227 on Build GRCh38
rs758279382 Chr.1: 53594417-53594417 on Build GRCh38
rs769631886 Chr.1: 53594092-53594092 on Build GRCh38
rs916598641 Chr.1: 53594056-53594056 on Build GRCh38
rs535647382 Chr.1: 53594078-53594078 on Build GRCh38
rs762158343 Chr.1: 53594401-53594401 on Build GRCh38
rs569988372 Chr.1: 53594327-53594327 on Build GRCh38
rs750049615 Chr.1: 53594404-53594404 on Build GRCh38
rs760018104 Chr.1: 53594391-53594391 on Build GRCh38
rs768935299 Chr.1: 53594439-53594439 on Build GRCh38
rs749215456 Chr.1: 53594359-53594359 on Build GRCh38
rs766864383 Chr.1: 53594233-53594233 on Build GRCh38
rs997401513 Chr.1: 53594190-53594190 on Build GRCh38
rs766719877 Chr.1: 53594100-53594100 on Build GRCh38
rs762031054 Chr.1: 53594117-53594117 on Build GRCh38
rs765828847 Chr.1: 53594408-53594408 on Build GRCh38
rs35288204 Chr.1: 53594132-53594132 on Build GRCh38
rs747842241 Chr.1: 53594062-53594062 on Build GRCh38
rs200339816 Chr.1: 53594258-53594258 on Build GRCh38
rs770329114 Chr.1: 53594402-53594402 on Build GRCh38
rs149706629 Chr.1: 53594322-53594322 on Build GRCh38
rs754771890 Chr.1: 53594360-53594360 on Build GRCh38
rs1038760577 Chr.1: 53594332-53594332 on Build GRCh38
rs34961060 Chr.1: 53594434-53594434 on Build GRCh38
rs763691093 Chr.1: 53594246-53594246 on Build GRCh38
rs769673939 Chr.1: 53594166-53594166 on Build GRCh38
rs780663060 Chr.1: 53594262-53594262 on Build GRCh38
rs763238878 Chr.1: 53594438-53594438 on Build GRCh38
rs375343014 Chr.1: 53594311-53594311 on Build GRCh38
rs990922501 Chr.1: 53594228-53594228 on Build GRCh38
rs781071820 Chr.1: 53594143-53594143 on Build GRCh38
rs143152577 Chr.1: 53594140-53594140 on Build GRCh38
rs770275773 Chr.1: 53594437-53594437 on Build GRCh38
rs756875095 Chr.1: 53594326-53594326 on Build GRCh38
rs977945259 Chr.1: 53594208-53594208 on Build GRCh38
rs755350809 Chr.1: 53594242-53594242 on Build GRCh38
rs145375877 Chr.1: 53594277-53594277 on Build GRCh38
rs936639156 Chr.1: 53594045-53594045 on Build GRCh38
rs949639558 Chr.1: 53594086-53594086 on Build GRCh38
rs185823869 Chr.1: 53594038-53594038 on Build GRCh38
rs200301349 Chr.1: 53594090-53594090 on Build GRCh38
rs970750183 Chr.1: 53594405-53594405 on Build GRCh38
rs761944746 Chr.1: 53594224-53594224 on Build GRCh38
rs949422881 Chr.1: 53594270-53594270 on Build GRCh38
rs756076762 Chr.1: 53594274-53594274 on Build GRCh38
rs140770740 Chr.1: 53594188-53594188 on Build GRCh38
rs776603321 Chr.1: 53594085-53594085 on Build GRCh38
rs1048572803 Chr.1: 53594316-53594316 on Build GRCh38

View Full Product Details