Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 47369234-47369766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPCAM
Entrez Gene ID
4072
Gene Name
epithelial cell adhesion molecule
Gene Aliases
DIAR5, EGP-2, EGP314, EGP40, ESA, HNPCC8, KS1/4, KSA, M4S1, MIC18, MK-1, TACSTD1, TROP1
UniGene
Hs.542050
Gene Chromosome Location
Chr.2: 47369148-47387028 on Build GRCh38
Transcripts
NM_002354.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6922710 c.70C>G p.Q24E Chr.2: 47369575-47369575 on Build GRCh38
6302867 c.49A>G p.T17A Chr.2: 47369554-47369554 on Build GRCh38
6375410 c.11C>A p.P4Q Chr.2: 47369516-47369516 on Build GRCh38
6413249 c.3G>T p.M1I Chr.2: 47369508-47369508 on Build GRCh38
7324302 c.57T>C p.T19T Chr.2: 47369562-47369562 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs878854494 Chr.2: 47369579-47369579 on Build GRCh38
rs1051432084 Chr.2: 47369328-47369328 on Build GRCh38
rs777516660 Chr.2: 47369636-47369636 on Build GRCh38
rs754801864 Chr.2: 47369358-47369358 on Build GRCh38
rs1009258421 Chr.2: 47369281-47369281 on Build GRCh38
rs1037569065 Chr.2: 47369303-47369303 on Build GRCh38
rs867177667 Chr.2: 47369521-47369521 on Build GRCh38
rs911761329 Chr.2: 47369670-47369670 on Build GRCh38
rs1046993012 Chr.2: 47369708-47369708 on Build GRCh38
rs752048938 Chr.2: 47369253-47369253 on Build GRCh38
rs774488035 Chr.2: 47369681-47369681 on Build GRCh38
rs977270594 Chr.2: 47369688-47369688 on Build GRCh38
rs903767290 Chr.2: 47369634-47369640 on Build GRCh38
rs878854490 Chr.2: 47369556-47369561 on Build GRCh38
rs1027687237 Chr.2: 47369733-47369733 on Build GRCh38
rs768375014 Chr.2: 47369283-47369283 on Build GRCh38
rs778641299 Chr.2: 47369516-47369516 on Build GRCh38
rs561842739 Chr.2: 47369398-47369398 on Build GRCh38
rs116429842 Chr.2: 47369555-47369555 on Build GRCh38
rs572608653 Chr.2: 47369285-47369285 on Build GRCh38
rs961670779 Chr.2: 47369548-47369548 on Build GRCh38
rs774722931 Chr.2: 47369587-47369587 on Build GRCh38
rs746990000 Chr.2: 47369539-47369539 on Build GRCh38
rs757917638 Chr.2: 47369723-47369723 on Build GRCh38
rs776646187 Chr.2: 47369543-47369543 on Build GRCh38
rs1032509550 Chr.2: 47369365-47369365 on Build GRCh38
rs1025227886 Chr.2: 47369388-47369388 on Build GRCh38
rs769957791 Chr.2: 47369655-47369655 on Build GRCh38
rs907976188 Chr.2: 47369291-47369291 on Build GRCh38
rs373746049 Chr.2: 47369727-47369727 on Build GRCh38
rs953510420 Chr.2: 47369668-47369668 on Build GRCh38
rs587780772 Chr.2: 47369512-47369512 on Build GRCh38
rs762123721 Chr.2: 47369602-47369602 on Build GRCh38
rs773336656 Chr.2: 47369486-47369486 on Build GRCh38
rs878854486 Chr.2: 47369525-47369525 on Build GRCh38
rs567762201 Chr.2: 47369362-47369362 on Build GRCh38
rs773832274 Chr.2: 47369697-47369697 on Build GRCh38
rs549177672 Chr.2: 47369568-47369568 on Build GRCh38
rs747738988 Chr.2: 47369518-47369518 on Build GRCh38
rs538922283 Chr.2: 47369729-47369729 on Build GRCh38
rs776827466 Chr.2: 47369500-47369500 on Build GRCh38
rs748991934 Chr.2: 47369280-47369280 on Build GRCh38
rs374563131 Chr.2: 47369511-47369511 on Build GRCh38
rs879892379 Chr.2: 47369738-47369738 on Build GRCh38
rs771965334 Chr.2: 47369522-47369522 on Build GRCh38
rs971034215 Chr.2: 47369499-47369499 on Build GRCh38
rs1000967615 Chr.2: 47369363-47369363 on Build GRCh38
rs973230051 Chr.2: 47369420-47369420 on Build GRCh38
rs758024258 Chr.2: 47369612-47369616 on Build GRCh38
rs764848112 Chr.2: 47369705-47369705 on Build GRCh38
rs863224709 Chr.2: 47369533-47369533 on Build GRCh38
rs983485905 Chr.2: 47369393-47369393 on Build GRCh38
rs763055107 Chr.2: 47369576-47369576 on Build GRCh38
rs900402744 Chr.2: 47369312-47369312 on Build GRCh38
rs199679894 Chr.2: 47369610-47369610 on Build GRCh38
rs1041354853 Chr.2: 47369536-47369536 on Build GRCh38
rs1015195227 Chr.2: 47369624-47369624 on Build GRCh38
rs894237303 Chr.2: 47369570-47369570 on Build GRCh38
rs1052079647 Chr.2: 47369437-47369437 on Build GRCh38
rs768364603 Chr.2: 47369647-47369648 on Build GRCh38
rs144077629 Chr.2: 47369714-47369715 on Build GRCh38
rs370806451 Chr.2: 47369607-47369607 on Build GRCh38
rs764645465 Chr.2: 47369618-47369618 on Build GRCh38
rs532872105 Chr.2: 47369514-47369514 on Build GRCh38
rs1046612207 Chr.2: 47369633-47369634 on Build GRCh38
rs927382710 Chr.2: 47369588-47369588 on Build GRCh38
rs1000530194 Chr.2: 47369489-47369489 on Build GRCh38
rs959636784 Chr.2: 47369261-47369261 on Build GRCh38
rs371271353 Chr.2: 47369259-47369259 on Build GRCh38
rs764492954 Chr.2: 47369581-47369581 on Build GRCh38
rs747979626 Chr.2: 47369466-47369466 on Build GRCh38
rs935639571 Chr.2: 47369690-47369690 on Build GRCh38
rs890085795 Chr.2: 47369345-47369345 on Build GRCh38
rs990831794 Chr.2: 47369277-47369277 on Build GRCh38
rs745792461 Chr.2: 47369643-47369643 on Build GRCh38
rs1009909046 Chr.2: 47369348-47369348 on Build GRCh38
rs960608240 Chr.2: 47369262-47369262 on Build GRCh38
rs768899723 Chr.2: 47369663-47369663 on Build GRCh38
rs941815712 Chr.2: 47369302-47369302 on Build GRCh38
rs895265671 Chr.2: 47369472-47369472 on Build GRCh38
rs886056129 Chr.2: 47369344-47369344 on Build GRCh38
rs921798972 Chr.2: 47369423-47369423 on Build GRCh38
rs1020636182 Chr.2: 47369478-47369478 on Build GRCh38
rs749341277 Chr.2: 47369661-47369661 on Build GRCh38
rs767854724 Chr.2: 47369693-47369693 on Build GRCh38
rs746695351 Chr.2: 47369641-47369647 on Build GRCh38
rs771025657 Chr.2: 47369541-47369541 on Build GRCh38
rs1041386144 Chr.2: 47369451-47369451 on Build GRCh38
rs757063495 Chr.2: 47369640-47369640 on Build GRCh38
rs750836547 Chr.2: 47369609-47369609 on Build GRCh38
rs562515601 Chr.2: 47369492-47369492 on Build GRCh38
rs931338743 Chr.2: 47369561-47369561 on Build GRCh38
rs879135112 Chr.2: 47369592-47369592 on Build GRCh38
rs954246217 Chr.2: 47369346-47369346 on Build GRCh38
rs780639335 Chr.2: 47369678-47369679 on Build GRCh38
rs752808238 Chr.2: 47369506-47369506 on Build GRCh38
rs769754089 Chr.2: 47369556-47369556 on Build GRCh38
rs971111660 Chr.2: 47369390-47369390 on Build GRCh38
rs779020022 Chr.2: 47369484-47369485 on Build GRCh38
rs36094436 Chr.2: 47369641-47369642 on Build GRCh38
rs912839724 Chr.2: 47369364-47369364 on Build GRCh38
rs945689704 Chr.2: 47369443-47369443 on Build GRCh38
rs925890818 Chr.2: 47369689-47369689 on Build GRCh38
rs752645618 Chr.2: 47369678-47369678 on Build GRCh38
rs550574145 Chr.2: 47369457-47369457 on Build GRCh38
rs959607406 Chr.2: 47369370-47369370 on Build GRCh38
rs886056130 Chr.2: 47369395-47369395 on Build GRCh38
rs991444746 Chr.2: 47369381-47369381 on Build GRCh38
rs539900942 Chr.2: 47369317-47369317 on Build GRCh38
rs781340035 Chr.2: 47369642-47369642 on Build GRCh38
rs752168467 Chr.2: 47369711-47369711 on Build GRCh38
rs921451344 Chr.2: 47369338-47369338 on Build GRCh38
rs938734907 Chr.2: 47369591-47369591 on Build GRCh38
rs864622092 Chr.2: 47369543-47369563 on Build GRCh38
rs878854487 Chr.2: 47369526-47369526 on Build GRCh38
rs537973815 Chr.2: 47369620-47369620 on Build GRCh38
rs528859440 Chr.2: 47369408-47369408 on Build GRCh38
rs760681394 Chr.2: 47369491-47369491 on Build GRCh38
rs1012434428 Chr.2: 47369482-47369482 on Build GRCh38
rs863224712 Chr.2: 47369586-47369586 on Build GRCh38
rs757963053 Chr.2: 47369630-47369630 on Build GRCh38
rs949871689 Chr.2: 47369701-47369701 on Build GRCh38
rs1034069212 Chr.2: 47369469-47369469 on Build GRCh38
rs762178837 Chr.2: 47369682-47369682 on Build GRCh38
rs779614438 Chr.2: 47369624-47369625 on Build GRCh38
rs777677935 Chr.2: 47369529-47369529 on Build GRCh38
rs112970128 Chr.2: 47369286-47369286 on Build GRCh38
rs1029464211 Chr.2: 47369651-47369651 on Build GRCh38
rs755689254 Chr.2: 47369254-47369254 on Build GRCh38
rs1048235899 Chr.2: 47369289-47369289 on Build GRCh38
rs987706438 Chr.2: 47369669-47369669 on Build GRCh38
rs1020424576 Chr.2: 47369356-47369356 on Build GRCh38
rs886056128 Chr.2: 47369303-47369303 on Build GRCh38
rs924274340 Chr.2: 47369440-47369440 on Build GRCh38
rs534099440 Chr.2: 47369611-47369612 on Build GRCh38
rs931844088 Chr.2: 47369427-47369427 on Build GRCh38
rs746808024 Chr.2: 47369638-47369638 on Build GRCh38
rs556133821 Chr.2: 47369660-47369660 on Build GRCh38
rs868121750 Chr.2: 47369623-47369623 on Build GRCh38
rs533255660 Chr.2: 47369505-47369505 on Build GRCh38
rs771705322 Chr.2: 47369484-47369484 on Build GRCh38
rs973357252 Chr.2: 47369554-47369554 on Build GRCh38
rs1005172606 Chr.2: 47369622-47369622 on Build GRCh38
rs945026534 Chr.2: 47369333-47369334 on Build GRCh38
rs201402370 Chr.2: 47369510-47369510 on Build GRCh38
rs886056131 Chr.2: 47369427-47369427 on Build GRCh38
rs376344699 Chr.2: 47369314-47369314 on Build GRCh38

View Full Product Details