Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 53774820-53775348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB3
Entrez Gene ID
51130
Gene Name
ankyrin repeat and SOCS box containing 3
Gene Aliases
ASB-3
UniGene
Hs.40763
Gene Chromosome Location
Chr.2: 53669980-53787009 on Build GRCh38
Transcripts
NM_001201965.1 NM_016115.4 NM_145863.2 NM_001008708.2 NM_001164165.1

Gene Symbol
CHAC2
Entrez Gene ID
494143
Gene Name
ChaC cation transport regulator homolog 2
Gene Aliases
-
UniGene
Hs.585944
Gene Chromosome Location
Chr.2: 53767792-53775150 on Build GRCh38
Transcripts
NM_001201965.1 NM_016115.4 NM_145863.2 NM_001008708.2 NM_001164165.1

Gene Symbol
GPR75-ASB3
Entrez Gene ID
100302652
Gene Name
GPR75-ASB3 readthrough
Gene Aliases
-
UniGene
Hs.40763
Gene Chromosome Location
Chr.2: 53670293-53860033 on Build GRCh38
Transcripts
NM_001201965.1 NM_016115.4 NM_145863.2 NM_001008708.2 NM_001164165.1

SNP Detail

SNP ID Chromosome Location
rs2542581 Chr.2: 53774927-53774927 on Build GRCh38
rs890037811 Chr.2: 53774889-53774889 on Build GRCh38
rs35914824 Chr.2: 53775288-53775289 on Build GRCh38
rs955618384 Chr.2: 53775189-53775189 on Build GRCh38
rs894815300 Chr.2: 53775102-53775102 on Build GRCh38
rs1048089166 Chr.2: 53775002-53775002 on Build GRCh38
rs941401212 Chr.2: 53775220-53775220 on Build GRCh38
rs7586453 Chr.2: 53775131-53775131 on Build GRCh38
rs978306159 Chr.2: 53775173-53775173 on Build GRCh38
rs750218629 Chr.2: 53775304-53775304 on Build GRCh38
rs1049912532 Chr.2: 53775116-53775117 on Build GRCh38
rs906427457 Chr.2: 53775043-53775043 on Build GRCh38
rs567217961 Chr.2: 53774894-53774894 on Build GRCh38
rs1015240645 Chr.2: 53775315-53775318 on Build GRCh38
rs564475953 Chr.2: 53775061-53775061 on Build GRCh38
rs775635151 Chr.2: 53775112-53775114 on Build GRCh38
rs1010642088 Chr.2: 53775193-53775193 on Build GRCh38
rs986121348 Chr.2: 53775203-53775203 on Build GRCh38
rs985451221 Chr.2: 53774903-53774903 on Build GRCh38
rs933965687 Chr.2: 53774902-53774902 on Build GRCh38
rs767387118 Chr.2: 53775222-53775222 on Build GRCh38
rs929184237 Chr.2: 53774977-53774977 on Build GRCh38
rs536067228 Chr.2: 53774909-53774909 on Build GRCh38
rs551163136 Chr.2: 53774980-53774980 on Build GRCh38
rs922906061 Chr.2: 53774895-53774895 on Build GRCh38
rs1047529864 Chr.2: 53775272-53775272 on Build GRCh38
rs940445785 Chr.2: 53775064-53775064 on Build GRCh38
rs6736808 Chr.2: 53775079-53775079 on Build GRCh38
rs1022086373 Chr.2: 53775163-53775163 on Build GRCh38
rs966852989 Chr.2: 53775165-53775165 on Build GRCh38
rs533562303 Chr.2: 53775105-53775105 on Build GRCh38
rs1030259785 Chr.2: 53775180-53775180 on Build GRCh38
rs927253892 Chr.2: 53774930-53774930 on Build GRCh38
rs930070998 Chr.2: 53775259-53775259 on Build GRCh38
rs541823225 Chr.2: 53774852-53774852 on Build GRCh38
rs918695448 Chr.2: 53775250-53775250 on Build GRCh38
rs922996956 Chr.2: 53775109-53775109 on Build GRCh38
rs898996686 Chr.2: 53774946-53774946 on Build GRCh38
rs764123225 Chr.2: 53774886-53774886 on Build GRCh38
rs1010297668 Chr.2: 53774922-53774922 on Build GRCh38
rs984693777 Chr.2: 53775273-53775273 on Build GRCh38
rs1033310688 Chr.2: 53775078-53775078 on Build GRCh38
rs555922066 Chr.2: 53774913-53774913 on Build GRCh38
rs890050760 Chr.2: 53775123-53775123 on Build GRCh38
rs561786853 Chr.2: 53774926-53774926 on Build GRCh38
rs951928523 Chr.2: 53775039-53775039 on Build GRCh38
rs911230936 Chr.2: 53774918-53774918 on Build GRCh38
rs934186216 Chr.2: 53775114-53775115 on Build GRCh38
rs961760457 Chr.2: 53775316-53775316 on Build GRCh38
rs189451949 Chr.2: 53774844-53774844 on Build GRCh38
rs181537428 Chr.2: 53775211-53775211 on Build GRCh38

View Full Product Details