Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 55288876-55289415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC88A
Entrez Gene ID
55704
Gene Name
coiled-coil domain containing 88A
Gene Aliases
APE, GIRDIN, GIV, GRDN, HkRP1, KIAA1212
UniGene
Hs.292925
Gene Chromosome Location
Chr.2: 55287817-55419921 on Build GRCh38
Transcripts
NM_001135597.1 NM_001254943.1 NM_018084.4 XM_005264421.2 XM_011532965.2 XM_011532966.2 XM_011532967.2 XM_011532968.2 XM_017004481.1

SNP Detail

SNP ID Chromosome Location
rs531834073 Chr.2: 55289379-55289379 on Build GRCh38
rs773880059 Chr.2: 55289094-55289094 on Build GRCh38
rs1011890525 Chr.2: 55289265-55289265 on Build GRCh38
rs560160195 Chr.2: 55289247-55289247 on Build GRCh38
rs529240313 Chr.2: 55289287-55289287 on Build GRCh38
rs886382092 Chr.2: 55288960-55288960 on Build GRCh38
rs978926513 Chr.2: 55289051-55289051 on Build GRCh38
rs778830841 Chr.2: 55289375-55289375 on Build GRCh38
rs954894854 Chr.2: 55289082-55289082 on Build GRCh38
rs376551012 Chr.2: 55289024-55289024 on Build GRCh38
rs911897322 Chr.2: 55289107-55289107 on Build GRCh38
rs900354572 Chr.2: 55288934-55288934 on Build GRCh38
rs1039057208 Chr.2: 55288911-55288911 on Build GRCh38
rs938491566 Chr.2: 55289318-55289318 on Build GRCh38
rs759157498 Chr.2: 55289182-55289182 on Build GRCh38
rs542670262 Chr.2: 55289104-55289104 on Build GRCh38
rs77490309 Chr.2: 55289072-55289072 on Build GRCh38
rs930704134 Chr.2: 55289167-55289167 on Build GRCh38
rs896150150 Chr.2: 55288981-55288981 on Build GRCh38
rs916604519 Chr.2: 55289197-55289197 on Build GRCh38
rs1037398395 Chr.2: 55289006-55289006 on Build GRCh38
rs1005870568 Chr.2: 55289004-55289004 on Build GRCh38
rs549101437 Chr.2: 55289332-55289332 on Build GRCh38
rs896161489 Chr.2: 55289249-55289249 on Build GRCh38
rs903623393 Chr.2: 55289299-55289299 on Build GRCh38
rs974741108 Chr.2: 55289385-55289385 on Build GRCh38
rs769418059 Chr.2: 55288910-55288910 on Build GRCh38
rs12619486 Chr.2: 55288967-55288967 on Build GRCh38
rs533571213 Chr.2: 55289033-55289033 on Build GRCh38
rs1056356120 Chr.2: 55289199-55289199 on Build GRCh38
rs974813290 Chr.2: 55289143-55289143 on Build GRCh38
rs956364468 Chr.2: 55289365-55289365 on Build GRCh38
rs753657973 Chr.2: 55289345-55289345 on Build GRCh38
rs540271318 Chr.2: 55289213-55289213 on Build GRCh38
rs2589113 Chr.2: 55289187-55289187 on Build GRCh38
rs186635834 Chr.2: 55289055-55289055 on Build GRCh38
rs970643482 Chr.2: 55289046-55289046 on Build GRCh38
rs886227118 Chr.2: 55288990-55288990 on Build GRCh38
rs74971692 Chr.2: 55289064-55289064 on Build GRCh38
rs922016456 Chr.2: 55289162-55289162 on Build GRCh38
rs866070151 Chr.2: 55289183-55289183 on Build GRCh38
rs1031028415 Chr.2: 55289175-55289175 on Build GRCh38
rs772222152 Chr.2: 55289114-55289115 on Build GRCh38
rs1034718947 Chr.2: 55288980-55288980 on Build GRCh38
rs75314859 Chr.2: 55289136-55289136 on Build GRCh38
rs868228772 Chr.2: 55289305-55289305 on Build GRCh38
rs569281591 Chr.2: 55289341-55289341 on Build GRCh38
rs1049469301 Chr.2: 55288955-55288955 on Build GRCh38
rs144150568 Chr.2: 55289198-55289198 on Build GRCh38
rs572263189 Chr.2: 55289206-55289206 on Build GRCh38
rs6545485 Chr.2: 55289366-55289366 on Build GRCh38
rs948677776 Chr.2: 55288935-55288935 on Build GRCh38
rs1047503959 Chr.2: 55288982-55288982 on Build GRCh38
rs1012005123 Chr.2: 55289010-55289010 on Build GRCh38
rs980050133 Chr.2: 55289236-55289236 on Build GRCh38
rs1045613 Chr.2: 55288954-55288954 on Build GRCh38
rs1044852970 Chr.2: 55289272-55289272 on Build GRCh38
rs766191434 Chr.2: 55289326-55289330 on Build GRCh38
rs569149179 Chr.2: 55288946-55288946 on Build GRCh38
rs760458378 Chr.2: 55288951-55288954 on Build GRCh38

View Full Product Details