Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 55289941-55290487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC88A
Entrez Gene ID
55704
Gene Name
coiled-coil domain containing 88A
Gene Aliases
APE, GIRDIN, GIV, GRDN, HkRP1, KIAA1212
UniGene
Hs.292925
Gene Chromosome Location
Chr.2: 55287817-55419921 on Build GRCh38
Transcripts
NM_001135597.1 NM_001254943.1 NM_018084.4 XM_005264421.2 XM_011532965.2 XM_011532966.2 XM_011532967.2 XM_011532968.2 XM_017004481.1

SNP Detail

SNP ID Chromosome Location
rs976482352 Chr.2: 55290351-55290351 on Build GRCh38
rs1025035852 Chr.2: 55290187-55290187 on Build GRCh38
rs1004204598 Chr.2: 55290361-55290361 on Build GRCh38
rs17347807 Chr.2: 55290151-55290151 on Build GRCh38
rs1052629346 Chr.2: 55289983-55289983 on Build GRCh38
rs754755044 Chr.2: 55290012-55290015 on Build GRCh38
rs1036576581 Chr.2: 55290383-55290383 on Build GRCh38
rs932112563 Chr.2: 55290123-55290123 on Build GRCh38
rs925169781 Chr.2: 55290038-55290038 on Build GRCh38
rs144826545 Chr.2: 55290357-55290357 on Build GRCh38
rs888047743 Chr.2: 55290144-55290144 on Build GRCh38
rs17046828 Chr.2: 55289986-55289986 on Build GRCh38
rs1050716802 Chr.2: 55290128-55290128 on Build GRCh38
rs995167203 Chr.2: 55290426-55290426 on Build GRCh38
rs981037647 Chr.2: 55290247-55290247 on Build GRCh38
rs980728030 Chr.2: 55290036-55290036 on Build GRCh38
rs763745123 Chr.2: 55290060-55290060 on Build GRCh38
rs969268278 Chr.2: 55290189-55290189 on Build GRCh38
rs1047948 Chr.2: 55290355-55290355 on Build GRCh38
rs17046829 Chr.2: 55290083-55290083 on Build GRCh38
rs760386464 Chr.2: 55290048-55290048 on Build GRCh38
rs964982401 Chr.2: 55290333-55290333 on Build GRCh38
rs773223623 Chr.2: 55290058-55290059 on Build GRCh38
rs748285809 Chr.2: 55290126-55290126 on Build GRCh38
rs899059647 Chr.2: 55290391-55290392 on Build GRCh38
rs1038393577 Chr.2: 55290372-55290372 on Build GRCh38
rs569653151 Chr.2: 55290269-55290269 on Build GRCh38
rs897826123 Chr.2: 55290170-55290170 on Build GRCh38
rs1040733090 Chr.2: 55290112-55290112 on Build GRCh38
rs567930277 Chr.2: 55290117-55290117 on Build GRCh38
rs909374404 Chr.2: 55290364-55290364 on Build GRCh38
rs202194714 Chr.2: 55290093-55290093 on Build GRCh38
rs1013678273 Chr.2: 55290182-55290182 on Build GRCh38
rs533580704 Chr.2: 55290213-55290214 on Build GRCh38
rs973233838 Chr.2: 55290098-55290098 on Build GRCh38
rs920987204 Chr.2: 55290354-55290354 on Build GRCh38
rs77513179 Chr.2: 55290096-55290096 on Build GRCh38
rs146721738 Chr.2: 55290450-55290450 on Build GRCh38
rs750530297 Chr.2: 55290268-55290268 on Build GRCh38
rs775052347 Chr.2: 55290031-55290031 on Build GRCh38
rs780295403 Chr.2: 55290430-55290430 on Build GRCh38
rs548135278 Chr.2: 55290111-55290111 on Build GRCh38
rs936583844 Chr.2: 55290057-55290057 on Build GRCh38
rs988075623 Chr.2: 55290300-55290300 on Build GRCh38
rs751784431 Chr.2: 55290452-55290452 on Build GRCh38
rs1032163155 Chr.2: 55290251-55290251 on Build GRCh38
rs1024852873 Chr.2: 55289967-55289967 on Build GRCh38
rs958231118 Chr.2: 55290252-55290252 on Build GRCh38
rs1045211313 Chr.2: 55290214-55290214 on Build GRCh38
rs913574485 Chr.2: 55290303-55290303 on Build GRCh38
rs1047946 Chr.2: 55290377-55290377 on Build GRCh38
rs549755711 Chr.2: 55290213-55290213 on Build GRCh38
rs1014966412 Chr.2: 55290161-55290161 on Build GRCh38
rs942316249 Chr.2: 55290376-55290376 on Build GRCh38
rs915479168 Chr.2: 55290198-55290198 on Build GRCh38

View Full Product Details