Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 113931158-113931691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR3
Entrez Gene ID
10096
Gene Name
ARP3 actin related protein 3 homolog
Gene Aliases
ARP3
UniGene
Hs.433512
Gene Chromosome Location
Chr.2: 113889934-113961554 on Build GRCh38
Transcripts
NM_001277140.1 NM_005721.4 NR_102318.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
714677 c.411A>T p.P137P Chr.2: 113931375-113931375 on Build GRCh38
3040445 c.375T>C p.Y125Y Chr.2: 113931339-113931339 on Build GRCh38
6593105 c.377C>G p.T126S Chr.2: 113931341-113931341 on Build GRCh38
3379739 c.391T>G p.F131V Chr.2: 113931355-113931355 on Build GRCh38
399067 c.376A>C p.T126P Chr.2: 113931340-113931340 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047320092 Chr.2: 113931606-113931606 on Build GRCh38
rs775821415 Chr.2: 113931389-113931389 on Build GRCh38
rs931595494 Chr.2: 113931243-113931243 on Build GRCh38
rs769172240 Chr.2: 113931411-113931411 on Build GRCh38
rs780578497 Chr.2: 113931616-113931616 on Build GRCh38
rs779061992 Chr.2: 113931417-113931422 on Build GRCh38
rs970579438 Chr.2: 113931639-113931639 on Build GRCh38
rs750511269 Chr.2: 113931430-113931431 on Build GRCh38
rs968344146 Chr.2: 113931466-113931466 on Build GRCh38
rs748742756 Chr.2: 113931534-113931534 on Build GRCh38
rs758482294 Chr.2: 113931444-113931445 on Build GRCh38
rs938872981 Chr.2: 113931621-113931621 on Build GRCh38
rs532079265 Chr.2: 113931467-113931467 on Build GRCh38
rs79567640 Chr.2: 113931484-113931484 on Build GRCh38
rs878943838 Chr.2: 113931471-113931471 on Build GRCh38
rs1031217461 Chr.2: 113931489-113931489 on Build GRCh38
rs931518868 Chr.2: 113931591-113931591 on Build GRCh38
rs756054890 Chr.2: 113931440-113931440 on Build GRCh38
rs781669596 Chr.2: 113931237-113931237 on Build GRCh38
rs975791444 Chr.2: 113931557-113931557 on Build GRCh38
rs879026605 Chr.2: 113931310-113931310 on Build GRCh38
rs751421952 Chr.2: 113931270-113931270 on Build GRCh38
rs1056210702 Chr.2: 113931635-113931635 on Build GRCh38
rs1000791350 Chr.2: 113931488-113931488 on Build GRCh38
rs528273687 Chr.2: 113931654-113931654 on Build GRCh38
rs200938275 Chr.2: 113931470-113931471 on Build GRCh38
rs766029374 Chr.2: 113931263-113931263 on Build GRCh38
rs746240721 Chr.2: 113931385-113931385 on Build GRCh38
rs898771573 Chr.2: 113931226-113931226 on Build GRCh38
rs973533823 Chr.2: 113931286-113931286 on Build GRCh38
rs374819129 Chr.2: 113931387-113931387 on Build GRCh38
rs777756922 Chr.2: 113931323-113931323 on Build GRCh38
rs749365317 Chr.2: 113931345-113931345 on Build GRCh38
rs771081642 Chr.2: 113931360-113931360 on Build GRCh38
rs559323859 Chr.2: 113931576-113931576 on Build GRCh38
rs374572880 Chr.2: 113931418-113931418 on Build GRCh38
rs751892514 Chr.2: 113931212-113931212 on Build GRCh38
rs185321834 Chr.2: 113931256-113931256 on Build GRCh38
rs762549449 Chr.2: 113931262-113931262 on Build GRCh38
rs551851710 Chr.2: 113931490-113931490 on Build GRCh38
rs75286930 Chr.2: 113931483-113931483 on Build GRCh38
rs908704095 Chr.2: 113931618-113931618 on Build GRCh38
rs772642332 Chr.2: 113931413-113931413 on Build GRCh38
rs1038126858 Chr.2: 113931348-113931348 on Build GRCh38
rs957580464 Chr.2: 113931572-113931572 on Build GRCh38
rs964001436 Chr.2: 113931556-113931556 on Build GRCh38
rs141317503 Chr.2: 113931415-113931415 on Build GRCh38
rs571387045 Chr.2: 113931330-113931330 on Build GRCh38
rs912669418 Chr.2: 113931539-113931539 on Build GRCh38
rs756117571 Chr.2: 113931303-113931303 on Build GRCh38
rs572600321 Chr.2: 113931223-113931223 on Build GRCh38
rs752621921 Chr.2: 113931298-113931298 on Build GRCh38
rs761234265 Chr.2: 113931409-113931409 on Build GRCh38
rs781277293 Chr.2: 113931289-113931289 on Build GRCh38
rs34090058 Chr.2: 113931368-113931368 on Build GRCh38
rs754780143 Chr.2: 113931288-113931288 on Build GRCh38
rs561613185 Chr.2: 113931264-113931264 on Build GRCh38
rs770488395 Chr.2: 113931542-113931542 on Build GRCh38
rs112631500 Chr.2: 113931432-113931432 on Build GRCh38
rs201774265 Chr.2: 113931439-113931439 on Build GRCh38
rs755471873 Chr.2: 113931236-113931236 on Build GRCh38

View Full Product Details