Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 117915792-117916317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC93
Entrez Gene ID
54520
Gene Name
coiled-coil domain containing 93
Gene Aliases
-
UniGene
Hs.107845
Gene Chromosome Location
Chr.2: 117915478-118014163 on Build GRCh38
Transcripts
NM_019044.4 XM_011511359.1 XM_011511361.1 XM_011511362.1

SNP Detail

SNP ID Chromosome Location
rs567321866 Chr.2: 117915971-117915971 on Build GRCh38
rs937385001 Chr.2: 117916065-117916065 on Build GRCh38
rs772931976 Chr.2: 117916188-117916188 on Build GRCh38
rs1016906968 Chr.2: 117915868-117915868 on Build GRCh38
rs1025806511 Chr.2: 117915930-117915930 on Build GRCh38
rs912847477 Chr.2: 117915954-117915954 on Build GRCh38
rs1032432208 Chr.2: 117915822-117915822 on Build GRCh38
rs532094421 Chr.2: 117916112-117916112 on Build GRCh38
rs373330389 Chr.2: 117916051-117916051 on Build GRCh38
rs965619255 Chr.2: 117916199-117916199 on Build GRCh38
rs540473964 Chr.2: 117915995-117915995 on Build GRCh38
rs902310583 Chr.2: 117916132-117916132 on Build GRCh38
rs1047290818 Chr.2: 117916168-117916168 on Build GRCh38
rs960854992 Chr.2: 117915941-117915941 on Build GRCh38
rs566455420 Chr.2: 117916156-117916156 on Build GRCh38
rs1019286113 Chr.2: 117916192-117916192 on Build GRCh38
rs528538990 Chr.2: 117916154-117916154 on Build GRCh38
rs952889362 Chr.2: 117916238-117916238 on Build GRCh38
rs749502150 Chr.2: 117916026-117916026 on Build GRCh38
rs937416643 Chr.2: 117916286-117916286 on Build GRCh38
rs965610907 Chr.2: 117915957-117915957 on Build GRCh38
rs942765930 Chr.2: 117916148-117916148 on Build GRCh38
rs1046330 Chr.2: 117916087-117916087 on Build GRCh38
rs994390065 Chr.2: 117915889-117915889 on Build GRCh38
rs898750287 Chr.2: 117915872-117915872 on Build GRCh38
rs969142125 Chr.2: 117915998-117915998 on Build GRCh38
rs896442085 Chr.2: 117916094-117916094 on Build GRCh38
rs1006594239 Chr.2: 117916166-117916166 on Build GRCh38
rs923127574 Chr.2: 117916111-117916111 on Build GRCh38
rs985510640 Chr.2: 117915930-117915930 on Build GRCh38
rs144623830 Chr.2: 117915836-117915837 on Build GRCh38
rs1023732346 Chr.2: 117916273-117916273 on Build GRCh38
rs143681604 Chr.2: 117915842-117915843 on Build GRCh38
rs931394681 Chr.2: 117916010-117916010 on Build GRCh38
rs972933096 Chr.2: 117915965-117915965 on Build GRCh38
rs542808591 Chr.2: 117916128-117916128 on Build GRCh38
rs534333662 Chr.2: 117916002-117916002 on Build GRCh38
rs552889563 Chr.2: 117916003-117916003 on Build GRCh38
rs186935876 Chr.2: 117916004-117916004 on Build GRCh38
rs575667925 Chr.2: 117916262-117916262 on Build GRCh38
rs769621532 Chr.2: 117916178-117916178 on Build GRCh38
rs536252038 Chr.2: 117916028-117916028 on Build GRCh38
rs555065723 Chr.2: 117915835-117915835 on Build GRCh38
rs1013453533 Chr.2: 117916120-117916120 on Build GRCh38
rs1046328 Chr.2: 117916162-117916162 on Build GRCh38
rs539239914 Chr.2: 117916137-117916137 on Build GRCh38
rs756464355 Chr.2: 117915910-117915910 on Build GRCh38
rs191406907 Chr.2: 117916164-117916164 on Build GRCh38
rs927431403 Chr.2: 117916283-117916283 on Build GRCh38
rs918761783 Chr.2: 117916233-117916233 on Build GRCh38
rs536778580 Chr.2: 117915823-117915823 on Build GRCh38
rs75855108 Chr.2: 117916032-117916032 on Build GRCh38
rs896682388 Chr.2: 117916203-117916203 on Build GRCh38
rs991899169 Chr.2: 117915968-117915968 on Build GRCh38
rs984678612 Chr.2: 117916282-117916282 on Build GRCh38
rs972944430 Chr.2: 117916222-117916222 on Build GRCh38
rs953712979 Chr.2: 117915847-117915847 on Build GRCh38
rs1038778539 Chr.2: 117916154-117916156 on Build GRCh38
rs776716829 Chr.2: 117916146-117916146 on Build GRCh38

View Full Product Details