Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 247960-248462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHL1
Entrez Gene ID
10752
Gene Name
cell adhesion molecule L1 like
Gene Aliases
CALL, L1CAM2
UniGene
Hs.148909
Gene Chromosome Location
Chr.3: 196588-409417 on Build GRCh38
Transcripts
NM_001253387.1 NM_006614.3 NR_045572.1 XM_006712938.1 XM_006712939.3 XM_006712940.3 XM_011533292.1 XM_011533294.1 XM_011533295.1 XM_011533296.1 XM_017005566.1 XM_017005567.1 XM_017005569.1 XM_017005570.1 XM_017005571.1 XM_017005572.1 XM_017005573.1

SNP Detail

SNP ID Chromosome Location
rs1041402402 Chr.3: 248106-248106 on Build GRCh38
rs375933522 Chr.3: 248107-248107 on Build GRCh38
rs537770004 Chr.3: 248153-248153 on Build GRCh38
rs370352202 Chr.3: 248376-248376 on Build GRCh38
rs778677466 Chr.3: 248128-248128 on Build GRCh38
rs745843936 Chr.3: 248150-248150 on Build GRCh38
rs568208303 Chr.3: 248023-248023 on Build GRCh38
rs984953838 Chr.3: 248420-248420 on Build GRCh38
rs756795821 Chr.3: 248027-248027 on Build GRCh38
rs1045723800 Chr.3: 248234-248234 on Build GRCh38
rs769829925 Chr.3: 248273-248273 on Build GRCh38
rs902894138 Chr.3: 248113-248113 on Build GRCh38
rs926735613 Chr.3: 248224-248224 on Build GRCh38
rs558629922 Chr.3: 248164-248164 on Build GRCh38
rs113666146 Chr.3: 248175-248175 on Build GRCh38
rs953189178 Chr.3: 248130-248130 on Build GRCh38
rs73814060 Chr.3: 248121-248121 on Build GRCh38
rs1011630254 Chr.3: 248102-248102 on Build GRCh38
rs1052361305 Chr.3: 248267-248267 on Build GRCh38
rs945652328 Chr.3: 248278-248278 on Build GRCh38
rs1007268981 Chr.3: 248288-248288 on Build GRCh38
rs944924196 Chr.3: 248148-248148 on Build GRCh38
rs541075446 Chr.3: 248206-248206 on Build GRCh38
rs953162451 Chr.3: 248073-248073 on Build GRCh38
rs989761463 Chr.3: 248057-248057 on Build GRCh38
rs332476 Chr.3: 248417-248417 on Build GRCh38
rs938234565 Chr.3: 248083-248083 on Build GRCh38
rs956560913 Chr.3: 248053-248053 on Build GRCh38
rs1051492848 Chr.3: 248336-248336 on Build GRCh38
rs552001545 Chr.3: 248392-248392 on Build GRCh38
rs1007414717 Chr.3: 248135-248135 on Build GRCh38
rs1016115885 Chr.3: 248404-248404 on Build GRCh38
rs758110492 Chr.3: 248275-248275 on Build GRCh38
rs1018682075 Chr.3: 248367-248367 on Build GRCh38
rs894534222 Chr.3: 248219-248219 on Build GRCh38
rs73814061 Chr.3: 248279-248279 on Build GRCh38
rs576820608 Chr.3: 248194-248194 on Build GRCh38
rs902841952 Chr.3: 248284-248284 on Build GRCh38
rs938165312 Chr.3: 248266-248266 on Build GRCh38
rs1016580872 Chr.3: 248137-248137 on Build GRCh38
rs999296403 Chr.3: 248259-248259 on Build GRCh38
rs1032083259 Chr.3: 248096-248096 on Build GRCh38
rs1031219082 Chr.3: 248026-248026 on Build GRCh38
rs374009341 Chr.3: 248002-248002 on Build GRCh38
rs1022029955 Chr.3: 248070-248070 on Build GRCh38
rs1010728950 Chr.3: 248277-248277 on Build GRCh38
rs36117231 Chr.3: 248205-248206 on Build GRCh38
rs1054430893 Chr.3: 248094-248094 on Build GRCh38
rs533064259 Chr.3: 248255-248255 on Build GRCh38
rs570216290 Chr.3: 248139-248139 on Build GRCh38
rs972163043 Chr.3: 248387-248387 on Build GRCh38
rs73814062 Chr.3: 248394-248394 on Build GRCh38
rs965679648 Chr.3: 248384-248384 on Build GRCh38
rs972450111 Chr.3: 248143-248143 on Build GRCh38
rs949488585 Chr.3: 248019-248019 on Build GRCh38
rs894086786 Chr.3: 248101-248101 on Build GRCh38
rs933012034 Chr.3: 248285-248285 on Build GRCh38
rs1049723656 Chr.3: 248203-248203 on Build GRCh38
rs907212236 Chr.3: 248235-248235 on Build GRCh38
rs1053971118 Chr.3: 248272-248272 on Build GRCh38
rs948801565 Chr.3: 248231-248231 on Build GRCh38

View Full Product Details