Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 4516384-4516885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPR1
Entrez Gene ID
3708
Gene Name
inositol 1,4,5-trisphosphate receptor type 1
Gene Aliases
ACV, CLA4, INSP3R1, IP3R, IP3R1, PPP1R94, SCA15, SCA16, SCA29
UniGene
Hs.567295
Gene Chromosome Location
Chr.3: 4493348-4847840 on Build GRCh38
Transcripts
NM_001099952.2 NM_001168272.1 NM_002222.5 XM_005265109.2 XM_005265110.2 XM_006713131.2 XM_011533681.1 XM_011533682.2 XM_011533683.2 XM_011533684.1 XM_011533685.1 XM_011533686.1 XM_011533687.1 XM_011533688.1 XM_011533690.1 XM_011533691.1 XM_011533692.2 XM_017006357.1 XM_017006358.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1045163 c.78T>G p.F26L Chr.3: 4516569-4516569 on Build GRCh38
3067655 c.66G>A p.S22S Chr.3: 4516557-4516557 on Build GRCh38
5588078 c.65C>T p.S22L Chr.3: 4516556-4516556 on Build GRCh38
3067656 c.66G>A p.S22S Chr.3: 4516557-4516557 on Build GRCh38
5588077 c.65C>T p.S22L Chr.3: 4516556-4516556 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922741219 Chr.3: 4516737-4516737 on Build GRCh38
rs1050355334 Chr.3: 4516648-4516649 on Build GRCh38
rs951742390 Chr.3: 4516547-4516547 on Build GRCh38
rs1011539694 Chr.3: 4516663-4516663 on Build GRCh38
rs755384756 Chr.3: 4516542-4516542 on Build GRCh38
rs923132519 Chr.3: 4516844-4516844 on Build GRCh38
rs369723935 Chr.3: 4516490-4516490 on Build GRCh38
rs9829878 Chr.3: 4516420-4516420 on Build GRCh38
rs763876605 Chr.3: 4516460-4516460 on Build GRCh38
rs761720611 Chr.3: 4516618-4516618 on Build GRCh38
rs957298813 Chr.3: 4516627-4516627 on Build GRCh38
rs757223877 Chr.3: 4516750-4516750 on Build GRCh38
rs778551906 Chr.3: 4516555-4516555 on Build GRCh38
rs112944532 Chr.3: 4516557-4516557 on Build GRCh38
rs570325460 Chr.3: 4516671-4516671 on Build GRCh38
rs76009854 Chr.3: 4516755-4516755 on Build GRCh38
rs984443400 Chr.3: 4516760-4516760 on Build GRCh38
rs1014423440 Chr.3: 4516524-4516524 on Build GRCh38
rs377352614 Chr.3: 4516613-4516613 on Build GRCh38
rs1027198310 Chr.3: 4516748-4516748 on Build GRCh38
rs893168265 Chr.3: 4516659-4516659 on Build GRCh38
rs909854175 Chr.3: 4516777-4516777 on Build GRCh38
rs775218272 Chr.3: 4516591-4516591 on Build GRCh38
rs931912416 Chr.3: 4516634-4516634 on Build GRCh38
rs756249640 Chr.3: 4516483-4516483 on Build GRCh38
rs985976883 Chr.3: 4516670-4516670 on Build GRCh38
rs991683914 Chr.3: 4516530-4516530 on Build GRCh38
rs373630095 Chr.3: 4516545-4516545 on Build GRCh38
rs200534989 Chr.3: 4516548-4516548 on Build GRCh38
rs780392647 Chr.3: 4516506-4516506 on Build GRCh38
rs34564211 Chr.3: 4516762-4516763 on Build GRCh38
rs1035844807 Chr.3: 4516700-4516700 on Build GRCh38
rs910380777 Chr.3: 4516676-4516676 on Build GRCh38
rs761422967 Chr.3: 4516684-4516684 on Build GRCh38
rs772632331 Chr.3: 4516458-4516458 on Build GRCh38
rs753790681 Chr.3: 4516462-4516462 on Build GRCh38
rs73807272 Chr.3: 4516612-4516612 on Build GRCh38
rs550392909 Chr.3: 4516664-4516664 on Build GRCh38
rs559949135 Chr.3: 4516437-4516437 on Build GRCh38
rs765219630 Chr.3: 4516619-4516619 on Build GRCh38
rs770468426 Chr.3: 4516809-4516810 on Build GRCh38
rs760334522 Chr.3: 4516592-4516592 on Build GRCh38
rs761845456 Chr.3: 4516468-4516468 on Build GRCh38
rs961812957 Chr.3: 4516791-4516791 on Build GRCh38
rs760519532 Chr.3: 4516456-4516456 on Build GRCh38
rs769176795 Chr.3: 4516833-4516833 on Build GRCh38
rs930029036 Chr.3: 4516743-4516743 on Build GRCh38
rs751806213 Chr.3: 4516520-4516520 on Build GRCh38
rs148305093 Chr.3: 4516699-4516699 on Build GRCh38
rs907583200 Chr.3: 4516550-4516550 on Build GRCh38
rs528083247 Chr.3: 4516472-4516472 on Build GRCh38
rs770471562 Chr.3: 4516551-4516551 on Build GRCh38
rs566721357 Chr.3: 4516856-4516856 on Build GRCh38
rs958851519 Chr.3: 4516709-4516709 on Build GRCh38
rs775141055 Chr.3: 4516452-4516452 on Build GRCh38
rs866671942 Chr.3: 4516543-4516547 on Build GRCh38

View Full Product Details