Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 4627616-4628100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPR1
Entrez Gene ID
3708
Gene Name
inositol 1,4,5-trisphosphate receptor type 1
Gene Aliases
ACV, CLA4, INSP3R1, IP3R, IP3R1, PPP1R94, SCA15, SCA16, SCA29
UniGene
Hs.567295
Gene Chromosome Location
Chr.3: 4493348-4847840 on Build GRCh38
Transcripts
NM_001099952.2 NM_001168272.1 NM_002222.5 XM_005265109.2 XM_005265110.2 XM_006713131.2 XM_011533681.1 XM_011533682.2 XM_011533683.2 XM_011533684.1 XM_011533685.1 XM_011533686.1 XM_011533687.1 XM_011533688.1 XM_011533690.1 XM_011533691.1 XM_011533692.2 XM_017006357.1 XM_017006358.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3408691 c.193C>T p.R65C Chr.3: 4627792-4627792 on Build GRCh38
4977233 c.203C>T p.A68V Chr.3: 4627802-4627802 on Build GRCh38
1137037 c.238G>A p.A80T Chr.3: 4627837-4627837 on Build GRCh38
3068271 c.256G>A p.A86T Chr.3: 4627855-4627855 on Build GRCh38
3940534 c.261G>A p.V87V Chr.3: 4627860-4627860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs933070642 Chr.3: 4627989-4627989 on Build GRCh38
rs764644504 Chr.3: 4627730-4627730 on Build GRCh38
rs765734360 Chr.3: 4627898-4627898 on Build GRCh38
rs762648966 Chr.3: 4627901-4627901 on Build GRCh38
rs551498897 Chr.3: 4628068-4628068 on Build GRCh38
rs570124103 Chr.3: 4628076-4628078 on Build GRCh38
rs558784142 Chr.3: 4627824-4627824 on Build GRCh38
rs1031725122 Chr.3: 4627760-4627760 on Build GRCh38
rs1021573166 Chr.3: 4627692-4627692 on Build GRCh38
rs757773705 Chr.3: 4627894-4627894 on Build GRCh38
rs1004272877 Chr.3: 4627679-4627679 on Build GRCh38
rs371299264 Chr.3: 4627878-4627878 on Build GRCh38
rs183746937 Chr.3: 4627696-4627696 on Build GRCh38
rs374995790 Chr.3: 4627712-4627712 on Build GRCh38
rs536347927 Chr.3: 4627652-4627652 on Build GRCh38
rs770832119 Chr.3: 4627844-4627844 on Build GRCh38
rs532018150 Chr.3: 4628016-4628016 on Build GRCh38
rs943070510 Chr.3: 4627906-4627906 on Build GRCh38
rs541174693 Chr.3: 4627976-4627976 on Build GRCh38
rs754396958 Chr.3: 4627741-4627741 on Build GRCh38
rs374495990 Chr.3: 4627835-4627835 on Build GRCh38
rs934774125 Chr.3: 4627864-4627864 on Build GRCh38
rs1030414860 Chr.3: 4627649-4627649 on Build GRCh38
rs1013360959 Chr.3: 4628067-4628067 on Build GRCh38
rs1051558030 Chr.3: 4627998-4627998 on Build GRCh38
rs754498803 Chr.3: 4627909-4627909 on Build GRCh38
rs901789046 Chr.3: 4627941-4627941 on Build GRCh38
rs1007884339 Chr.3: 4627668-4627668 on Build GRCh38
rs367814655 Chr.3: 4627854-4627854 on Build GRCh38
rs766252600 Chr.3: 4627902-4627902 on Build GRCh38
rs2307067 Chr.3: 4628008-4628008 on Build GRCh38
rs933253975 Chr.3: 4628004-4628004 on Build GRCh38
rs761076544 Chr.3: 4627866-4627866 on Build GRCh38
rs971660373 Chr.3: 4627841-4627841 on Build GRCh38
rs572262537 Chr.3: 4627953-4627953 on Build GRCh38
rs910154477 Chr.3: 4627944-4627944 on Build GRCh38
rs963042376 Chr.3: 4627701-4627701 on Build GRCh38
rs757740509 Chr.3: 4627754-4627754 on Build GRCh38
rs190299782 Chr.3: 4628007-4628007 on Build GRCh38
rs531991826 Chr.3: 4627740-4627740 on Build GRCh38
rs1037169857 Chr.3: 4627958-4627958 on Build GRCh38
rs774483090 Chr.3: 4627847-4627847 on Build GRCh38
rs888767459 Chr.3: 4627998-4627998 on Build GRCh38
rs563347395 Chr.3: 4627794-4627794 on Build GRCh38
rs746562377 Chr.3: 4627770-4627770 on Build GRCh38
rs1038679417 Chr.3: 4627907-4627907 on Build GRCh38
rs2307066 Chr.3: 4628033-4628033 on Build GRCh38
rs536328664 Chr.3: 4627893-4627893 on Build GRCh38
rs780777144 Chr.3: 4627921-4627921 on Build GRCh38
rs529031125 Chr.3: 4627843-4627843 on Build GRCh38
rs754207637 Chr.3: 4627889-4627889 on Build GRCh38
rs376974334 Chr.3: 4627777-4627777 on Build GRCh38
rs780830383 Chr.3: 4627775-4627775 on Build GRCh38
rs767787371 Chr.3: 4627855-4627855 on Build GRCh38
rs562670402 Chr.3: 4628032-4628032 on Build GRCh38
rs754623990 Chr.3: 4627773-4627773 on Build GRCh38
rs764519723 Chr.3: 4627882-4627882 on Build GRCh38
rs543538622 Chr.3: 4627779-4627779 on Build GRCh38
rs139104291 Chr.3: 4627701-4627702 on Build GRCh38
rs576089005 Chr.3: 4627698-4627698 on Build GRCh38
rs370953766 Chr.3: 4627787-4627787 on Build GRCh38
rs898561046 Chr.3: 4628048-4628048 on Build GRCh38
rs776148416 Chr.3: 4627666-4627666 on Build GRCh38
rs888829083 Chr.3: 4627665-4627665 on Build GRCh38
rs182590802 Chr.3: 4628028-4628028 on Build GRCh38
rs756558937 Chr.3: 4627717-4627717 on Build GRCh38
rs915889019 Chr.3: 4627785-4627785 on Build GRCh38
rs779598045 Chr.3: 4627765-4627765 on Build GRCh38
rs567321039 Chr.3: 4627651-4627651 on Build GRCh38
rs756522212 Chr.3: 4627746-4627746 on Build GRCh38

View Full Product Details