Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 5179329-5179835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL8B
Entrez Gene ID
55207
Gene Name
ADP ribosylation factor like GTPase 8B
Gene Aliases
ARL10C, Gie1
UniGene
Hs.250009
Gene Chromosome Location
Chr.3: 5122245-5180916 on Build GRCh38
Transcripts
NM_018184.2

SNP Detail

SNP ID Chromosome Location
rs759326584 Chr.3: 5179359-5179359 on Build GRCh38
rs553146456 Chr.3: 5179757-5179757 on Build GRCh38
rs948748861 Chr.3: 5179796-5179796 on Build GRCh38
rs117819224 Chr.3: 5179383-5179383 on Build GRCh38
rs775099928 Chr.3: 5179412-5179412 on Build GRCh38
rs961839623 Chr.3: 5179636-5179636 on Build GRCh38
rs191037023 Chr.3: 5179673-5179673 on Build GRCh38
rs1048480054 Chr.3: 5179597-5179597 on Build GRCh38
rs564019185 Chr.3: 5179704-5179704 on Build GRCh38
rs551622386 Chr.3: 5179610-5179610 on Build GRCh38
rs1030201581 Chr.3: 5179666-5179666 on Build GRCh38
rs182631176 Chr.3: 5179443-5179443 on Build GRCh38
rs186302873 Chr.3: 5179503-5179503 on Build GRCh38
rs986162378 Chr.3: 5179681-5179681 on Build GRCh38
rs767505448 Chr.3: 5179409-5179409 on Build GRCh38
rs968844630 Chr.3: 5179709-5179712 on Build GRCh38
rs763113793 Chr.3: 5179726-5179726 on Build GRCh38
rs914272999 Chr.3: 5179463-5179463 on Build GRCh38
rs760581365 Chr.3: 5179476-5179476 on Build GRCh38
rs34105324 Chr.3: 5179442-5179443 on Build GRCh38
rs572931584 Chr.3: 5179556-5179556 on Build GRCh38
rs1006069385 Chr.3: 5179616-5179616 on Build GRCh38
rs1014794734 Chr.3: 5179379-5179379 on Build GRCh38
rs1057414769 Chr.3: 5179773-5179773 on Build GRCh38
rs977157594 Chr.3: 5179712-5179712 on Build GRCh38
rs766777903 Chr.3: 5179454-5179454 on Build GRCh38
rs914346230 Chr.3: 5179689-5179689 on Build GRCh38
rs938830470 Chr.3: 5179750-5179750 on Build GRCh38
rs866574379 Chr.3: 5179736-5179736 on Build GRCh38
rs183020948 Chr.3: 5179740-5179740 on Build GRCh38
rs947583020 Chr.3: 5179464-5179464 on Build GRCh38
rs997411531 Chr.3: 5179645-5179646 on Build GRCh38
rs953142452 Chr.3: 5179434-5179434 on Build GRCh38
rs938946962 Chr.3: 5179544-5179544 on Build GRCh38
rs894521036 Chr.3: 5179549-5179549 on Build GRCh38
rs143418261 Chr.3: 5179763-5179763 on Build GRCh38
rs975732965 Chr.3: 5179406-5179406 on Build GRCh38
rs542488238 Chr.3: 5179526-5179526 on Build GRCh38
rs765528301 Chr.3: 5179547-5179547 on Build GRCh38
rs1014681997 Chr.3: 5179632-5179632 on Build GRCh38
rs1042255442 Chr.3: 5179505-5179505 on Build GRCh38
rs916129948 Chr.3: 5179779-5179779 on Build GRCh38
rs750726405 Chr.3: 5179569-5179569 on Build GRCh38
rs535475138 Chr.3: 5179772-5179772 on Build GRCh38
rs796912475 Chr.3: 5179660-5179660 on Build GRCh38

View Full Product Details