Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9740678-9741212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRPF1
Entrez Gene ID
7862
Gene Name
bromodomain and PHD finger containing 1
Gene Aliases
BR140
UniGene
Hs.1004
Gene Chromosome Location
Chr.3: 9731729-9748015 on Build GRCh38
Transcripts
NM_001003694.1 NM_001319049.1 NM_001319050.1 NM_004634.2 XM_005265449.1 XM_005265450.1 XM_005265452.1 XM_011534101.1 XM_011534102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
265774 c.1652G>A p.R551Q Chr.3: 9740871-9740871 on Build GRCh38
1049131 c.1675C>T p.R559C Chr.3: 9740894-9740894 on Build GRCh38
5995012 c.1672C>T p.R558C Chr.3: 9740891-9740891 on Build GRCh38
731348 c.1659G>T p.G553G Chr.3: 9740878-9740878 on Build GRCh38
5865407 c.1697C>T p.S566F Chr.3: 9740916-9740916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995709909 Chr.3: 9741188-9741188 on Build GRCh38
rs1024318345 Chr.3: 9740872-9740872 on Build GRCh38
rs202091926 Chr.3: 9740971-9740971 on Build GRCh38
rs762329210 Chr.3: 9740960-9740960 on Build GRCh38
rs770715314 Chr.3: 9740984-9740984 on Build GRCh38
rs149893819 Chr.3: 9740925-9740925 on Build GRCh38
rs558924805 Chr.3: 9741154-9741154 on Build GRCh38
rs772270535 Chr.3: 9741115-9741115 on Build GRCh38
rs767573342 Chr.3: 9740767-9740767 on Build GRCh38
rs748879941 Chr.3: 9740981-9740981 on Build GRCh38
rs757662316 Chr.3: 9740857-9740857 on Build GRCh38
rs772075231 Chr.3: 9740932-9740932 on Build GRCh38
rs760910675 Chr.3: 9740799-9740799 on Build GRCh38
rs748043133 Chr.3: 9741109-9741109 on Build GRCh38
rs141454094 Chr.3: 9740721-9740721 on Build GRCh38
rs182220710 Chr.3: 9741182-9741182 on Build GRCh38
rs373715117 Chr.3: 9740722-9740722 on Build GRCh38
rs894410502 Chr.3: 9741078-9741078 on Build GRCh38
rs957637248 Chr.3: 9741054-9741054 on Build GRCh38
rs750360933 Chr.3: 9740983-9740983 on Build GRCh38
rs370240712 Chr.3: 9740947-9740947 on Build GRCh38
rs971454177 Chr.3: 9740881-9740881 on Build GRCh38
rs138963007 Chr.3: 9740704-9740704 on Build GRCh38
rs780103175 Chr.3: 9740703-9740703 on Build GRCh38
rs985576180 Chr.3: 9741046-9741046 on Build GRCh38
rs981164557 Chr.3: 9741177-9741177 on Build GRCh38
rs368143990 Chr.3: 9740975-9740975 on Build GRCh38
rs905665268 Chr.3: 9740805-9740805 on Build GRCh38
rs1054751303 Chr.3: 9741060-9741060 on Build GRCh38
rs1045596834 Chr.3: 9740798-9740798 on Build GRCh38
rs762900143 Chr.3: 9740743-9740743 on Build GRCh38
rs372753144 Chr.3: 9740983-9740984 on Build GRCh38
rs907245522 Chr.3: 9741118-9741118 on Build GRCh38
rs759590481 Chr.3: 9740754-9740754 on Build GRCh38
rs754299765 Chr.3: 9740839-9740839 on Build GRCh38
rs898632345 Chr.3: 9741179-9741179 on Build GRCh38
rs779628014 Chr.3: 9740862-9740862 on Build GRCh38
rs750956735 Chr.3: 9740879-9740879 on Build GRCh38
rs533900598 Chr.3: 9741105-9741105 on Build GRCh38
rs777470249 Chr.3: 9740980-9740980 on Build GRCh38
rs750973515 Chr.3: 9740966-9740966 on Build GRCh38
rs1015709656 Chr.3: 9741171-9741171 on Build GRCh38
rs1029801425 Chr.3: 9741026-9741026 on Build GRCh38
rs369772859 Chr.3: 9741083-9741083 on Build GRCh38
rs944432140 Chr.3: 9741036-9741036 on Build GRCh38
rs766355261 Chr.3: 9740750-9740750 on Build GRCh38
rs572308400 Chr.3: 9740991-9740991 on Build GRCh38
rs759450597 Chr.3: 9740930-9740930 on Build GRCh38
rs774242990 Chr.3: 9740751-9740751 on Build GRCh38
rs769451679 Chr.3: 9741186-9741186 on Build GRCh38
rs754462866 Chr.3: 9740884-9740884 on Build GRCh38
rs147002530 Chr.3: 9740986-9740986 on Build GRCh38
rs754105866 Chr.3: 9740958-9740958 on Build GRCh38
rs765331570 Chr.3: 9740760-9740761 on Build GRCh38
rs949790461 Chr.3: 9740784-9740784 on Build GRCh38
rs774284168 Chr.3: 9740914-9740914 on Build GRCh38
rs913007507 Chr.3: 9740762-9740762 on Build GRCh38
rs764384206 Chr.3: 9740836-9740836 on Build GRCh38
rs201724083 Chr.3: 9740953-9740953 on Build GRCh38
rs142175258 Chr.3: 9740890-9740890 on Build GRCh38
rs961764693 Chr.3: 9740990-9740990 on Build GRCh38
rs998192273 Chr.3: 9741020-9741020 on Build GRCh38
rs200132847 Chr.3: 9740899-9740899 on Build GRCh38
rs780590390 Chr.3: 9740972-9740972 on Build GRCh38
rs138117634 Chr.3: 9741090-9741090 on Build GRCh38
rs926226906 Chr.3: 9741047-9741047 on Build GRCh38
rs755549839 Chr.3: 9740978-9740978 on Build GRCh38
rs1004249917 Chr.3: 9741170-9741170 on Build GRCh38
rs772951644 Chr.3: 9740738-9740738 on Build GRCh38
rs988923885 Chr.3: 9741064-9741064 on Build GRCh38
rs953003597 Chr.3: 9740979-9740979 on Build GRCh38
rs765641628 Chr.3: 9740961-9740961 on Build GRCh38
rs954190083 Chr.3: 9741040-9741040 on Build GRCh38
rs770709359 Chr.3: 9740911-9740911 on Build GRCh38
rs775590646 Chr.3: 9740946-9740946 on Build GRCh38
rs745814695 Chr.3: 9740988-9740988 on Build GRCh38
rs749123930 Chr.3: 9740906-9740906 on Build GRCh38
rs777382821 Chr.3: 9740896-9740896 on Build GRCh38
rs752865774 Chr.3: 9740771-9740771 on Build GRCh38
rs745581160 Chr.3: 9741068-9741068 on Build GRCh38
rs769439953 Chr.3: 9740891-9740891 on Build GRCh38

View Full Product Details