Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10028518-10029061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCD2
Entrez Gene ID
2177
Gene Name
Fanconi anemia complementation group D2
Gene Aliases
FA-D2, FA4, FACD, FAD, FAD2, FANCD
UniGene
Hs.208388
Gene Chromosome Location
Chr.3: 10026387-10101937 on Build GRCh38
Transcripts
NM_001018115.2 NM_001319984.1 NM_033084.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1308311 c.14G>C p.R5T Chr.3: 10028671-10028671 on Build GRCh38
7156838 c.55G>T p.D19Y Chr.3: 10028712-10028712 on Build GRCh38
7156837 c.55G>T p.D19Y Chr.3: 10028712-10028712 on Build GRCh38
1308312 c.14G>C p.R5T Chr.3: 10028671-10028671 on Build GRCh38
350685 c.42G>C p.E14D Chr.3: 10028699-10028699 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762544551 Chr.3: 10028764-10028764 on Build GRCh38
rs147426418 Chr.3: 10028690-10028690 on Build GRCh38
rs777719596 Chr.3: 10028722-10028723 on Build GRCh38
rs996281316 Chr.3: 10028912-10028912 on Build GRCh38
rs3732974 Chr.3: 10028652-10028652 on Build GRCh38
rs144926145 Chr.3: 10028735-10028735 on Build GRCh38
rs370843473 Chr.3: 10028646-10028646 on Build GRCh38
rs989100220 Chr.3: 10028787-10028787 on Build GRCh38
rs55892753 Chr.3: 10028913-10028913 on Build GRCh38
rs150075366 Chr.3: 10028685-10028685 on Build GRCh38
rs749402179 Chr.3: 10028702-10028702 on Build GRCh38
rs751263095 Chr.3: 10028770-10028770 on Build GRCh38
rs745937059 Chr.3: 10029025-10029025 on Build GRCh38
rs954129525 Chr.3: 10028742-10028742 on Build GRCh38
rs761151603 Chr.3: 10028734-10028734 on Build GRCh38
rs756374537 Chr.3: 10028885-10028885 on Build GRCh38
rs963475972 Chr.3: 10028555-10028555 on Build GRCh38
rs967641358 Chr.3: 10029000-10029000 on Build GRCh38
rs765845616 Chr.3: 10028767-10028767 on Build GRCh38
rs56098809 Chr.3: 10028886-10028886 on Build GRCh38
rs113338938 Chr.3: 10028612-10028612 on Build GRCh38
rs545556102 Chr.3: 10029034-10029034 on Build GRCh38
rs949190517 Chr.3: 10028837-10028837 on Build GRCh38
rs371700840 Chr.3: 10028773-10028773 on Build GRCh38
rs776943888 Chr.3: 10028758-10028758 on Build GRCh38
rs538558340 Chr.3: 10028942-10028942 on Build GRCh38
rs936977732 Chr.3: 10028595-10028595 on Build GRCh38
rs774743051 Chr.3: 10028721-10028721 on Build GRCh38
rs913387411 Chr.3: 10028843-10028843 on Build GRCh38
rs899134053 Chr.3: 10028888-10028888 on Build GRCh38
rs887492533 Chr.3: 10028895-10028895 on Build GRCh38
rs781445211 Chr.3: 10029007-10029007 on Build GRCh38
rs771088798 Chr.3: 10028707-10028707 on Build GRCh38
rs901774987 Chr.3: 10029010-10029010 on Build GRCh38
rs954835992 Chr.3: 10028648-10028648 on Build GRCh38
rs190220233 Chr.3: 10028540-10028540 on Build GRCh38
rs754809720 Chr.3: 10028682-10028682 on Build GRCh38
rs925051964 Chr.3: 10028705-10028705 on Build GRCh38
rs1029178487 Chr.3: 10028647-10028647 on Build GRCh38
rs9833228 Chr.3: 10028733-10028733 on Build GRCh38
rs887658416 Chr.3: 10028628-10028628 on Build GRCh38
rs751175161 Chr.3: 10028676-10028676 on Build GRCh38
rs777509355 Chr.3: 10028700-10028700 on Build GRCh38
rs950189890 Chr.3: 10028855-10028855 on Build GRCh38
rs56176124 Chr.3: 10028868-10028868 on Build GRCh38
rs985829609 Chr.3: 10028761-10028761 on Build GRCh38
rs1004838289 Chr.3: 10028637-10028637 on Build GRCh38
rs528736110 Chr.3: 10028589-10028589 on Build GRCh38
rs754592042 Chr.3: 10028611-10028611 on Build GRCh38
rs890662104 Chr.3: 10028925-10028925 on Build GRCh38
rs754613426 Chr.3: 10028772-10028772 on Build GRCh38
rs981593040 Chr.3: 10028865-10028865 on Build GRCh38
rs997328477 Chr.3: 10029024-10029024 on Build GRCh38
rs536056640 Chr.3: 10028856-10028856 on Build GRCh38
rs1032024224 Chr.3: 10029018-10029018 on Build GRCh38
rs746053143 Chr.3: 10028722-10028722 on Build GRCh38
rs56123930 Chr.3: 10028869-10028869 on Build GRCh38
rs148612270 Chr.3: 10028774-10028774 on Build GRCh38
rs181838959 Chr.3: 10028655-10028655 on Build GRCh38
rs957660465 Chr.3: 10029019-10029019 on Build GRCh38
rs756078235 Chr.3: 10028695-10028695 on Build GRCh38
rs764665698 Chr.3: 10028740-10028740 on Build GRCh38
rs189329374 Chr.3: 10028982-10028982 on Build GRCh38
rs1020979186 Chr.3: 10028989-10028989 on Build GRCh38
rs4019853 Chr.3: 10028587-10028587 on Build GRCh38
rs772610095 Chr.3: 10028732-10028732 on Build GRCh38

View Full Product Details