Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 197749602-197750106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FYTTD1
Entrez Gene ID
84248
Gene Name
forty-two-three domain containing 1
Gene Aliases
UIF
UniGene
Hs.277533
Gene Chromosome Location
Chr.3: 197749553-197784446 on Build GRCh38
Transcripts
NM_001011537.2 NM_032288.6 NM_001145642.3 XM_006713829.3 XM_006713831.3 XM_017007545.1

Gene Symbol
RUBCN
Entrez Gene ID
9711
Gene Name
RUN and cysteine rich domain containing beclin 1 interacting protein
Gene Aliases
KIAA0226, RUBICON, SCAR15
UniGene
Hs.478868
Gene Chromosome Location
Chr.3: 197670213-197749851 on Build GRCh38
Transcripts
NM_001011537.2 NM_032288.6 NM_001145642.3 XM_006713829.3 XM_006713831.3 XM_017007545.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
311324 c.60G>T p.K20N Chr.3: 197750031-197750031 on Build GRCh38
4685943 c.36G>A p.T12T Chr.3: 197750007-197750007 on Build GRCh38
6668091 c.20G>A p.R7Q Chr.3: 197749991-197749991 on Build GRCh38
7125732 c.44C>A p.S15Y Chr.3: 197750015-197750015 on Build GRCh38
6431435 c.7C>T p.R3W Chr.3: 197749978-197749978 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199936115 Chr.3: 197750027-197750027 on Build GRCh38
rs983803886 Chr.3: 197749858-197749858 on Build GRCh38
rs1032866638 Chr.3: 197749907-197749907 on Build GRCh38
rs756715292 Chr.3: 197749697-197749697 on Build GRCh38
rs1030420456 Chr.3: 197749705-197749705 on Build GRCh38
rs768955266 Chr.3: 197749989-197749989 on Build GRCh38
rs909631656 Chr.3: 197749985-197749985 on Build GRCh38
rs779902502 Chr.3: 197749674-197749674 on Build GRCh38
rs985730736 Chr.3: 197749915-197749915 on Build GRCh38
rs144778477 Chr.3: 197749724-197749724 on Build GRCh38
rs901042475 Chr.3: 197749670-197749670 on Build GRCh38
rs769842762 Chr.3: 197750012-197750012 on Build GRCh38
rs535995443 Chr.3: 197749704-197749704 on Build GRCh38
rs748005833 Chr.3: 197750009-197750009 on Build GRCh38
rs769643926 Chr.3: 197750084-197750084 on Build GRCh38
rs750541611 Chr.3: 197750063-197750064 on Build GRCh38
rs140342002 Chr.3: 197749970-197749970 on Build GRCh38
rs747953023 Chr.3: 197750079-197750079 on Build GRCh38
rs1037336189 Chr.3: 197749635-197749635 on Build GRCh38
rs780198355 Chr.3: 197749978-197749978 on Build GRCh38
rs758930561 Chr.3: 197750042-197750042 on Build GRCh38
rs751106503 Chr.3: 197749951-197749951 on Build GRCh38
rs1032187346 Chr.3: 197749817-197749817 on Build GRCh38
rs956200210 Chr.3: 197749818-197749819 on Build GRCh38
rs944365988 Chr.3: 197749932-197749932 on Build GRCh38
rs747153175 Chr.3: 197749981-197749981 on Build GRCh38
rs914776982 Chr.3: 197749859-197749859 on Build GRCh38
rs755134168 Chr.3: 197750066-197750066 on Build GRCh38
rs541967493 Chr.3: 197750086-197750086 on Build GRCh38
rs778922334 Chr.3: 197749930-197749930 on Build GRCh38
rs779441651 Chr.3: 197749966-197749966 on Build GRCh38
rs887695183 Chr.3: 197749747-197749747 on Build GRCh38
rs915671552 Chr.3: 197749806-197749806 on Build GRCh38
rs933850495 Chr.3: 197749898-197749898 on Build GRCh38
rs764316335 Chr.3: 197749956-197749956 on Build GRCh38
rs757725379 Chr.3: 197749963-197749963 on Build GRCh38
rs948420045 Chr.3: 197749826-197749826 on Build GRCh38
rs866742144 Chr.3: 197749979-197749979 on Build GRCh38
rs765594054 Chr.3: 197750032-197750032 on Build GRCh38
rs546111062 Chr.3: 197749934-197749934 on Build GRCh38
rs376413160 Chr.3: 197749947-197749947 on Build GRCh38
rs776510184 Chr.3: 197749940-197749940 on Build GRCh38
rs778201963 Chr.3: 197749924-197749924 on Build GRCh38
rs955842361 Chr.3: 197749914-197749914 on Build GRCh38
rs909571151 Chr.3: 197749862-197749862 on Build GRCh38
rs1049999355 Chr.3: 197749901-197749901 on Build GRCh38
rs749698800 Chr.3: 197749925-197749925 on Build GRCh38
rs781536770 Chr.3: 197750071-197750071 on Build GRCh38
rs374077120 Chr.3: 197749942-197749942 on Build GRCh38
rs944418887 Chr.3: 197749666-197749666 on Build GRCh38
rs572117288 Chr.3: 197749920-197749920 on Build GRCh38
rs933877638 Chr.3: 197750069-197750069 on Build GRCh38
rs1036992498 Chr.3: 197749882-197749882 on Build GRCh38
rs750874230 Chr.3: 197750033-197750033 on Build GRCh38
rs112679200 Chr.3: 197749829-197749829 on Build GRCh38
rs796592343 Chr.3: 197749776-197749776 on Build GRCh38
rs897627125 Chr.3: 197749720-197749720 on Build GRCh38
rs772826694 Chr.3: 197749945-197749945 on Build GRCh38
rs779027689 Chr.3: 197749950-197749962 on Build GRCh38
rs754257971 Chr.3: 197749958-197749958 on Build GRCh38
rs553803476 Chr.3: 197749843-197749843 on Build GRCh38
rs28373227 Chr.3: 197749811-197749811 on Build GRCh38
rs773932677 Chr.3: 197750022-197750022 on Build GRCh38
rs576134538 Chr.3: 197749990-197749990 on Build GRCh38
rs977731716 Chr.3: 197749763-197749763 on Build GRCh38
rs775759906 Chr.3: 197749935-197749935 on Build GRCh38
rs543620468 Chr.3: 197750019-197750019 on Build GRCh38
rs756502645 Chr.3: 197749921-197749921 on Build GRCh38
rs773349004 Chr.3: 197750015-197750015 on Build GRCh38
rs907184919 Chr.3: 197749961-197749961 on Build GRCh38
rs765307103 Chr.3: 197749943-197749943 on Build GRCh38
rs901047881 Chr.3: 197749890-197749890 on Build GRCh38
rs768281186 Chr.3: 197749938-197749938 on Build GRCh38
rs1000046794 Chr.3: 197749714-197749714 on Build GRCh38
rs968673548 Chr.3: 197749734-197749734 on Build GRCh38
rs373199162 Chr.3: 197750028-197750028 on Build GRCh38
rs1022910648 Chr.3: 197749787-197749787 on Build GRCh38
rs889218642 Chr.3: 197749913-197749913 on Build GRCh38
rs1021557697 Chr.3: 197749729-197749729 on Build GRCh38
rs993366521 Chr.3: 197749799-197749799 on Build GRCh38
rs374640680 Chr.3: 197749774-197749774 on Build GRCh38
rs960273830 Chr.3: 197749781-197749781 on Build GRCh38
rs889166654 Chr.3: 197749716-197749716 on Build GRCh38
rs759068407 Chr.3: 197750023-197750023 on Build GRCh38
rs1012822957 Chr.3: 197749748-197749748 on Build GRCh38
rs368846031 Chr.3: 197749926-197749926 on Build GRCh38
rs990720034 Chr.3: 197749824-197749824 on Build GRCh38
rs370833584 Chr.3: 197749953-197749953 on Build GRCh38
rs1011690349 Chr.3: 197750031-197750031 on Build GRCh38
rs747154398 Chr.3: 197750052-197750052 on Build GRCh38
rs939652306 Chr.3: 197749874-197749874 on Build GRCh38
rs530137433 Chr.3: 197750044-197750044 on Build GRCh38

View Full Product Details