Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 197750557-197751093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FYTTD1
Entrez Gene ID
84248
Gene Name
forty-two-three domain containing 1
Gene Aliases
UIF
UniGene
Hs.277533
Gene Chromosome Location
Chr.3: 197749553-197784446 on Build GRCh38
Transcripts
NM_001011537.2 NM_032288.6 NR_027840.1

SNP Detail

SNP ID Chromosome Location
rs978648801 Chr.3: 197751024-197751024 on Build GRCh38
rs1010432366 Chr.3: 197750995-197750995 on Build GRCh38
rs557734199 Chr.3: 197750623-197750623 on Build GRCh38
rs1006524853 Chr.3: 197750720-197750720 on Build GRCh38
rs368287719 Chr.3: 197750716-197750716 on Build GRCh38
rs941620181 Chr.3: 197750653-197750653 on Build GRCh38
rs905502355 Chr.3: 197750914-197750914 on Build GRCh38
rs184533961 Chr.3: 197750745-197750745 on Build GRCh38
rs1017963519 Chr.3: 197750968-197750968 on Build GRCh38
rs1015924400 Chr.3: 197750599-197750599 on Build GRCh38
rs1006619244 Chr.3: 197750912-197750912 on Build GRCh38
rs1038604478 Chr.3: 197750662-197750662 on Build GRCh38
rs532033279 Chr.3: 197750843-197750843 on Build GRCh38
rs577958179 Chr.3: 197750908-197750908 on Build GRCh38
rs897367225 Chr.3: 197750855-197750855 on Build GRCh38
rs913090343 Chr.3: 197750803-197750803 on Build GRCh38
rs994345675 Chr.3: 197750682-197750682 on Build GRCh38
rs112293928 Chr.3: 197750881-197750881 on Build GRCh38
rs9844344 Chr.3: 197750578-197750578 on Build GRCh38
rs576296208 Chr.3: 197750624-197750624 on Build GRCh38
rs1043071661 Chr.3: 197750685-197750685 on Build GRCh38
rs889822077 Chr.3: 197750689-197750689 on Build GRCh38
rs956784979 Chr.3: 197750774-197750774 on Build GRCh38
rs910087882 Chr.3: 197750836-197750836 on Build GRCh38
rs547493870 Chr.3: 197750921-197750921 on Build GRCh38
rs984424323 Chr.3: 197750597-197750597 on Build GRCh38
rs980166505 Chr.3: 197750797-197750797 on Build GRCh38
rs1005734639 Chr.3: 197750911-197750911 on Build GRCh38
rs913149630 Chr.3: 197750663-197750663 on Build GRCh38
rs572082032 Chr.3: 197750767-197750767 on Build GRCh38
rs927307488 Chr.3: 197750814-197750814 on Build GRCh38
rs1041156197 Chr.3: 197750732-197750732 on Build GRCh38
rs989334076 Chr.3: 197750581-197750581 on Build GRCh38
rs1019782618 Chr.3: 197750577-197750577 on Build GRCh38
rs1017872120 Chr.3: 197750739-197750739 on Build GRCh38
rs1030908002 Chr.3: 197750768-197750768 on Build GRCh38
rs1001208510 Chr.3: 197750929-197750929 on Build GRCh38
rs1029830490 Chr.3: 197750717-197750717 on Build GRCh38
rs938122148 Chr.3: 197750810-197750810 on Build GRCh38
rs994355456 Chr.3: 197750740-197750740 on Build GRCh38
rs1020099678 Chr.3: 197750997-197750997 on Build GRCh38
rs964001115 Chr.3: 197751050-197751050 on Build GRCh38
rs79784259 Chr.3: 197750975-197750975 on Build GRCh38
rs115802401 Chr.3: 197750676-197750676 on Build GRCh38
rs73894338 Chr.3: 197751030-197751030 on Build GRCh38
rs9883096 Chr.3: 197750580-197750580 on Build GRCh38
rs555410989 Chr.3: 197751073-197751073 on Build GRCh38
rs551360235 Chr.3: 197750965-197750965 on Build GRCh38
rs941334542 Chr.3: 197750834-197750834 on Build GRCh38
rs890844752 Chr.3: 197750718-197750718 on Build GRCh38
rs539467220 Chr.3: 197750604-197750604 on Build GRCh38
rs989098916 Chr.3: 197750655-197750655 on Build GRCh38
rs950041176 Chr.3: 197750626-197750626 on Build GRCh38
rs546191994 Chr.3: 197750788-197750788 on Build GRCh38
rs565801635 Chr.3: 197750936-197750936 on Build GRCh38
rs539024478 Chr.3: 197750940-197750940 on Build GRCh38
rs753829305 Chr.3: 197751020-197751020 on Build GRCh38
rs182060171 Chr.3: 197750648-197750648 on Build GRCh38
rs938539630 Chr.3: 197750650-197750650 on Build GRCh38
rs917336648 Chr.3: 197750582-197750582 on Build GRCh38
rs562256414 Chr.3: 197750894-197750894 on Build GRCh38
rs536800106 Chr.3: 197750627-197750627 on Build GRCh38
rs564581670 Chr.3: 197750796-197750796 on Build GRCh38
rs991973447 Chr.3: 197750783-197750783 on Build GRCh38
rs979844577 Chr.3: 197750647-197750647 on Build GRCh38
rs541906439 Chr.3: 197750741-197750741 on Build GRCh38
rs1035801867 Chr.3: 197750945-197750945 on Build GRCh38
rs984157594 Chr.3: 197750824-197750824 on Build GRCh38
rs189191450 Chr.3: 197750822-197750822 on Build GRCh38

View Full Product Details