Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73449143-73449652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFP
Entrez Gene ID
174
Gene Name
alpha fetoprotein
Gene Aliases
AFPD, FETA, HPAFP
UniGene
Hs.518808
Gene Chromosome Location
Chr.4: 73436219-73455785 on Build GRCh38
Transcripts
NM_001134.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4342876 c.1070A>G p.E357G Chr.4: 73449346-73449346 on Build GRCh38
6875565 c.1127G>A p.G376E Chr.4: 73449403-73449403 on Build GRCh38
5500605 c.1063G>C p.V355L Chr.4: 73449339-73449339 on Build GRCh38
5928549 c.1189G>A p.G397R Chr.4: 73449465-73449465 on Build GRCh38
5401514 c.1167C>T p.N389N Chr.4: 73449443-73449443 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936277322 Chr.4: 73449246-73449246 on Build GRCh38
rs937151428 Chr.4: 73449417-73449417 on Build GRCh38
rs926181494 Chr.4: 73449188-73449188 on Build GRCh38
rs986121502 Chr.4: 73449241-73449241 on Build GRCh38
rs954360723 Chr.4: 73449598-73449598 on Build GRCh38
rs957605394 Chr.4: 73449533-73449533 on Build GRCh38
rs774956431 Chr.4: 73449506-73449506 on Build GRCh38
rs777307383 Chr.4: 73449532-73449535 on Build GRCh38
rs943696006 Chr.4: 73449251-73449251 on Build GRCh38
rs775901894 Chr.4: 73449461-73449461 on Build GRCh38
rs927706059 Chr.4: 73449625-73449625 on Build GRCh38
rs767883317 Chr.4: 73449510-73449510 on Build GRCh38
rs937625225 Chr.4: 73449323-73449323 on Build GRCh38
rs746621443 Chr.4: 73449401-73449408 on Build GRCh38
rs35252463 Chr.4: 73449523-73449524 on Build GRCh38
rs1054808899 Chr.4: 73449334-73449334 on Build GRCh38
rs547434390 Chr.4: 73449256-73449256 on Build GRCh38
rs745967306 Chr.4: 73449437-73449437 on Build GRCh38
rs746089770 Chr.4: 73449472-73449472 on Build GRCh38
rs761426050 Chr.4: 73449418-73449418 on Build GRCh38
rs557363224 Chr.4: 73449200-73449200 on Build GRCh38
rs868732013 Chr.4: 73449432-73449432 on Build GRCh38
rs902641454 Chr.4: 73449537-73449537 on Build GRCh38
rs912783011 Chr.4: 73449247-73449247 on Build GRCh38
rs780662719 Chr.4: 73449300-73449300 on Build GRCh38
rs781503568 Chr.4: 73449458-73449458 on Build GRCh38
rs73827496 Chr.4: 73449302-73449302 on Build GRCh38
rs116657170 Chr.4: 73449168-73449168 on Build GRCh38
rs374624843 Chr.4: 73449308-73449308 on Build GRCh38
rs989057242 Chr.4: 73449560-73449560 on Build GRCh38
rs762119727 Chr.4: 73449507-73449507 on Build GRCh38
rs764510228 Chr.4: 73449408-73449408 on Build GRCh38
rs1033000186 Chr.4: 73449530-73449530 on Build GRCh38
rs759636523 Chr.4: 73449367-73449367 on Build GRCh38
rs770235744 Chr.4: 73449509-73449511 on Build GRCh38
rs552654353 Chr.4: 73449289-73449289 on Build GRCh38
rs749586751 Chr.4: 73449220-73449220 on Build GRCh38
rs753811251 Chr.4: 73449444-73449444 on Build GRCh38
rs765812136 Chr.4: 73449443-73449443 on Build GRCh38
rs763433563 Chr.4: 73449403-73449403 on Build GRCh38
rs376112674 Chr.4: 73449339-73449339 on Build GRCh38
rs569289389 Chr.4: 73449273-73449273 on Build GRCh38
rs749198913 Chr.4: 73449317-73449317 on Build GRCh38
rs747909610 Chr.4: 73449453-73449453 on Build GRCh38
rs979655616 Chr.4: 73449187-73449187 on Build GRCh38
rs772786892 Chr.4: 73449517-73449517 on Build GRCh38
rs755564892 Chr.4: 73449436-73449436 on Build GRCh38
rs1001358272 Chr.4: 73449594-73449594 on Build GRCh38
rs777023996 Chr.4: 73449357-73449357 on Build GRCh38
rs985406204 Chr.4: 73449515-73449515 on Build GRCh38
rs768431928 Chr.4: 73449332-73449332 on Build GRCh38
rs771280920 Chr.4: 73449374-73449374 on Build GRCh38
rs555037837 Chr.4: 73449423-73449423 on Build GRCh38
rs905689356 Chr.4: 73449394-73449394 on Build GRCh38
rs775995318 Chr.4: 73449398-73449398 on Build GRCh38
rs1032917972 Chr.4: 73449193-73449193 on Build GRCh38
rs769086953 Chr.4: 73449496-73449496 on Build GRCh38
rs373915432 Chr.4: 73449447-73449447 on Build GRCh38
rs774632101 Chr.4: 73449410-73449410 on Build GRCh38
rs758289615 Chr.4: 73449455-73449455 on Build GRCh38
rs34255749 Chr.4: 73449309-73449310 on Build GRCh38
rs540821622 Chr.4: 73449563-73449563 on Build GRCh38
rs913389755 Chr.4: 73449596-73449596 on Build GRCh38
rs200626144 Chr.4: 73449343-73449343 on Build GRCh38
rs950208315 Chr.4: 73449620-73449620 on Build GRCh38
rs761222590 Chr.4: 73449623-73449623 on Build GRCh38
rs779218170 Chr.4: 73449409-73449409 on Build GRCh38
rs1005493160 Chr.4: 73449362-73449362 on Build GRCh38
rs778438327 Chr.4: 73449283-73449283 on Build GRCh38
rs769986000 Chr.4: 73449478-73449478 on Build GRCh38
rs775486922 Chr.4: 73449486-73449486 on Build GRCh38
rs750100359 Chr.4: 73449433-73449433 on Build GRCh38
rs906275915 Chr.4: 73449305-73449305 on Build GRCh38
rs777608132 Chr.4: 73449203-73449203 on Build GRCh38
rs1015828029 Chr.4: 73449381-73449381 on Build GRCh38
rs749783288 Chr.4: 73449490-73449490 on Build GRCh38
rs778640868 Chr.4: 73449335-73449335 on Build GRCh38

View Full Product Details